Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 106924385-106924925 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DKK2
Entrez Gene ID
27123
Gene Name
dickkopf WNT signaling pathway inhibitor 2
Gene Aliases
DKK-2
UniGene
Hs.211869
Gene Chromosome Location
Chr.4: 106921802-107036296 on Build GRCh38
Transcripts
NM_014421.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
345579 c.456C>A p.H152Q Chr.4: 106924618-106924618 on Build GRCh38
7424897 c.429T>A p.D143E Chr.4: 106924645-106924645 on Build GRCh38
7417105 c.486G>A p.W162* Chr.4: 106924588-106924588 on Build GRCh38
149724 c.437G>A p.R146Q Chr.4: 106924637-106924637 on Build GRCh38
6098734 c.483A>T p.G161G Chr.4: 106924591-106924591 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs913992437 Chr.4: 106924484-106924484 on Build GRCh38
rs144369104 Chr.4: 106924659-106924659 on Build GRCh38
rs745406082 Chr.4: 106924584-106924584 on Build GRCh38
rs1032247062 Chr.4: 106924431-106924431 on Build GRCh38
rs964380997 Chr.4: 106924740-106924740 on Build GRCh38
rs958156862 Chr.4: 106924754-106924754 on Build GRCh38
rs17037102 Chr.4: 106924637-106924637 on Build GRCh38
rs763852133 Chr.4: 106924611-106924611 on Build GRCh38
rs549927928 Chr.4: 106924822-106924822 on Build GRCh38
rs372180616 Chr.4: 106924654-106924654 on Build GRCh38
rs755352007 Chr.4: 106924639-106924639 on Build GRCh38
rs913105323 Chr.4: 106924532-106924532 on Build GRCh38
rs982630431 Chr.4: 106924831-106924831 on Build GRCh38
rs543088091 Chr.4: 106924494-106924494 on Build GRCh38
rs200275573 Chr.4: 106924513-106924514 on Build GRCh38
rs573677337 Chr.4: 106924565-106924565 on Build GRCh38
rs773285393 Chr.4: 106924690-106924690 on Build GRCh38
rs761722065 Chr.4: 106924667-106924667 on Build GRCh38
rs779183411 Chr.4: 106924550-106924550 on Build GRCh38
rs913102327 Chr.4: 106924860-106924860 on Build GRCh38
rs746149718 Chr.4: 106924472-106924472 on Build GRCh38
rs745318292 Chr.4: 106924655-106924655 on Build GRCh38
rs752706600 Chr.4: 106924531-106924532 on Build GRCh38
rs184796669 Chr.4: 106924899-106924899 on Build GRCh38
rs577559877 Chr.4: 106924449-106924449 on Build GRCh38
rs771657399 Chr.4: 106924588-106924588 on Build GRCh38
rs773814398 Chr.4: 106924580-106924580 on Build GRCh38
rs781718513 Chr.4: 106924571-106924571 on Build GRCh38
rs770400633 Chr.4: 106924577-106924577 on Build GRCh38
rs558531820 Chr.4: 106924440-106924440 on Build GRCh38
rs767805994 Chr.4: 106924743-106924743 on Build GRCh38
rs368574320 Chr.4: 106924610-106924610 on Build GRCh38
rs143888191 Chr.4: 106924598-106924598 on Build GRCh38
rs761694951 Chr.4: 106924618-106924618 on Build GRCh38
rs753785515 Chr.4: 106924535-106924535 on Build GRCh38
rs774392569 Chr.4: 106924509-106924510 on Build GRCh38
rs987999458 Chr.4: 106924768-106924768 on Build GRCh38
rs1016325740 Chr.4: 106924736-106924736 on Build GRCh38
rs771201779 Chr.4: 106924506-106924509 on Build GRCh38
rs1047973276 Chr.4: 106924896-106924897 on Build GRCh38
rs750640591 Chr.4: 106924553-106924553 on Build GRCh38
rs890641921 Chr.4: 106924898-106924898 on Build GRCh38
rs775195730 Chr.4: 106924658-106924658 on Build GRCh38
rs771637130 Chr.4: 106924657-106924657 on Build GRCh38
rs757346025 Chr.4: 106924540-106924540 on Build GRCh38
rs879039026 Chr.4: 106924463-106924463 on Build GRCh38
rs938030403 Chr.4: 106924864-106924864 on Build GRCh38
rs920851227 Chr.4: 106924870-106924870 on Build GRCh38
rs767191866 Chr.4: 106924519-106924519 on Build GRCh38
rs756566924 Chr.4: 106924652-106924652 on Build GRCh38
rs34264321 Chr.4: 106924592-106924592 on Build GRCh38
rs111479771 Chr.4: 106924625-106924625 on Build GRCh38
rs748655485 Chr.4: 106924572-106924572 on Build GRCh38
rs34890926 Chr.4: 106924644-106924644 on Build GRCh38
rs115538369 Chr.4: 106924867-106924867 on Build GRCh38
rs763957261 Chr.4: 106924721-106924726 on Build GRCh38
rs988594831 Chr.4: 106924846-106924846 on Build GRCh38
rs779273379 Chr.4: 106924470-106924470 on Build GRCh38
rs974398047 Chr.4: 106924774-106924774 on Build GRCh38
rs984830722 Chr.4: 106924451-106924451 on Build GRCh38
rs759652252 Chr.4: 106924512-106924513 on Build GRCh38
rs1015965534 Chr.4: 106924466-106924466 on Build GRCh38
rs866243871 Chr.4: 106924427-106924427 on Build GRCh38
rs764026579 Chr.4: 106924527-106924527 on Build GRCh38
rs748580596 Chr.4: 106924646-106924646 on Build GRCh38
rs146683175 Chr.4: 106924662-106924662 on Build GRCh38
rs773975742 Chr.4: 106924514-106924514 on Build GRCh38
rs868430389 Chr.4: 106924593-106924593 on Build GRCh38
rs1021241212 Chr.4: 106924799-106924799 on Build GRCh38
rs777013395 Chr.4: 106924504-106924504 on Build GRCh38
rs565067628 Chr.4: 106924638-106924638 on Build GRCh38
rs376498901 Chr.4: 106924692-106924692 on Build GRCh38
rs764567781 Chr.4: 106924818-106924819 on Build GRCh38
rs760332429 Chr.4: 106924713-106924717 on Build GRCh38
rs372423907 Chr.4: 106924721-106924721 on Build GRCh38
rs539665202 Chr.4: 106924826-106924826 on Build GRCh38
rs1026546184 Chr.4: 106924447-106924447 on Build GRCh38
rs755119981 Chr.4: 106924722-106924722 on Build GRCh38
rs749456498 Chr.4: 106924522-106924522 on Build GRCh38
rs747361659 Chr.4: 106924497-106924497 on Build GRCh38
rs757495574 Chr.4: 106924617-106924617 on Build GRCh38
rs758651321 Chr.4: 106924558-106924558 on Build GRCh38
rs374743911 Chr.4: 106924560-106924560 on Build GRCh38
rs766423497 Chr.4: 106924699-106924699 on Build GRCh38
rs954468841 Chr.4: 106924805-106924805 on Build GRCh38
rs926698067 Chr.4: 106924850-106924850 on Build GRCh38
rs200137828 Chr.4: 106924513-106924513 on Build GRCh38
rs370396790 Chr.4: 106924509-106924509 on Build GRCh38
rs752061642 Chr.4: 106924521-106924521 on Build GRCh38
rs765206473 Chr.4: 106924686-106924686 on Build GRCh38
rs769155630 Chr.4: 106924501-106924501 on Build GRCh38
rs542381258 Chr.4: 106924569-106924569 on Build GRCh38
rs188609214 Chr.4: 106924525-106924525 on Build GRCh38
rs1029996707 Chr.4: 106924795-106924795 on Build GRCh38
rs753640771 Chr.4: 106924742-106924742 on Build GRCh38

View Full Product Details