Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 119030916-119031425 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SYNPO2
Entrez Gene ID
171024
Gene Name
synaptopodin 2
Gene Aliases
-
UniGene
Hs.655519
Gene Chromosome Location
Chr.4: 118850688-119061247 on Build GRCh38
Transcripts
NM_001128933.2 NM_001128934.2 NM_001286754.1 NM_001286755.1 NM_133477.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4169047 c.2578G>T p.G860* Chr.4: 119031353-119031353 on Build GRCh38
1050413 c.2469C>T p.N823N Chr.4: 119031244-119031244 on Build GRCh38
1594260 c.2470G>A p.V824M Chr.4: 119031245-119031245 on Build GRCh38
4479501 c.2330C>T p.S777F Chr.4: 119031105-119031105 on Build GRCh38
6166159 c.2275G>C p.V759L Chr.4: 119031050-119031050 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756886095 Chr.4: 119031308-119031308 on Build GRCh38
rs370135118 Chr.4: 119031141-119031141 on Build GRCh38
rs754962373 Chr.4: 119031225-119031225 on Build GRCh38
rs767049854 Chr.4: 119031320-119031320 on Build GRCh38
rs201894708 Chr.4: 119030945-119030945 on Build GRCh38
rs760829273 Chr.4: 119030944-119030944 on Build GRCh38
rs147039167 Chr.4: 119030985-119030985 on Build GRCh38
rs547181288 Chr.4: 119031019-119031019 on Build GRCh38
rs568868405 Chr.4: 119030977-119030977 on Build GRCh38
rs771131778 Chr.4: 119031272-119031272 on Build GRCh38
rs781028904 Chr.4: 119031162-119031162 on Build GRCh38
rs746022678 Chr.4: 119031019-119031021 on Build GRCh38
rs988764824 Chr.4: 119031309-119031309 on Build GRCh38
rs371041061 Chr.4: 119031172-119031172 on Build GRCh38
rs1049475650 Chr.4: 119031123-119031123 on Build GRCh38
rs201165841 Chr.4: 119031046-119031046 on Build GRCh38
rs966346835 Chr.4: 119031300-119031300 on Build GRCh38
rs374516196 Chr.4: 119031152-119031152 on Build GRCh38
rs562890418 Chr.4: 119031200-119031200 on Build GRCh38
rs768406595 Chr.4: 119031153-119031153 on Build GRCh38
rs777594117 Chr.4: 119030986-119030986 on Build GRCh38
rs753596181 Chr.4: 119030991-119030991 on Build GRCh38
rs745805028 Chr.4: 119031027-119031027 on Build GRCh38
rs557525605 Chr.4: 119031032-119031032 on Build GRCh38
rs761461634 Chr.4: 119031206-119031206 on Build GRCh38
rs138266651 Chr.4: 119031202-119031202 on Build GRCh38
rs369367759 Chr.4: 119030948-119030948 on Build GRCh38
rs749459875 Chr.4: 119031364-119031364 on Build GRCh38
rs759959851 Chr.4: 119031090-119031090 on Build GRCh38
rs753538835 Chr.4: 119031102-119031102 on Build GRCh38
rs896417952 Chr.4: 119031100-119031100 on Build GRCh38
rs746835891 Chr.4: 119031040-119031040 on Build GRCh38
rs751078933 Chr.4: 119031333-119031333 on Build GRCh38
rs756919263 Chr.4: 119031107-119031107 on Build GRCh38
rs772712106 Chr.4: 119031277-119031277 on Build GRCh38
rs75493275 Chr.4: 119031197-119031197 on Build GRCh38
rs545390527 Chr.4: 119031222-119031222 on Build GRCh38
rs34163579 Chr.4: 119031154-119031155 on Build GRCh38
rs537882356 Chr.4: 119031018-119031018 on Build GRCh38
rs976720630 Chr.4: 119031354-119031354 on Build GRCh38
rs761977430 Chr.4: 119031056-119031056 on Build GRCh38
rs771897583 Chr.4: 119030939-119030939 on Build GRCh38
rs540441313 Chr.4: 119031070-119031070 on Build GRCh38
rs867828048 Chr.4: 119031234-119031234 on Build GRCh38
rs772327047 Chr.4: 119031041-119031043 on Build GRCh38
rs777692059 Chr.4: 119031363-119031363 on Build GRCh38
rs771186840 Chr.4: 119031191-119031191 on Build GRCh38
rs373300504 Chr.4: 119030952-119030952 on Build GRCh38
rs760188668 Chr.4: 119031198-119031198 on Build GRCh38
rs745835000 Chr.4: 119030938-119030938 on Build GRCh38
rs763455307 Chr.4: 119031073-119031073 on Build GRCh38
rs758315263 Chr.4: 119031128-119031128 on Build GRCh38
rs755771437 Chr.4: 119031367-119031367 on Build GRCh38
rs768815773 Chr.4: 119031045-119031045 on Build GRCh38
rs775206514 Chr.4: 119030942-119030942 on Build GRCh38
rs762968549 Chr.4: 119031183-119031183 on Build GRCh38
rs763433488 Chr.4: 119030967-119030967 on Build GRCh38
rs149649606 Chr.4: 119031321-119031321 on Build GRCh38
rs763005134 Chr.4: 119030971-119030971 on Build GRCh38
rs747928984 Chr.4: 119031253-119031253 on Build GRCh38
rs768036976 Chr.4: 119031098-119031098 on Build GRCh38
rs745975778 Chr.4: 119031372-119031372 on Build GRCh38
rs756761123 Chr.4: 119030999-119030999 on Build GRCh38
rs760969009 Chr.4: 119031317-119031317 on Build GRCh38
rs533172135 Chr.4: 119031117-119031117 on Build GRCh38
rs751981895 Chr.4: 119030978-119030978 on Build GRCh38
rs751926863 Chr.4: 119031076-119031076 on Build GRCh38
rs779070957 Chr.4: 119031370-119031370 on Build GRCh38
rs141800282 Chr.4: 119031341-119031341 on Build GRCh38
rs374744931 Chr.4: 119031368-119031368 on Build GRCh38
rs749898945 Chr.4: 119031218-119031218 on Build GRCh38
rs767468007 Chr.4: 119031344-119031344 on Build GRCh38
rs749567616 Chr.4: 119031271-119031271 on Build GRCh38
rs750085659 Chr.4: 119031350-119031350 on Build GRCh38
rs573930944 Chr.4: 119031171-119031171 on Build GRCh38
rs141598717 Chr.4: 119030983-119030983 on Build GRCh38
rs762658094 Chr.4: 119031324-119031324 on Build GRCh38
rs769909024 Chr.4: 119031065-119031065 on Build GRCh38
rs578130565 Chr.4: 119031217-119031217 on Build GRCh38
rs777838583 Chr.4: 119031255-119031255 on Build GRCh38
rs746319591 Chr.4: 119031273-119031273 on Build GRCh38
rs773307460 Chr.4: 119031174-119031174 on Build GRCh38
rs1013908941 Chr.4: 119031139-119031139 on Build GRCh38
rs764827268 Chr.4: 119031212-119031212 on Build GRCh38
rs376703168 Chr.4: 119031223-119031223 on Build GRCh38
rs1040558660 Chr.4: 119030966-119030966 on Build GRCh38
rs1035713957 Chr.4: 119031283-119031283 on Build GRCh38
rs779756923 Chr.4: 119031037-119031037 on Build GRCh38
rs370623734 Chr.4: 119031330-119031330 on Build GRCh38
rs772657285 Chr.4: 119031378-119031378 on Build GRCh38
rs775883109 Chr.4: 119031310-119031310 on Build GRCh38
rs775836528 Chr.4: 119031393-119031393 on Build GRCh38
rs749906229 Chr.4: 119031122-119031122 on Build GRCh38
rs796928730 Chr.4: 119030979-119030979 on Build GRCh38
rs765279340 Chr.4: 119030962-119030962 on Build GRCh38
rs778428957 Chr.4: 119031120-119031120 on Build GRCh38
rs756314480 Chr.4: 119031254-119031254 on Build GRCh38
rs960137815 Chr.4: 119031287-119031287 on Build GRCh38
rs970546637 Chr.4: 119030968-119030968 on Build GRCh38
rs1015299966 Chr.4: 119031196-119031196 on Build GRCh38
rs775642547 Chr.4: 119031105-119031105 on Build GRCh38
rs751491761 Chr.4: 119031224-119031224 on Build GRCh38
rs747081179 Chr.4: 119031110-119031110 on Build GRCh38
rs565206988 Chr.4: 119031240-119031240 on Build GRCh38
rs138262112 Chr.4: 119031075-119031075 on Build GRCh38

View Full Product Details