Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 122827247-122827751 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGF2
Entrez Gene ID
2247
Gene Name
fibroblast growth factor 2
Gene Aliases
BFGF, FGF-2, FGFB, HBGF-2
UniGene
Hs.284244
Gene Chromosome Location
Chr.4: 122826708-122898235 on Build GRCh38
Transcripts
NM_002006.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4388139 c.530A>C p.H177P Chr.4: 122827305-122827305 on Build GRCh38
460698 c.477C>G p.F159L Chr.4: 122827252-122827252 on Build GRCh38
4613617 c.531delC p.D179fs*40 Chr.4: 122827306-122827306 on Build GRCh38
94308 c.529C>A p.H177N Chr.4: 122827304-122827304 on Build GRCh38
228405 c.485C>T p.P162L Chr.4: 122827260-122827260 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1042247517 Chr.4: 122827554-122827554 on Build GRCh38
rs113537066 Chr.4: 122827273-122827273 on Build GRCh38
rs746553829 Chr.4: 122827289-122827289 on Build GRCh38
rs542086639 Chr.4: 122827627-122827627 on Build GRCh38
rs556413775 Chr.4: 122827537-122827537 on Build GRCh38
rs368546149 Chr.4: 122827729-122827729 on Build GRCh38
rs1049108543 Chr.4: 122827585-122827585 on Build GRCh38
rs1025836377 Chr.4: 122827437-122827437 on Build GRCh38
rs779007855 Chr.4: 122827387-122827387 on Build GRCh38
rs761829108 Chr.4: 122827325-122827325 on Build GRCh38
rs3804144 Chr.4: 122827579-122827579 on Build GRCh38
rs1039401869 Chr.4: 122827622-122827622 on Build GRCh38
rs573090193 Chr.4: 122827597-122827597 on Build GRCh38
rs1019307071 Chr.4: 122827655-122827655 on Build GRCh38
rs1032697042 Chr.4: 122827314-122827314 on Build GRCh38
rs902336193 Chr.4: 122827547-122827547 on Build GRCh38
rs771842609 Chr.4: 122827313-122827313 on Build GRCh38
rs539693259 Chr.4: 122827574-122827574 on Build GRCh38
rs959219115 Chr.4: 122827701-122827701 on Build GRCh38
rs991898864 Chr.4: 122827480-122827480 on Build GRCh38
rs566071779 Chr.4: 122827418-122827418 on Build GRCh38
rs910161661 Chr.4: 122827470-122827470 on Build GRCh38
rs183060480 Chr.4: 122827318-122827318 on Build GRCh38
rs761028222 Chr.4: 122827319-122827319 on Build GRCh38
rs199668081 Chr.4: 122827367-122827367 on Build GRCh38
rs759255044 Chr.4: 122827285-122827285 on Build GRCh38
rs893753868 Chr.4: 122827594-122827594 on Build GRCh38
rs2922979 Chr.4: 122827461-122827461 on Build GRCh38
rs965348856 Chr.4: 122827405-122827405 on Build GRCh38
rs1006174514 Chr.4: 122827688-122827688 on Build GRCh38
rs1025860068 Chr.4: 122827673-122827673 on Build GRCh38
rs1006523772 Chr.4: 122827619-122827619 on Build GRCh38
rs904705723 Chr.4: 122827558-122827558 on Build GRCh38
rs1017150534 Chr.4: 122827697-122827697 on Build GRCh38
rs866587151 Chr.4: 122827656-122827656 on Build GRCh38
rs374332071 Chr.4: 122827309-122827309 on Build GRCh38
rs991418757 Chr.4: 122827707-122827707 on Build GRCh38
rs776742713 Chr.4: 122827324-122827324 on Build GRCh38
rs45613534 Chr.4: 122827377-122827377 on Build GRCh38
rs745880627 Chr.4: 122827402-122827402 on Build GRCh38
rs114399263 Chr.4: 122827515-122827515 on Build GRCh38
rs1053701650 Chr.4: 122827581-122827581 on Build GRCh38
rs936364529 Chr.4: 122827570-122827570 on Build GRCh38
rs1007001386 Chr.4: 122827336-122827336 on Build GRCh38
rs557453076 Chr.4: 122827514-122827514 on Build GRCh38
rs893459659 Chr.4: 122827332-122827332 on Build GRCh38
rs900900310 Chr.4: 122827636-122827636 on Build GRCh38
rs41333248 Chr.4: 122827369-122827369 on Build GRCh38
rs2922980 Chr.4: 122827639-122827639 on Build GRCh38
rs567568819 Chr.4: 122827478-122827478 on Build GRCh38
rs901057762 Chr.4: 122827662-122827662 on Build GRCh38
rs765761551 Chr.4: 122827328-122827328 on Build GRCh38
rs889581534 Chr.4: 122827582-122827582 on Build GRCh38
rs370687386 Chr.4: 122827366-122827366 on Build GRCh38
rs917296146 Chr.4: 122827495-122827495 on Build GRCh38
rs764272274 Chr.4: 122827320-122827320 on Build GRCh38
rs983953095 Chr.4: 122827720-122827720 on Build GRCh38
rs771017160 Chr.4: 122827267-122827267 on Build GRCh38
rs148965201 Chr.4: 122827667-122827667 on Build GRCh38
rs984173741 Chr.4: 122827464-122827464 on Build GRCh38
rs370928166 Chr.4: 122827370-122827370 on Build GRCh38
rs186985577 Chr.4: 122827460-122827460 on Build GRCh38
rs532522277 Chr.4: 122827376-122827376 on Build GRCh38
rs535310601 Chr.4: 122827459-122827459 on Build GRCh38
rs951573208 Chr.4: 122827680-122827680 on Build GRCh38
rs377621586 Chr.4: 122827362-122827362 on Build GRCh38
rs373341357 Chr.4: 122827409-122827409 on Build GRCh38

View Full Product Details