Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 155796291-155796788 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GUCY1B3
Entrez Gene ID
2983
Gene Name
guanylate cyclase 1 soluble subunit beta
Gene Aliases
GC-S-beta-1, GC-SB3, GUC1B3, GUCB3, GUCSB3, GUCY1B1
UniGene
Hs.77890
Gene Chromosome Location
Chr.4: 155758973-155807642 on Build GRCh38
Transcripts
NM_000857.3 NM_001291951.1 NM_001291952.1 NM_001291953.1 NM_001291954.1 NM_001291955.1 XM_011531901.1 XM_017008130.1 XM_017008131.1 XM_017008132.1 XM_017008133.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4514144 c.964C>T p.L322L Chr.4: 155796497-155796497 on Build GRCh38
6261220 c.871G>T p.E291* Chr.4: 155796404-155796404 on Build GRCh38
4425248 c.964C>G p.L322V Chr.4: 155796497-155796497 on Build GRCh38
4560982 c.864G>C p.E288D Chr.4: 155796397-155796397 on Build GRCh38
4441772 c.860T>C p.V287A Chr.4: 155796393-155796393 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1003873831 Chr.4: 155796676-155796676 on Build GRCh38
rs368471905 Chr.4: 155796522-155796523 on Build GRCh38
rs906336040 Chr.4: 155796655-155796655 on Build GRCh38
rs199558345 Chr.4: 155796428-155796428 on Build GRCh38
rs757133016 Chr.4: 155796468-155796468 on Build GRCh38
rs946133938 Chr.4: 155796601-155796601 on Build GRCh38
rs201309079 Chr.4: 155796391-155796391 on Build GRCh38
rs747517071 Chr.4: 155796330-155796330 on Build GRCh38
rs762495088 Chr.4: 155796722-155796722 on Build GRCh38
rs986653224 Chr.4: 155796549-155796549 on Build GRCh38
rs942602561 Chr.4: 155796617-155796617 on Build GRCh38
rs763136532 Chr.4: 155796539-155796539 on Build GRCh38
rs2229201 Chr.4: 155796472-155796472 on Build GRCh38
rs904484927 Chr.4: 155796652-155796652 on Build GRCh38
rs764971221 Chr.4: 155796516-155796516 on Build GRCh38
rs773218587 Chr.4: 155796499-155796499 on Build GRCh38
rs994534531 Chr.4: 155796734-155796734 on Build GRCh38
rs868775613 Chr.4: 155796380-155796380 on Build GRCh38
rs58885710 Chr.4: 155796709-155796709 on Build GRCh38
rs200359355 Chr.4: 155796421-155796421 on Build GRCh38
rs773442560 Chr.4: 155796375-155796375 on Build GRCh38
rs185629730 Chr.4: 155796536-155796536 on Build GRCh38
rs1033357682 Chr.4: 155796397-155796397 on Build GRCh38
rs763146891 Chr.4: 155796475-155796476 on Build GRCh38
rs201359483 Chr.4: 155796434-155796434 on Build GRCh38
rs913348093 Chr.4: 155796553-155796553 on Build GRCh38
rs1003438128 Chr.4: 155796344-155796344 on Build GRCh38
rs762977665 Chr.4: 155796407-155796407 on Build GRCh38
rs1055086754 Chr.4: 155796704-155796704 on Build GRCh38
rs1053127066 Chr.4: 155796662-155796662 on Build GRCh38
rs762166734 Chr.4: 155796343-155796343 on Build GRCh38
rs777047364 Chr.4: 155796338-155796338 on Build GRCh38
rs372089795 Chr.4: 155796498-155796498 on Build GRCh38
rs773395367 Chr.4: 155796538-155796538 on Build GRCh38
rs538361404 Chr.4: 155796543-155796543 on Build GRCh38
rs556596016 Chr.4: 155796327-155796327 on Build GRCh38
rs779488048 Chr.4: 155796486-155796486 on Build GRCh38
rs770055381 Chr.4: 155796464-155796465 on Build GRCh38
rs747546342 Chr.4: 155796490-155796490 on Build GRCh38
rs536997873 Chr.4: 155796414-155796414 on Build GRCh38
rs1027307569 Chr.4: 155796761-155796761 on Build GRCh38
rs766428766 Chr.4: 155796477-155796478 on Build GRCh38
rs370164801 Chr.4: 155796451-155796451 on Build GRCh38
rs762333446 Chr.4: 155796508-155796508 on Build GRCh38
rs571381412 Chr.4: 155796711-155796711 on Build GRCh38
rs376552151 Chr.4: 155796349-155796349 on Build GRCh38
rs773474777 Chr.4: 155796467-155796470 on Build GRCh38
rs556610219 Chr.4: 155796552-155796552 on Build GRCh38
rs768974193 Chr.4: 155796494-155796494 on Build GRCh38
rs763949175 Chr.4: 155796447-155796447 on Build GRCh38
rs1016571754 Chr.4: 155796460-155796460 on Build GRCh38
rs535320953 Chr.4: 155796448-155796448 on Build GRCh38
rs1042631728 Chr.4: 155796622-155796622 on Build GRCh38
rs769051561 Chr.4: 155796334-155796334 on Build GRCh38
rs778660514 Chr.4: 155796471-155796471 on Build GRCh38
rs1020773503 Chr.4: 155796316-155796316 on Build GRCh38
rs757900619 Chr.4: 155796485-155796485 on Build GRCh38
rs1044041404 Chr.4: 155796629-155796629 on Build GRCh38
rs748460677 Chr.4: 155796532-155796532 on Build GRCh38
rs1024406800 Chr.4: 155796721-155796721 on Build GRCh38

View Full Product Details