Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 163350155-163350653 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NPY5R
Entrez Gene ID
4889
Gene Name
neuropeptide Y receptor Y5
Gene Aliases
NPY5-R, NPYR5, NPYY5-R
UniGene
Hs.519058
Gene Chromosome Location
Chr.4: 163336968-163352277 on Build GRCh38
Transcripts
NM_001317091.1 NM_001317092.1 NM_006174.3 XM_005263038.3 XM_011532015.2 XM_011532017.2 XM_017008255.1 XM_017008256.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1309864 c.284C>T p.S95L Chr.4: 163350557-163350557 on Build GRCh38
5640774 c.239T>C p.I80T Chr.4: 163350512-163350512 on Build GRCh38
6166636 c.63C>A p.A21A Chr.4: 163350336-163350336 on Build GRCh38
234278 c.320G>A p.W107* Chr.4: 163350593-163350593 on Build GRCh38
6099507 c.151G>T p.V51L Chr.4: 163350424-163350424 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764418498 Chr.4: 163350224-163350225 on Build GRCh38
rs1004379174 Chr.4: 163350190-163350190 on Build GRCh38
rs752819658 Chr.4: 163350515-163350515 on Build GRCh38
rs80086880 Chr.4: 163350482-163350482 on Build GRCh38
rs765566769 Chr.4: 163350601-163350601 on Build GRCh38
rs751738838 Chr.4: 163350377-163350377 on Build GRCh38
rs3967767 Chr.4: 163350385-163350385 on Build GRCh38
rs775716946 Chr.4: 163350617-163350617 on Build GRCh38
rs765809370 Chr.4: 163350357-163350357 on Build GRCh38
rs75269811 Chr.4: 163350303-163350303 on Build GRCh38
rs536920158 Chr.4: 163350540-163350540 on Build GRCh38
rs537633153 Chr.4: 163350442-163350442 on Build GRCh38
rs777746508 Chr.4: 163350340-163350340 on Build GRCh38
rs371917653 Chr.4: 163350291-163350291 on Build GRCh38
rs200912302 Chr.4: 163350374-163350374 on Build GRCh38
rs1017040292 Chr.4: 163350202-163350202 on Build GRCh38
rs963890058 Chr.4: 163350623-163350623 on Build GRCh38
rs780729323 Chr.4: 163350561-163350561 on Build GRCh38
rs375564689 Chr.4: 163350473-163350473 on Build GRCh38
rs78060816 Chr.4: 163350271-163350271 on Build GRCh38
rs867560945 Chr.4: 163350474-163350474 on Build GRCh38
rs776148718 Chr.4: 163350331-163350331 on Build GRCh38
rs745630054 Chr.4: 163350313-163350313 on Build GRCh38
rs201405862 Chr.4: 163350546-163350546 on Build GRCh38
rs779770681 Chr.4: 163350537-163350537 on Build GRCh38
rs1042954323 Chr.4: 163350381-163350381 on Build GRCh38
rs571795802 Chr.4: 163350409-163350409 on Build GRCh38
rs774621918 Chr.4: 163350583-163350583 on Build GRCh38
rs758155411 Chr.4: 163350453-163350453 on Build GRCh38
rs747764021 Chr.4: 163350572-163350572 on Build GRCh38
rs765708962 Chr.4: 163350261-163350261 on Build GRCh38
rs769434625 Chr.4: 163350577-163350577 on Build GRCh38
rs1001289694 Chr.4: 163350414-163350414 on Build GRCh38
rs763606723 Chr.4: 163350237-163350237 on Build GRCh38
rs757149914 Chr.4: 163350612-163350612 on Build GRCh38
rs147087846 Chr.4: 163350484-163350484 on Build GRCh38
rs921335137 Chr.4: 163350239-163350239 on Build GRCh38
rs776885611 Chr.4: 163350437-163350438 on Build GRCh38
rs768056385 Chr.4: 163350329-163350329 on Build GRCh38
rs913841290 Chr.4: 163350284-163350284 on Build GRCh38
rs201640301 Chr.4: 163350314-163350314 on Build GRCh38
rs749822591 Chr.4: 163350582-163350582 on Build GRCh38
rs779831413 Chr.4: 163350458-163350458 on Build GRCh38
rs773762046 Chr.4: 163350369-163350369 on Build GRCh38
rs759920239 Chr.4: 163350592-163350592 on Build GRCh38
rs945935121 Chr.4: 163350345-163350345 on Build GRCh38
rs78861810 Chr.4: 163350304-163350304 on Build GRCh38
rs933010286 Chr.4: 163350217-163350217 on Build GRCh38
rs747640202 Chr.4: 163350481-163350481 on Build GRCh38
rs79555174 Chr.4: 163350438-163350438 on Build GRCh38
rs757222561 Chr.4: 163350424-163350424 on Build GRCh38
rs962869909 Chr.4: 163350547-163350547 on Build GRCh38
rs764114710 Chr.4: 163350519-163350519 on Build GRCh38
rs79962221 Chr.4: 163350575-163350575 on Build GRCh38
rs761905399 Chr.4: 163350554-163350554 on Build GRCh38
rs996930289 Chr.4: 163350586-163350586 on Build GRCh38
rs781270157 Chr.4: 163350300-163350300 on Build GRCh38
rs767555011 Chr.4: 163350407-163350407 on Build GRCh38
rs538370021 Chr.4: 163350267-163350267 on Build GRCh38
rs772794090 Chr.4: 163350578-163350578 on Build GRCh38
rs975481562 Chr.4: 163350226-163350226 on Build GRCh38
rs767751082 Chr.4: 163350512-163350512 on Build GRCh38
rs962798022 Chr.4: 163350223-163350223 on Build GRCh38
rs79693996 Chr.4: 163350601-163350601 on Build GRCh38
rs750361929 Chr.4: 163350522-163350522 on Build GRCh38
rs1024920404 Chr.4: 163350517-163350517 on Build GRCh38
rs772273448 Chr.4: 163350603-163350603 on Build GRCh38
rs759746938 Chr.4: 163350511-163350511 on Build GRCh38
rs755044465 Chr.4: 163350294-163350294 on Build GRCh38
rs866991067 Chr.4: 163350447-163350447 on Build GRCh38
rs552707275 Chr.4: 163350497-163350497 on Build GRCh38
rs758297007 Chr.4: 163350533-163350533 on Build GRCh38
rs566434611 Chr.4: 163350265-163350265 on Build GRCh38
rs574392131 Chr.4: 163350518-163350518 on Build GRCh38
rs778748928 Chr.4: 163350309-163350309 on Build GRCh38
rs375176738 Chr.4: 163350341-163350341 on Build GRCh38
rs768340023 Chr.4: 163350471-163350471 on Build GRCh38
rs372200526 Chr.4: 163350469-163350469 on Build GRCh38
rs951021955 Chr.4: 163350520-163350520 on Build GRCh38
rs754332295 Chr.4: 163350241-163350241 on Build GRCh38
rs760331232 Chr.4: 163350328-163350328 on Build GRCh38
rs762343362 Chr.4: 163350248-163350248 on Build GRCh38
rs774768573 Chr.4: 163350498-163350498 on Build GRCh38
rs780119025 Chr.4: 163350289-163350289 on Build GRCh38
rs755174120 Chr.4: 163350386-163350386 on Build GRCh38
rs775024515 Chr.4: 163350315-163350315 on Build GRCh38

View Full Product Details