Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 165079544-165080047 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM192
Entrez Gene ID
201931
Gene Name
transmembrane protein 192
Gene Aliases
-
UniGene
Hs.704511
Gene Chromosome Location
Chr.4: 165076078-165112913 on Build GRCh38
Transcripts
NM_001100389.1 XM_011531717.2 XM_011531718.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3993533 c.691C>A p.L231I Chr.4: 165079783-165079783 on Build GRCh38
4868021 c.719A>G p.Y240C Chr.4: 165079743-165079743 on Build GRCh38
4123295 c.750G>A p.A250A Chr.4: 165079724-165079724 on Build GRCh38
5991296 c.779T>A p.L260Q Chr.4: 165079683-165079683 on Build GRCh38
5991295 c.791T>A p.L264Q Chr.4: 165079683-165079683 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758592980 Chr.4: 165079730-165079730 on Build GRCh38
rs201314834 Chr.4: 165079756-165079756 on Build GRCh38
rs980025441 Chr.4: 165079609-165079609 on Build GRCh38
rs922103884 Chr.4: 165079783-165079783 on Build GRCh38
rs749142352 Chr.4: 165079699-165079699 on Build GRCh38
rs879468477 Chr.4: 165079654-165079654 on Build GRCh38
rs374733693 Chr.4: 165079847-165079847 on Build GRCh38
rs200586876 Chr.4: 165079743-165079743 on Build GRCh38
rs761286842 Chr.4: 165079643-165079643 on Build GRCh38
rs200755080 Chr.4: 165079786-165079786 on Build GRCh38
rs765550331 Chr.4: 165079726-165079726 on Build GRCh38
rs201065650 Chr.4: 165079669-165079669 on Build GRCh38
rs763829640 Chr.4: 165079711-165079711 on Build GRCh38
rs766564993 Chr.4: 165079834-165079834 on Build GRCh38
rs556977126 Chr.4: 165079710-165079710 on Build GRCh38
rs200266889 Chr.4: 165079808-165079808 on Build GRCh38
rs766944244 Chr.4: 165079638-165079638 on Build GRCh38
rs750421987 Chr.4: 165079653-165079655 on Build GRCh38
rs1013668958 Chr.4: 165079946-165079946 on Build GRCh38
rs533933004 Chr.4: 165079904-165079904 on Build GRCh38
rs771816502 Chr.4: 165079813-165079813 on Build GRCh38
rs774914155 Chr.4: 165079719-165079719 on Build GRCh38
rs77771595 Chr.4: 165079998-165079998 on Build GRCh38
rs754295931 Chr.4: 165079649-165079649 on Build GRCh38
rs1004205882 Chr.4: 165079856-165079856 on Build GRCh38
rs896513611 Chr.4: 165079941-165079941 on Build GRCh38
rs764677479 Chr.4: 165079648-165079648 on Build GRCh38
rs374995608 Chr.4: 165079725-165079725 on Build GRCh38
rs151083192 Chr.4: 165079614-165079614 on Build GRCh38
rs540435585 Chr.4: 165080021-165080022 on Build GRCh38
rs773484601 Chr.4: 165079826-165079826 on Build GRCh38
rs756083102 Chr.4: 165079684-165079684 on Build GRCh38
rs771664264 Chr.4: 165079781-165079781 on Build GRCh38
rs372603967 Chr.4: 165080022-165080023 on Build GRCh38
rs114703118 Chr.4: 165079934-165079934 on Build GRCh38
rs888046939 Chr.4: 165079822-165079822 on Build GRCh38
rs755040059 Chr.4: 165079735-165079735 on Build GRCh38
rs770309404 Chr.4: 165079821-165079821 on Build GRCh38
rs528707594 Chr.4: 165079734-165079734 on Build GRCh38
rs758387411 Chr.4: 165079772-165079772 on Build GRCh38
rs766590037 Chr.4: 165079731-165079731 on Build GRCh38
rs953253565 Chr.4: 165079599-165079599 on Build GRCh38
rs925890331 Chr.4: 165079637-165079637 on Build GRCh38
rs367897333 Chr.4: 165079763-165079763 on Build GRCh38
rs763369586 Chr.4: 165079827-165079827 on Build GRCh38
rs745475104 Chr.4: 165079705-165079705 on Build GRCh38
rs1024134530 Chr.4: 165079580-165079580 on Build GRCh38
rs759174436 Chr.4: 165079628-165079628 on Build GRCh38
rs145157032 Chr.4: 165079709-165079709 on Build GRCh38
rs760233507 Chr.4: 165079723-165079723 on Build GRCh38
rs778596030 Chr.4: 165079758-165079758 on Build GRCh38
rs1053910124 Chr.4: 165079777-165079777 on Build GRCh38
rs750747805 Chr.4: 165079727-165079727 on Build GRCh38
rs770727784 Chr.4: 165079700-165079700 on Build GRCh38
rs897518032 Chr.4: 165079929-165079929 on Build GRCh38
rs757680935 Chr.4: 165079650-165079650 on Build GRCh38
rs750614559 Chr.4: 165079661-165079661 on Build GRCh38
rs1035993512 Chr.4: 165079863-165079863 on Build GRCh38
rs141459493 Chr.4: 165079659-165079659 on Build GRCh38
rs368324061 Chr.4: 165079677-165079677 on Build GRCh38
rs34977148 Chr.4: 165079576-165079577 on Build GRCh38
rs576156835 Chr.4: 165079589-165079589 on Build GRCh38
rs751796966 Chr.4: 165079844-165079844 on Build GRCh38
rs564490422 Chr.4: 165079724-165079724 on Build GRCh38
rs777680484 Chr.4: 165079695-165079695 on Build GRCh38
rs781193015 Chr.4: 165079740-165079740 on Build GRCh38
rs775026124 Chr.4: 165079640-165079640 on Build GRCh38
rs751159387 Chr.4: 165079846-165079846 on Build GRCh38
rs758606457 Chr.4: 165079668-165079668 on Build GRCh38
rs906760586 Chr.4: 165079849-165079849 on Build GRCh38
rs768284329 Chr.4: 165079809-165079809 on Build GRCh38
rs569695093 Chr.4: 165079972-165079972 on Build GRCh38
rs1007319523 Chr.4: 165080009-165080009 on Build GRCh38
rs373709632 Chr.4: 165079811-165079811 on Build GRCh38

View Full Product Details