Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 168915917-168916462 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CBR4
Entrez Gene ID
84869
Gene Name
carbonyl reductase 4
Gene Aliases
SDR45C1
UniGene
Hs.659311
Gene Chromosome Location
Chr.4: 168894484-169010317 on Build GRCh38
Transcripts
XM_005263315.2 XM_017008782.1 NM_001166108.1 NM_001166109.1 NM_001166110.1 NM_016081.3 XM_005262861.4 XM_005262866.2 XM_011531768.2 XM_011531769.2 XM_011531770.2 XM_011531771.2 XM_011531772.2 XM_011531773.1 XM_011531774.1 XM_011531775.1 XM_017007910.1

Gene Symbol
PALLD
Entrez Gene ID
23022
Gene Name
palladin, cytoskeletal associated protein
Gene Aliases
CGI-151, CGI151, MYN, PNCA1, SIH002
UniGene
Hs.151220
Gene Chromosome Location
Chr.4: 168497039-168928457 on Build GRCh38
Transcripts
XM_005263315.2 XM_017008782.1 NM_001166108.1 NM_001166109.1 NM_001166110.1 NM_016081.3 XM_005262861.4 XM_005262866.2 XM_011531768.2 XM_011531769.2 XM_011531770.2 XM_011531771.2 XM_011531772.2 XM_011531773.1 XM_011531774.1 XM_011531775.1 XM_017007910.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6203483 c.2215A>C p.S739R Chr.4: 168915917-168915917 on Build GRCh38
3601982 c.2224G>A p.E742K Chr.4: 168915926-168915926 on Build GRCh38
4853181 c.2758C>G p.L920V Chr.4: 168915986-168915986 on Build GRCh38
6737076 c.2719C>T p.R907* Chr.4: 168915947-168915947 on Build GRCh38
6099933 c.2260C>T p.H754Y Chr.4: 168915962-168915962 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773447112 Chr.4: 168916064-168916064 on Build GRCh38
rs745655806 Chr.4: 168916031-168916031 on Build GRCh38
rs778696691 Chr.4: 168916106-168916107 on Build GRCh38
rs777454544 Chr.4: 168916049-168916054 on Build GRCh38
rs755556663 Chr.4: 168915974-168915974 on Build GRCh38
rs897773656 Chr.4: 168916176-168916176 on Build GRCh38
rs774232784 Chr.4: 168916224-168916225 on Build GRCh38
rs986221733 Chr.4: 168916057-168916057 on Build GRCh38
rs967680431 Chr.4: 168916298-168916298 on Build GRCh38
rs771821579 Chr.4: 168916034-168916034 on Build GRCh38
rs756261237 Chr.4: 168915979-168915979 on Build GRCh38
rs370652817 Chr.4: 168915967-168915967 on Build GRCh38
rs886242319 Chr.4: 168916192-168916192 on Build GRCh38
rs935514520 Chr.4: 168916293-168916293 on Build GRCh38
rs541659949 Chr.4: 168916359-168916359 on Build GRCh38
rs780035680 Chr.4: 168916038-168916038 on Build GRCh38
rs563135659 Chr.4: 168916381-168916381 on Build GRCh38
rs756934356 Chr.4: 168915978-168915978 on Build GRCh38
rs960732231 Chr.4: 168916254-168916254 on Build GRCh38
rs763223193 Chr.4: 168916068-168916068 on Build GRCh38
rs1001928773 Chr.4: 168916264-168916264 on Build GRCh38
rs7693134 Chr.4: 168916438-168916438 on Build GRCh38
rs113605627 Chr.4: 168916268-168916268 on Build GRCh38
rs900452373 Chr.4: 168916114-168916114 on Build GRCh38
rs769936930 Chr.4: 168916062-168916062 on Build GRCh38
rs986948932 Chr.4: 168916311-168916311 on Build GRCh38
rs994681166 Chr.4: 168916150-168916150 on Build GRCh38
rs867904654 Chr.4: 168916382-168916382 on Build GRCh38
rs755336512 Chr.4: 168915948-168915948 on Build GRCh38
rs968331227 Chr.4: 168916272-168916272 on Build GRCh38
rs941881301 Chr.4: 168916108-168916108 on Build GRCh38
rs376808728 Chr.4: 168916055-168916055 on Build GRCh38
rs986211779 Chr.4: 168916424-168916424 on Build GRCh38
rs780140461 Chr.4: 168916069-168916069 on Build GRCh38
rs906207077 Chr.4: 168916248-168916248 on Build GRCh38
rs978095414 Chr.4: 168916313-168916313 on Build GRCh38
rs924119137 Chr.4: 168916280-168916280 on Build GRCh38
rs1034754097 Chr.4: 168916199-168916199 on Build GRCh38
rs954876447 Chr.4: 168916401-168916401 on Build GRCh38
rs960430324 Chr.4: 168915941-168915941 on Build GRCh38
rs1039717043 Chr.4: 168916436-168916436 on Build GRCh38
rs990533767 Chr.4: 168916267-168916267 on Build GRCh38
rs977479187 Chr.4: 168916042-168916042 on Build GRCh38
rs1043925326 Chr.4: 168916002-168916002 on Build GRCh38
rs1037613400 Chr.4: 168916148-168916148 on Build GRCh38
rs912688255 Chr.4: 168916350-168916350 on Build GRCh38
rs146787957 Chr.4: 168915976-168915976 on Build GRCh38
rs551597541 Chr.4: 168916281-168916281 on Build GRCh38
rs540983021 Chr.4: 168916200-168916200 on Build GRCh38
rs769167363 Chr.4: 168916183-168916183 on Build GRCh38
rs114250766 Chr.4: 168915961-168915961 on Build GRCh38
rs1004657860 Chr.4: 168916195-168916195 on Build GRCh38
rs576733987 Chr.4: 168916104-168916104 on Build GRCh38
rs1027450113 Chr.4: 168916185-168916185 on Build GRCh38
rs942773268 Chr.4: 168916407-168916407 on Build GRCh38
rs543534073 Chr.4: 168916091-168916091 on Build GRCh38
rs185765257 Chr.4: 168916093-168916093 on Build GRCh38
rs778471055 Chr.4: 168915992-168915992 on Build GRCh38
rs910672276 Chr.4: 168916434-168916434 on Build GRCh38
rs935421039 Chr.4: 168916048-168916048 on Build GRCh38
rs189876363 Chr.4: 168916273-168916273 on Build GRCh38

View Full Product Details