Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 174936821-174937339 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAM29
Entrez Gene ID
11086
Gene Name
ADAM metallopeptidase domain 29
Gene Aliases
CT73, svph1
UniGene
Hs.126838
Gene Chromosome Location
Chr.4: 174918358-174978180 on Build GRCh38
Transcripts
NM_001130703.1 NM_001130704.1 NM_001130705.1 NM_001278125.1 NM_001278126.1 NM_001278127.1 NM_014269.4 XM_011531556.1 XM_011531557.1 XM_011531559.1 XM_011531560.1 XM_011531561.1

SNP Detail

SNP ID Chromosome Location
rs189292358 Chr.4: 174937098-174937098 on Build GRCh38
rs1000609446 Chr.4: 174937291-174937291 on Build GRCh38
rs79106343 Chr.4: 174937290-174937290 on Build GRCh38
rs753344853 Chr.4: 174936853-174936853 on Build GRCh38
rs13144382 Chr.4: 174936878-174936878 on Build GRCh38
rs758264183 Chr.4: 174937259-174937259 on Build GRCh38
rs560883690 Chr.4: 174937066-174937066 on Build GRCh38
rs62334421 Chr.4: 174936846-174936846 on Build GRCh38
rs116434596 Chr.4: 174937269-174937269 on Build GRCh38
rs916697657 Chr.4: 174937294-174937294 on Build GRCh38
rs149199514 Chr.4: 174937309-174937309 on Build GRCh38
rs1016777599 Chr.4: 174937278-174937278 on Build GRCh38
rs1028020844 Chr.4: 174937296-174937296 on Build GRCh38
rs145239713 Chr.4: 174937049-174937049 on Build GRCh38
rs912772637 Chr.4: 174937096-174937096 on Build GRCh38
rs764187005 Chr.4: 174936988-174936989 on Build GRCh38
rs979136285 Chr.4: 174936859-174936859 on Build GRCh38
rs570885135 Chr.4: 174937129-174937129 on Build GRCh38
rs765229147 Chr.4: 174937097-174937097 on Build GRCh38
rs951364240 Chr.4: 174936922-174936922 on Build GRCh38
rs893930553 Chr.4: 174937099-174937099 on Build GRCh38
rs533379669 Chr.4: 174937015-174937015 on Build GRCh38
rs899573717 Chr.4: 174937171-174937171 on Build GRCh38
rs73001898 Chr.4: 174937264-174937264 on Build GRCh38
rs551520299 Chr.4: 174937298-174937298 on Build GRCh38
rs909684518 Chr.4: 174937028-174937028 on Build GRCh38
rs954219845 Chr.4: 174937300-174937300 on Build GRCh38
rs1029725904 Chr.4: 174937241-174937241 on Build GRCh38
rs1056384868 Chr.4: 174937237-174937237 on Build GRCh38
rs937811008 Chr.4: 174937233-174937233 on Build GRCh38
rs948094475 Chr.4: 174937087-174937087 on Build GRCh38
rs1018074124 Chr.4: 174937140-174937140 on Build GRCh38
rs905064089 Chr.4: 174937218-174937218 on Build GRCh38
rs997881717 Chr.4: 174937180-174937180 on Build GRCh38
rs954208060 Chr.4: 174936956-174936956 on Build GRCh38
rs991279948 Chr.4: 174937293-174937293 on Build GRCh38
rs931410120 Chr.4: 174937119-174937119 on Build GRCh38
rs369677071 Chr.4: 174937009-174937009 on Build GRCh38
rs186357130 Chr.4: 174936955-174936955 on Build GRCh38
rs560359514 Chr.4: 174937267-174937267 on Build GRCh38
rs867745358 Chr.4: 174937250-174937250 on Build GRCh38
rs916584503 Chr.4: 174937078-174937078 on Build GRCh38
rs919913023 Chr.4: 174937113-174937113 on Build GRCh38
rs1044498419 Chr.4: 174937198-174937198 on Build GRCh38

View Full Product Details