Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 459270-459771 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXOC3
Entrez Gene ID
11336
Gene Name
exocyst complex component 3
Gene Aliases
SEC6, SEC6L1, Sec6p
UniGene
Hs.481464
Gene Chromosome Location
Chr.5: 443184-471938 on Build GRCh38
Transcripts
NM_007277.4 XM_011513948.2 XM_011513949.2 XM_011513950.2 XM_011513951.2 XM_017009000.1 XM_017009001.1 XM_017009002.1 XM_017009003.1 XM_017009004.1 XM_017009005.1 XM_017009006.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3343078 c.1343C>G p.T448R Chr.5: 459411-459411 on Build GRCh38
5647144 c.1366C>T p.Q456* Chr.5: 459434-459434 on Build GRCh38
5459696 c.1389C>A p.S463R Chr.5: 459457-459457 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs922676879 Chr.5: 459323-459323 on Build GRCh38
rs530304095 Chr.5: 459514-459514 on Build GRCh38
rs758868550 Chr.5: 459460-459461 on Build GRCh38
rs925012719 Chr.5: 459701-459701 on Build GRCh38
rs751058427 Chr.5: 459396-459396 on Build GRCh38
rs997396691 Chr.5: 459685-459685 on Build GRCh38
rs767424178 Chr.5: 459412-459412 on Build GRCh38
rs895662458 Chr.5: 459706-459706 on Build GRCh38
rs182873651 Chr.5: 459479-459479 on Build GRCh38
rs976912129 Chr.5: 459321-459321 on Build GRCh38
rs954089400 Chr.5: 459639-459639 on Build GRCh38
rs193285524 Chr.5: 459692-459692 on Build GRCh38
rs1056947839 Chr.5: 459705-459705 on Build GRCh38
rs373527336 Chr.5: 459411-459411 on Build GRCh38
rs904295249 Chr.5: 459745-459745 on Build GRCh38
rs765925516 Chr.5: 459381-459381 on Build GRCh38
rs548392700 Chr.5: 459521-459521 on Build GRCh38
rs777676282 Chr.5: 459451-459451 on Build GRCh38
rs761810579 Chr.5: 459388-459392 on Build GRCh38
rs2561664 Chr.5: 459678-459678 on Build GRCh38
rs924746067 Chr.5: 459316-459316 on Build GRCh38
rs1012672824 Chr.5: 459477-459477 on Build GRCh38
rs141734504 Chr.5: 459418-459418 on Build GRCh38
rs375802350 Chr.5: 459416-459416 on Build GRCh38
rs77253901 Chr.5: 459619-459619 on Build GRCh38
rs199851464 Chr.5: 459440-459440 on Build GRCh38
rs929249922 Chr.5: 459700-459700 on Build GRCh38
rs979047342 Chr.5: 459314-459314 on Build GRCh38
rs369067591 Chr.5: 459511-459511 on Build GRCh38
rs762978188 Chr.5: 459496-459497 on Build GRCh38
rs80046645 Chr.5: 459300-459300 on Build GRCh38
rs1037635750 Chr.5: 459417-459417 on Build GRCh38
rs572385357 Chr.5: 459672-459672 on Build GRCh38
rs386684258 Chr.5: 459656-459657 on Build GRCh38
rs1030082149 Chr.5: 459691-459691 on Build GRCh38
rs762645195 Chr.5: 459679-459679 on Build GRCh38
rs1044170958 Chr.5: 459709-459709 on Build GRCh38
rs537226771 Chr.5: 459553-459553 on Build GRCh38
rs763893988 Chr.5: 459720-459720 on Build GRCh38
rs553461347 Chr.5: 459667-459667 on Build GRCh38
rs747042270 Chr.5: 459503-459503 on Build GRCh38
rs547012886 Chr.5: 459331-459331 on Build GRCh38
rs895567886 Chr.5: 459474-459474 on Build GRCh38
rs957837471 Chr.5: 459707-459707 on Build GRCh38
rs780344738 Chr.5: 459498-459498 on Build GRCh38
rs1050188981 Chr.5: 459332-459332 on Build GRCh38
rs188768527 Chr.5: 459532-459532 on Build GRCh38
rs1031905542 Chr.5: 459580-459580 on Build GRCh38
rs77955022 Chr.5: 459656-459656 on Build GRCh38
rs916149294 Chr.5: 459563-459563 on Build GRCh38
rs1006254156 Chr.5: 459340-459340 on Build GRCh38
rs956319477 Chr.5: 459618-459618 on Build GRCh38
rs1054829770 Chr.5: 459542-459542 on Build GRCh38
rs2561663 Chr.5: 459657-459657 on Build GRCh38
rs746137344 Chr.5: 459468-459468 on Build GRCh38
rs1013055272 Chr.5: 459708-459708 on Build GRCh38
rs143640519 Chr.5: 459465-459468 on Build GRCh38
rs760016446 Chr.5: 459617-459617 on Build GRCh38
rs752452703 Chr.5: 459414-459414 on Build GRCh38
rs59635824 Chr.5: 459566-459566 on Build GRCh38
rs956227005 Chr.5: 459319-459319 on Build GRCh38
rs747483071 Chr.5: 459622-459622 on Build GRCh38

View Full Product Details