Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 6714424-6714925 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PAPD7
Entrez Gene ID
11044
Gene Name
PAP associated domain containing 7
Gene Aliases
LAK-1, LAK1, POLK, POLS, TRF4, TRF4-1, TRF41, TUTASE5
UniGene
Hs.481542
Gene Chromosome Location
Chr.5: 6713952-6757048 on Build GRCh38
Transcripts
NM_001171805.1 NM_006999.4 XM_005248234.3 XM_017008989.1

SNP Detail

SNP ID Chromosome Location
rs111451280 Chr.5: 6714677-6714677 on Build GRCh38
rs541110146 Chr.5: 6714481-6714481 on Build GRCh38
rs959160106 Chr.5: 6714844-6714845 on Build GRCh38
rs949827087 Chr.5: 6714697-6714697 on Build GRCh38
rs551439140 Chr.5: 6714783-6714783 on Build GRCh38
rs917713582 Chr.5: 6714661-6714661 on Build GRCh38
rs1042376699 Chr.5: 6714468-6714468 on Build GRCh38
rs983671160 Chr.5: 6714574-6714574 on Build GRCh38
rs1030894444 Chr.5: 6714556-6714556 on Build GRCh38
rs961029166 Chr.5: 6714602-6714602 on Build GRCh38
rs527413169 Chr.5: 6714858-6714858 on Build GRCh38
rs1050605498 Chr.5: 6714827-6714827 on Build GRCh38
rs1000942678 Chr.5: 6714473-6714473 on Build GRCh38
rs963594246 Chr.5: 6714805-6714805 on Build GRCh38
rs547538973 Chr.5: 6714867-6714867 on Build GRCh38
rs1027026653 Chr.5: 6714831-6714831 on Build GRCh38
rs924503950 Chr.5: 6714726-6714726 on Build GRCh38
rs542766991 Chr.5: 6714688-6714688 on Build GRCh38
rs919699923 Chr.5: 6714612-6714612 on Build GRCh38
rs199781238 Chr.5: 6714763-6714764 on Build GRCh38
rs867175536 Chr.5: 6714741-6714741 on Build GRCh38
rs902512589 Chr.5: 6714470-6714470 on Build GRCh38
rs936675334 Chr.5: 6714718-6714718 on Build GRCh38
rs973783668 Chr.5: 6714830-6714830 on Build GRCh38
rs571249917 Chr.5: 6714786-6714786 on Build GRCh38
rs996626468 Chr.5: 6714751-6714751 on Build GRCh38
rs990670099 Chr.5: 6714631-6714631 on Build GRCh38
rs929023128 Chr.5: 6714735-6714735 on Build GRCh38
rs984517356 Chr.5: 6714740-6714740 on Build GRCh38
rs754996277 Chr.5: 6714893-6714893 on Build GRCh38
rs529094777 Chr.5: 6714679-6714679 on Build GRCh38
rs760754068 Chr.5: 6714829-6714829 on Build GRCh38
rs937734022 Chr.5: 6714622-6714622 on Build GRCh38
rs890621656 Chr.5: 6714758-6714758 on Build GRCh38
rs1015303364 Chr.5: 6714594-6714594 on Build GRCh38
rs1042280816 Chr.5: 6714717-6714717 on Build GRCh38
rs567317963 Chr.5: 6714895-6714895 on Build GRCh38
rs562644152 Chr.5: 6714732-6714732 on Build GRCh38
rs918757547 Chr.5: 6714495-6714495 on Build GRCh38
rs559941746 Chr.5: 6714489-6714489 on Build GRCh38
rs946734127 Chr.5: 6714804-6714804 on Build GRCh38
rs151312829 Chr.5: 6714741-6714742 on Build GRCh38
rs376790335 Chr.5: 6714766-6714766 on Build GRCh38
rs867135528 Chr.5: 6714504-6714504 on Build GRCh38
rs974253536 Chr.5: 6714608-6714608 on Build GRCh38
rs1009141537 Chr.5: 6714880-6714880 on Build GRCh38
rs954946303 Chr.5: 6714568-6714568 on Build GRCh38
rs751442126 Chr.5: 6714861-6714861 on Build GRCh38
rs981271170 Chr.5: 6714710-6714710 on Build GRCh38
rs902543687 Chr.5: 6714718-6714719 on Build GRCh38
rs893776880 Chr.5: 6714787-6714787 on Build GRCh38

View Full Product Details