Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 9036851-9037390 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SEMA5A
Entrez Gene ID
9037
Gene Name
semaphorin 5A
Gene Aliases
SEMAF, semF
UniGene
Hs.27621
Gene Chromosome Location
Chr.5: 9035026-9546121 on Build GRCh38
Transcripts
NM_003966.2 XM_006714506.2 XM_006714507.3 XM_011514155.2 XM_011514156.1 XM_011514157.1 XM_011514158.1 XM_011514159.2 XM_017010016.1

SNP Detail

SNP ID Chromosome Location
rs748272292 Chr.5: 9036905-9036905 on Build GRCh38
rs189618508 Chr.5: 9037231-9037231 on Build GRCh38
rs926893362 Chr.5: 9037268-9037268 on Build GRCh38
rs573873364 Chr.5: 9037135-9037135 on Build GRCh38
rs570358912 Chr.5: 9037215-9037215 on Build GRCh38
rs1040402824 Chr.5: 9037343-9037343 on Build GRCh38
rs533815484 Chr.5: 9037322-9037322 on Build GRCh38
rs200112336 Chr.5: 9037161-9037161 on Build GRCh38
rs938297871 Chr.5: 9037279-9037279 on Build GRCh38
rs181463926 Chr.5: 9037319-9037319 on Build GRCh38
rs192437221 Chr.5: 9036989-9036989 on Build GRCh38
rs968052917 Chr.5: 9037097-9037097 on Build GRCh38
rs1022877499 Chr.5: 9037183-9037183 on Build GRCh38
rs1002380032 Chr.5: 9037011-9037011 on Build GRCh38
rs896560948 Chr.5: 9037014-9037014 on Build GRCh38
rs866748627 Chr.5: 9036910-9036910 on Build GRCh38
rs184169866 Chr.5: 9036896-9036896 on Build GRCh38
rs968226286 Chr.5: 9037249-9037249 on Build GRCh38
rs964343802 Chr.5: 9037202-9037202 on Build GRCh38
rs36201828 Chr.5: 9037157-9037163 on Build GRCh38
rs763131654 Chr.5: 9036959-9036959 on Build GRCh38
rs1031258034 Chr.5: 9037222-9037222 on Build GRCh38
rs922453855 Chr.5: 9037214-9037214 on Build GRCh38
rs975263193 Chr.5: 9037209-9037209 on Build GRCh38
rs530152532 Chr.5: 9037073-9037073 on Build GRCh38
rs905336958 Chr.5: 9036993-9036993 on Build GRCh38
rs578149590 Chr.5: 9037013-9037013 on Build GRCh38
rs1040896332 Chr.5: 9037057-9037057 on Build GRCh38
rs779685950 Chr.5: 9037156-9037157 on Build GRCh38
rs1009625017 Chr.5: 9037321-9037321 on Build GRCh38
rs542346981 Chr.5: 9037149-9037149 on Build GRCh38
rs776379565 Chr.5: 9037280-9037280 on Build GRCh38
rs113631514 Chr.5: 9037162-9037162 on Build GRCh38
rs769749893 Chr.5: 9036925-9036925 on Build GRCh38
rs368985722 Chr.5: 9037028-9037029 on Build GRCh38
rs903814522 Chr.5: 9037346-9037346 on Build GRCh38
rs910800014 Chr.5: 9037306-9037306 on Build GRCh38
rs201071968 Chr.5: 9037159-9037159 on Build GRCh38
rs113910924 Chr.5: 9037163-9037163 on Build GRCh38
rs78456783 Chr.5: 9037228-9037228 on Build GRCh38
rs764026504 Chr.5: 9037263-9037263 on Build GRCh38
rs202131312 Chr.5: 9037160-9037160 on Build GRCh38
rs527555814 Chr.5: 9037341-9037341 on Build GRCh38
rs189395513 Chr.5: 9036942-9036942 on Build GRCh38
rs924356028 Chr.5: 9036933-9036933 on Build GRCh38
rs562235665 Chr.5: 9037216-9037216 on Build GRCh38
rs946973549 Chr.5: 9037002-9037003 on Build GRCh38
rs185680580 Chr.5: 9037050-9037050 on Build GRCh38
rs1032930373 Chr.5: 9037148-9037148 on Build GRCh38
rs754086656 Chr.5: 9037365-9037365 on Build GRCh38
rs1610939 Chr.5: 9037159-9037165 on Build GRCh38
rs148006277 Chr.5: 9037026-9037026 on Build GRCh38
rs941593845 Chr.5: 9036968-9036968 on Build GRCh38
rs915385775 Chr.5: 9036995-9036995 on Build GRCh38
rs773550434 Chr.5: 9036928-9036928 on Build GRCh38
rs933766415 Chr.5: 9036894-9036894 on Build GRCh38
rs993543632 Chr.5: 9037099-9037099 on Build GRCh38
rs751184710 Chr.5: 9037182-9037182 on Build GRCh38

View Full Product Details