Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 36673860-36674360 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC1A3
Entrez Gene ID
6507
Gene Name
solute carrier family 1 member 3
Gene Aliases
EA6, EAAT1, GLAST, GLAST1
UniGene
Hs.481918
Gene Chromosome Location
Chr.5: 36606355-36688334 on Build GRCh38
Transcripts
NM_001166695.2 NM_001289939.1 NM_001289940.1 NM_004172.4 XM_005248342.2 XM_011514084.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1067630 c.526A>G p.N176D Chr.5: 36674050-36674050 on Build GRCh38
7418217 c.557G>A p.C186Y Chr.5: 36674081-36674081 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs200014819 Chr.5: 36674123-36674123 on Build GRCh38
rs773616587 Chr.5: 36674114-36674114 on Build GRCh38
rs945187734 Chr.5: 36674226-36674226 on Build GRCh38
rs1018210007 Chr.5: 36673932-36673932 on Build GRCh38
rs372718413 Chr.5: 36674011-36674011 on Build GRCh38
rs977000555 Chr.5: 36674073-36674073 on Build GRCh38
rs1018347566 Chr.5: 36674300-36674300 on Build GRCh38
rs1006923831 Chr.5: 36674284-36674284 on Build GRCh38
rs949736033 Chr.5: 36674220-36674220 on Build GRCh38
rs112436413 Chr.5: 36673905-36673905 on Build GRCh38
rs779052167 Chr.5: 36674068-36674068 on Build GRCh38
rs779967208 Chr.5: 36674023-36674023 on Build GRCh38
rs771481177 Chr.5: 36674133-36674133 on Build GRCh38
rs186052446 Chr.5: 36673895-36673895 on Build GRCh38
rs1054347632 Chr.5: 36674246-36674246 on Build GRCh38
rs888548463 Chr.5: 36674271-36674271 on Build GRCh38
rs137852620 Chr.5: 36674080-36674080 on Build GRCh38
rs910300550 Chr.5: 36674145-36674145 on Build GRCh38
rs760494602 Chr.5: 36674283-36674283 on Build GRCh38
rs938026185 Chr.5: 36674242-36674242 on Build GRCh38
rs1002185083 Chr.5: 36673906-36673906 on Build GRCh38
rs902674457 Chr.5: 36673883-36673883 on Build GRCh38
rs756967365 Chr.5: 36673999-36673999 on Build GRCh38
rs1055093167 Chr.5: 36674253-36674253 on Build GRCh38
rs772359617 Chr.5: 36674082-36674082 on Build GRCh38
rs1054045506 Chr.5: 36674163-36674163 on Build GRCh38
rs922839111 Chr.5: 36674099-36674099 on Build GRCh38
rs374100855 Chr.5: 36674009-36674009 on Build GRCh38
rs146581375 Chr.5: 36673899-36673899 on Build GRCh38
rs1015741005 Chr.5: 36674054-36674054 on Build GRCh38
rs763673944 Chr.5: 36674318-36674318 on Build GRCh38
rs112687916 Chr.5: 36674227-36674227 on Build GRCh38
rs2033224 Chr.5: 36673977-36673977 on Build GRCh38
rs532433761 Chr.5: 36674334-36674334 on Build GRCh38
rs749384702 Chr.5: 36674141-36674145 on Build GRCh38
rs749277303 Chr.5: 36674047-36674047 on Build GRCh38
rs913706243 Chr.5: 36674173-36674173 on Build GRCh38
rs564972278 Chr.5: 36674243-36674243 on Build GRCh38
rs768557336 Chr.5: 36674066-36674066 on Build GRCh38
rs1006138377 Chr.5: 36674037-36674037 on Build GRCh38
rs909605238 Chr.5: 36674158-36674158 on Build GRCh38
rs936394385 Chr.5: 36674160-36674160 on Build GRCh38
rs750200485 Chr.5: 36674010-36674010 on Build GRCh38
rs991260960 Chr.5: 36674198-36674198 on Build GRCh38
rs1033802672 Chr.5: 36673924-36673924 on Build GRCh38
rs901146631 Chr.5: 36674324-36674324 on Build GRCh38
rs916919128 Chr.5: 36674210-36674210 on Build GRCh38
rs559313950 Chr.5: 36673912-36673912 on Build GRCh38
rs951205343 Chr.5: 36674147-36674147 on Build GRCh38
rs748275359 Chr.5: 36674076-36674076 on Build GRCh38
rs540430171 Chr.5: 36674238-36674238 on Build GRCh38

View Full Product Details