Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 43121932-43122442 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF131
Entrez Gene ID
7690
Gene Name
zinc finger protein 131
Gene Aliases
ZBTB35, pHZ-10
UniGene
Hs.535804
Gene Chromosome Location
Chr.5: 43120848-43176324 on Build GRCh38
Transcripts
NM_001297548.1 NM_003432.2 NR_123719.1 NR_123720.1 XM_005248359.2 XM_005248360.2 XM_005248361.2 XM_005248362.4 XM_005248363.4 XM_005248365.3 XM_005248367.2 XM_017009830.1 XM_017009832.1 XM_017009833.1 XM_017009834.1 XM_017009835.1 XM_017009836.1 XM_017009837.1 XM_017009839.1 XM_017009840.1 XM_017009841.1 XM_017009842.1 XM_017009843.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6007129 c.48T>A p.H16Q Chr.5: 43122101-43122101 on Build GRCh38
5556348 c.50A>G p.H17R Chr.5: 43122103-43122103 on Build GRCh38
4808828 c.63C>T p.L21L Chr.5: 43122116-43122116 on Build GRCh38
4347942 c.63C>A p.L21L Chr.5: 43122116-43122116 on Build GRCh38
3776719 c.85G>C p.E29Q Chr.5: 43122138-43122138 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs545415564 Chr.5: 43122227-43122227 on Build GRCh38
rs775870112 Chr.5: 43122026-43122028 on Build GRCh38
rs749781006 Chr.5: 43121998-43121998 on Build GRCh38
rs973032026 Chr.5: 43122146-43122146 on Build GRCh38
rs1005134496 Chr.5: 43122073-43122073 on Build GRCh38
rs921299622 Chr.5: 43122293-43122293 on Build GRCh38
rs553456627 Chr.5: 43122046-43122046 on Build GRCh38
rs935996751 Chr.5: 43122241-43122241 on Build GRCh38
rs780447324 Chr.5: 43121994-43121994 on Build GRCh38
rs977364947 Chr.5: 43122235-43122235 on Build GRCh38
rs749105585 Chr.5: 43122221-43122221 on Build GRCh38
rs779507395 Chr.5: 43122062-43122062 on Build GRCh38
rs963846163 Chr.5: 43121958-43121958 on Build GRCh38
rs745685804 Chr.5: 43122014-43122014 on Build GRCh38
rs775802733 Chr.5: 43122164-43122164 on Build GRCh38
rs368322196 Chr.5: 43122005-43122005 on Build GRCh38
rs11287008 Chr.5: 43122269-43122269 on Build GRCh38
rs780744251 Chr.5: 43122199-43122199 on Build GRCh38
rs745498385 Chr.5: 43122077-43122077 on Build GRCh38
rs776359711 Chr.5: 43122012-43122012 on Build GRCh38
rs944161791 Chr.5: 43122247-43122254 on Build GRCh38
rs777419048 Chr.5: 43122004-43122004 on Build GRCh38
rs908692898 Chr.5: 43122242-43122244 on Build GRCh38
rs1055892746 Chr.5: 43122006-43122006 on Build GRCh38
rs753212370 Chr.5: 43122192-43122192 on Build GRCh38
rs778282173 Chr.5: 43122247-43122247 on Build GRCh38
rs749627217 Chr.5: 43122395-43122395 on Build GRCh38
rs761021661 Chr.5: 43122054-43122054 on Build GRCh38
rs1031890453 Chr.5: 43122053-43122053 on Build GRCh38
rs760947665 Chr.5: 43122159-43122159 on Build GRCh38
rs774077640 Chr.5: 43122248-43122249 on Build GRCh38
rs769540425 Chr.5: 43122081-43122081 on Build GRCh38
rs953053468 Chr.5: 43122232-43122232 on Build GRCh38
rs767756270 Chr.5: 43122027-43122027 on Build GRCh38
rs925821579 Chr.5: 43122240-43122240 on Build GRCh38
rs750609386 Chr.5: 43122030-43122030 on Build GRCh38
rs887993175 Chr.5: 43122382-43122382 on Build GRCh38
rs756484469 Chr.5: 43122032-43122032 on Build GRCh38
rs577377153 Chr.5: 43122291-43122291 on Build GRCh38
rs746900252 Chr.5: 43122247-43122249 on Build GRCh38
rs761812739 Chr.5: 43122022-43122022 on Build GRCh38
rs950094145 Chr.5: 43122313-43122313 on Build GRCh38
rs779516968 Chr.5: 43122216-43122216 on Build GRCh38
rs929585326 Chr.5: 43122350-43122350 on Build GRCh38
rs1046347391 Chr.5: 43122316-43122316 on Build GRCh38
rs774425272 Chr.5: 43122021-43122021 on Build GRCh38
rs950211003 Chr.5: 43122025-43122025 on Build GRCh38
rs904450355 Chr.5: 43122360-43122360 on Build GRCh38
rs775339286 Chr.5: 43122082-43122082 on Build GRCh38
rs1029486730 Chr.5: 43122186-43122186 on Build GRCh38
rs562986941 Chr.5: 43122336-43122336 on Build GRCh38
rs199924282 Chr.5: 43122185-43122185 on Build GRCh38
rs60096743 Chr.5: 43122268-43122269 on Build GRCh38
rs1025765212 Chr.5: 43122047-43122047 on Build GRCh38
rs749022747 Chr.5: 43122102-43122102 on Build GRCh38
rs756791883 Chr.5: 43122070-43122070 on Build GRCh38
rs151063287 Chr.5: 43121978-43121978 on Build GRCh38
rs761902665 Chr.5: 43122113-43122113 on Build GRCh38
rs544880548 Chr.5: 43122307-43122307 on Build GRCh38
rs75933834 Chr.5: 43122245-43122248 on Build GRCh38
rs759919117 Chr.5: 43122181-43122181 on Build GRCh38
rs149795884 Chr.5: 43122282-43122282 on Build GRCh38
rs767666296 Chr.5: 43121989-43121989 on Build GRCh38
rs192738655 Chr.5: 43121991-43121991 on Build GRCh38
rs556121503 Chr.5: 43121992-43121992 on Build GRCh38
rs775325604 Chr.5: 43122015-43122015 on Build GRCh38
rs773347302 Chr.5: 43122148-43122148 on Build GRCh38
rs772117297 Chr.5: 43122116-43122116 on Build GRCh38
rs755530335 Chr.5: 43122051-43122051 on Build GRCh38
rs1049319498 Chr.5: 43122353-43122353 on Build GRCh38
rs111680043 Chr.5: 43121999-43121999 on Build GRCh38
rs998716125 Chr.5: 43122035-43122035 on Build GRCh38
rs35497367 Chr.5: 43122246-43122247 on Build GRCh38
rs372521618 Chr.5: 43122207-43122207 on Build GRCh38
rs764355897 Chr.5: 43122207-43122207 on Build GRCh38
rs774230885 Chr.5: 43122110-43122110 on Build GRCh38
rs571772162 Chr.5: 43122198-43122198 on Build GRCh38
rs754233275 Chr.5: 43122176-43122176 on Build GRCh38
rs1027243671 Chr.5: 43122219-43122220 on Build GRCh38
rs369196197 Chr.5: 43122190-43122190 on Build GRCh38
rs200355739 Chr.5: 43122247-43122247 on Build GRCh38
rs991459136 Chr.5: 43122277-43122277 on Build GRCh38
rs750114096 Chr.5: 43122203-43122203 on Build GRCh38
rs770723168 Chr.5: 43122011-43122011 on Build GRCh38
rs768400672 Chr.5: 43122225-43122225 on Build GRCh38
rs375732228 Chr.5: 43122103-43122103 on Build GRCh38
rs372795836 Chr.5: 43122016-43122016 on Build GRCh38
rs754371005 Chr.5: 43122050-43122050 on Build GRCh38
rs764213044 Chr.5: 43122018-43122018 on Build GRCh38
rs11953246 Chr.5: 43122365-43122365 on Build GRCh38
rs552571467 Chr.5: 43121961-43121961 on Build GRCh38
rs746525850 Chr.5: 43122063-43122063 on Build GRCh38
rs377071760 Chr.5: 43122244-43122244 on Build GRCh38
rs934557665 Chr.5: 43122383-43122383 on Build GRCh38
rs878898278 Chr.5: 43122244-43122244 on Build GRCh38

View Full Product Details