Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 50777943-50778483 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PARP8
Entrez Gene ID
79668
Gene Name
poly(ADP-ribose) polymerase family member 8
Gene Aliases
ARTD16, pART16
UniGene
Hs.369581
Gene Chromosome Location
Chr.5: 50665899-50846522 on Build GRCh38
Transcripts
NM_001178055.1 NM_001178056.1 NM_024615.3 XM_005248596.4 XM_011543631.2 XM_011543632.2 XM_011543633.2 XM_011543634.2 XM_011543635.2 XM_011543636.2 XM_011543640.2 XM_011543641.2 XM_011543642.2 XM_011543643.2 XM_017009850.1 XM_017009851.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1068543 c.561C>T p.I187I Chr.5: 50778111-50778111 on Build GRCh38
1328774 c.574C>G p.L192V Chr.5: 50778124-50778124 on Build GRCh38
7167756 c.553C>T p.L185L Chr.5: 50778103-50778103 on Build GRCh38
3343184 c.536A>T p.D179V Chr.5: 50778086-50778086 on Build GRCh38
3730932 c.567T>C p.V189V Chr.5: 50778117-50778117 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs909670814 Chr.5: 50778403-50778403 on Build GRCh38
rs529988294 Chr.5: 50778359-50778359 on Build GRCh38
rs528605318 Chr.5: 50778430-50778430 on Build GRCh38
rs533041611 Chr.5: 50778195-50778195 on Build GRCh38
rs763603761 Chr.5: 50778079-50778079 on Build GRCh38
rs767140880 Chr.5: 50778174-50778178 on Build GRCh38
rs751278159 Chr.5: 50778040-50778040 on Build GRCh38
rs764184893 Chr.5: 50778297-50778297 on Build GRCh38
rs76168257 Chr.5: 50778398-50778398 on Build GRCh38
rs543820697 Chr.5: 50778239-50778239 on Build GRCh38
rs776518124 Chr.5: 50778049-50778049 on Build GRCh38
rs753217346 Chr.5: 50778180-50778180 on Build GRCh38
rs771245671 Chr.5: 50778046-50778046 on Build GRCh38
rs375021903 Chr.5: 50778045-50778045 on Build GRCh38
rs1039553547 Chr.5: 50778196-50778196 on Build GRCh38
rs575169208 Chr.5: 50778227-50778227 on Build GRCh38
rs925475013 Chr.5: 50778009-50778009 on Build GRCh38
rs376804637 Chr.5: 50778161-50778161 on Build GRCh38
rs762813123 Chr.5: 50778292-50778292 on Build GRCh38
rs776705402 Chr.5: 50778219-50778219 on Build GRCh38
rs758207644 Chr.5: 50778028-50778028 on Build GRCh38
rs902490439 Chr.5: 50778291-50778291 on Build GRCh38
rs777667574 Chr.5: 50778041-50778041 on Build GRCh38
rs978174753 Chr.5: 50777972-50777972 on Build GRCh38
rs750067490 Chr.5: 50778406-50778406 on Build GRCh38
rs760230087 Chr.5: 50778119-50778119 on Build GRCh38
rs561170790 Chr.5: 50778316-50778316 on Build GRCh38
rs774428207 Chr.5: 50778362-50778362 on Build GRCh38
rs770385784 Chr.5: 50778051-50778051 on Build GRCh38
rs1030763381 Chr.5: 50778251-50778251 on Build GRCh38
rs764522092 Chr.5: 50778103-50778103 on Build GRCh38
rs754566290 Chr.5: 50778132-50778132 on Build GRCh38
rs544175307 Chr.5: 50778039-50778040 on Build GRCh38
rs1046495888 Chr.5: 50778070-50778070 on Build GRCh38
rs754705235 Chr.5: 50778048-50778049 on Build GRCh38
rs1023719411 Chr.5: 50778318-50778318 on Build GRCh38
rs971400645 Chr.5: 50778329-50778329 on Build GRCh38
rs774845957 Chr.5: 50778111-50778111 on Build GRCh38
rs555984060 Chr.5: 50778013-50778013 on Build GRCh38
rs958108964 Chr.5: 50778306-50778306 on Build GRCh38
rs747894183 Chr.5: 50778026-50778026 on Build GRCh38
rs371730211 Chr.5: 50778062-50778062 on Build GRCh38
rs745948749 Chr.5: 50778050-50778050 on Build GRCh38
rs918229633 Chr.5: 50778096-50778096 on Build GRCh38
rs777429850 Chr.5: 50778455-50778455 on Build GRCh38
rs929732784 Chr.5: 50778147-50778147 on Build GRCh38
rs942384866 Chr.5: 50778420-50778420 on Build GRCh38
rs571553216 Chr.5: 50778315-50778315 on Build GRCh38
rs568077916 Chr.5: 50778363-50778363 on Build GRCh38
rs770946836 Chr.5: 50778203-50778203 on Build GRCh38
rs372488538 Chr.5: 50778046-50778047 on Build GRCh38

View Full Product Details