Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 64598256-64598763 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RGS7BP
Entrez Gene ID
401190
Gene Name
regulator of G-protein signaling 7 binding protein
Gene Aliases
R7BP
UniGene
Hs.657133
Gene Chromosome Location
Chr.5: 64504565-64614661 on Build GRCh38
Transcripts
NM_001029875.2 NM_001271890.1 NM_001271891.1 NR_073511.1 XM_005248502.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3617389 c.652G>A p.E218K Chr.5: 64598405-64598405 on Build GRCh38
3617388 c.651G>A p.R217R Chr.5: 64598404-64598404 on Build GRCh38
3617387 c.634C>T p.L212F Chr.5: 64598387-64598387 on Build GRCh38
3617386 c.627G>A p.M209I Chr.5: 64598380-64598380 on Build GRCh38
1069307 c.622G>T p.E208* Chr.5: 64598375-64598375 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1051631749 Chr.5: 64598513-64598513 on Build GRCh38
rs781223999 Chr.5: 64598411-64598411 on Build GRCh38
rs1014825004 Chr.5: 64598696-64598696 on Build GRCh38
rs775829800 Chr.5: 64598460-64598460 on Build GRCh38
rs978973140 Chr.5: 64598737-64598737 on Build GRCh38
rs756431967 Chr.5: 64598337-64598337 on Build GRCh38
rs1020487176 Chr.5: 64598730-64598730 on Build GRCh38
rs377268590 Chr.5: 64598335-64598335 on Build GRCh38
rs6898818 Chr.5: 64598661-64598661 on Build GRCh38
rs371003261 Chr.5: 64598338-64598338 on Build GRCh38
rs940650462 Chr.5: 64598569-64598569 on Build GRCh38
rs543513913 Chr.5: 64598511-64598511 on Build GRCh38
rs6877763 Chr.5: 64598653-64598653 on Build GRCh38
rs921396714 Chr.5: 64598721-64598721 on Build GRCh38
rs768845140 Chr.5: 64598483-64598483 on Build GRCh38
rs1009906372 Chr.5: 64598461-64598461 on Build GRCh38
rs771009091 Chr.5: 64598391-64598391 on Build GRCh38
rs551938904 Chr.5: 64598580-64598580 on Build GRCh38
rs745686977 Chr.5: 64598422-64598422 on Build GRCh38
rs886346604 Chr.5: 64598543-64598543 on Build GRCh38
rs545578294 Chr.5: 64598530-64598530 on Build GRCh38
rs762220457 Chr.5: 64598319-64598319 on Build GRCh38
rs958579014 Chr.5: 64598588-64598588 on Build GRCh38
rs936855481 Chr.5: 64598727-64598727 on Build GRCh38
rs150104966 Chr.5: 64598379-64598379 on Build GRCh38
rs1037586050 Chr.5: 64598574-64598574 on Build GRCh38
rs146036934 Chr.5: 64598344-64598344 on Build GRCh38
rs892765318 Chr.5: 64598374-64598374 on Build GRCh38
rs922728365 Chr.5: 64598481-64598481 on Build GRCh38
rs776338236 Chr.5: 64598579-64598579 on Build GRCh38
rs975535308 Chr.5: 64598709-64598709 on Build GRCh38
rs376946870 Chr.5: 64598442-64598442 on Build GRCh38
rs373375396 Chr.5: 64598423-64598423 on Build GRCh38
rs777148258 Chr.5: 64598346-64598346 on Build GRCh38
rs967622294 Chr.5: 64598736-64598736 on Build GRCh38
rs752166851 Chr.5: 64598315-64598315 on Build GRCh38
rs369413408 Chr.5: 64598462-64598462 on Build GRCh38
rs532141646 Chr.5: 64598576-64598576 on Build GRCh38
rs951332915 Chr.5: 64598648-64598648 on Build GRCh38
rs542354842 Chr.5: 64598357-64598357 on Build GRCh38
rs373339872 Chr.5: 64598465-64598465 on Build GRCh38
rs1001171873 Chr.5: 64598591-64598591 on Build GRCh38
rs913456800 Chr.5: 64598303-64598303 on Build GRCh38
rs993031777 Chr.5: 64598295-64598295 on Build GRCh38
rs1054048220 Chr.5: 64598366-64598366 on Build GRCh38
rs952652131 Chr.5: 64598531-64598531 on Build GRCh38
rs774297884 Chr.5: 64598486-64598486 on Build GRCh38
rs1026862150 Chr.5: 64598625-64598625 on Build GRCh38
rs752270797 Chr.5: 64598342-64598342 on Build GRCh38
rs944154927 Chr.5: 64598292-64598293 on Build GRCh38
rs149979769 Chr.5: 64598718-64598718 on Build GRCh38
rs762121707 Chr.5: 64598740-64598740 on Build GRCh38

View Full Product Details