Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 64774602-64775103 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CWC27
Entrez Gene ID
10283
Gene Name
CWC27 spliceosome associated protein homolog
Gene Aliases
NY-CO-10, SDCCAG-10, SDCCAG10
UniGene
Hs.371372
Gene Chromosome Location
Chr.5: 64768918-65018763 on Build GRCh38
Transcripts
NM_001297644.1 NM_001297645.1 NM_001318000.1 NM_005869.3 XM_011543095.2 XM_011543096.1 XM_017008943.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7176744 c.115_136del22 p.N39fs*17 Chr.5: 64774763-64774784 on Build GRCh38
6104450 c.91A>G p.K31E Chr.5: 64774739-64774739 on Build GRCh38
5499564 c.139+4_139+5insAATTATGAC p.? Chr.5: 64774791-64774792 on Build GRCh38
5934954 c.139+5G>A p.? Chr.5: 64774792-64774792 on Build GRCh38
6638371 c.92A>T p.K31I Chr.5: 64774740-64774740 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1036935498 Chr.5: 64775065-64775065 on Build GRCh38
rs1044552735 Chr.5: 64775009-64775009 on Build GRCh38
rs780571389 Chr.5: 64774654-64774654 on Build GRCh38
rs910505074 Chr.5: 64774906-64774906 on Build GRCh38
rs967895602 Chr.5: 64774887-64774887 on Build GRCh38
rs115861531 Chr.5: 64774936-64774936 on Build GRCh38
rs371215899 Chr.5: 64774813-64774813 on Build GRCh38
rs914976159 Chr.5: 64774903-64774903 on Build GRCh38
rs199695297 Chr.5: 64774791-64774791 on Build GRCh38
rs922927440 Chr.5: 64774742-64774742 on Build GRCh38
rs913753883 Chr.5: 64774640-64774640 on Build GRCh38
rs942240248 Chr.5: 64774986-64774986 on Build GRCh38
rs781194080 Chr.5: 64774719-64774719 on Build GRCh38
rs1040435835 Chr.5: 64774734-64774734 on Build GRCh38
rs1049073808 Chr.5: 64775023-64775023 on Build GRCh38
rs200858460 Chr.5: 64774792-64774792 on Build GRCh38
rs910452901 Chr.5: 64774716-64774716 on Build GRCh38
rs773081038 Chr.5: 64774679-64774680 on Build GRCh38
rs751712173 Chr.5: 64774774-64774774 on Build GRCh38
rs771787453 Chr.5: 64774671-64774671 on Build GRCh38
rs979370511 Chr.5: 64774699-64774699 on Build GRCh38
rs1010748321 Chr.5: 64774876-64774876 on Build GRCh38
rs762620409 Chr.5: 64774754-64774754 on Build GRCh38
rs566068642 Chr.5: 64774884-64774884 on Build GRCh38
rs1036104600 Chr.5: 64774849-64774849 on Build GRCh38
rs114597273 Chr.5: 64774789-64774789 on Build GRCh38
rs771287488 Chr.5: 64775048-64775048 on Build GRCh38
rs769658848 Chr.5: 64774746-64774746 on Build GRCh38
rs891817233 Chr.5: 64775061-64775061 on Build GRCh38
rs188737499 Chr.5: 64774864-64774864 on Build GRCh38
rs772707775 Chr.5: 64774682-64774682 on Build GRCh38
rs373625212 Chr.5: 64774812-64774812 on Build GRCh38
rs762917044 Chr.5: 64774789-64774790 on Build GRCh38
rs745330221 Chr.5: 64774835-64774835 on Build GRCh38
rs929067484 Chr.5: 64774747-64774747 on Build GRCh38
rs956040466 Chr.5: 64774872-64774872 on Build GRCh38
rs988870878 Chr.5: 64774638-64774638 on Build GRCh38
rs141974787 Chr.5: 64774748-64774748 on Build GRCh38
rs928974388 Chr.5: 64774957-64774957 on Build GRCh38
rs377209646 Chr.5: 64774934-64774934 on Build GRCh38
rs10057598 Chr.5: 64774985-64774985 on Build GRCh38
rs150844528 Chr.5: 64774778-64774778 on Build GRCh38
rs923843411 Chr.5: 64774639-64774639 on Build GRCh38
rs1459189 Chr.5: 64775056-64775056 on Build GRCh38
rs939186620 Chr.5: 64774935-64774935 on Build GRCh38
rs1047493576 Chr.5: 64774961-64774961 on Build GRCh38
rs780659334 Chr.5: 64774793-64774793 on Build GRCh38
rs201754441 Chr.5: 64774790-64774790 on Build GRCh38
rs184047222 Chr.5: 64774810-64774810 on Build GRCh38
rs574809154 Chr.5: 64774914-64774914 on Build GRCh38
rs376182212 Chr.5: 64774691-64774691 on Build GRCh38
rs948031920 Chr.5: 64774994-64774994 on Build GRCh38
rs568023065 Chr.5: 64774651-64774651 on Build GRCh38
rs977913950 Chr.5: 64774916-64774916 on Build GRCh38
rs559263544 Chr.5: 64775017-64775017 on Build GRCh38
rs1012208400 Chr.5: 64774708-64774708 on Build GRCh38
rs754295604 Chr.5: 64774807-64774807 on Build GRCh38
rs76939619 Chr.5: 64774888-64774888 on Build GRCh38
rs369814947 Chr.5: 64774697-64774697 on Build GRCh38
rs759472038 Chr.5: 64774739-64774739 on Build GRCh38
rs953709503 Chr.5: 64774861-64774861 on Build GRCh38
rs887848960 Chr.5: 64775025-64775025 on Build GRCh38
rs201232840 Chr.5: 64774769-64774769 on Build GRCh38
rs749607358 Chr.5: 64774666-64774666 on Build GRCh38
rs939319723 Chr.5: 64774643-64774643 on Build GRCh38
rs746366494 Chr.5: 64774645-64774645 on Build GRCh38
rs140062546 Chr.5: 64774790-64774791 on Build GRCh38
rs140865325 Chr.5: 64774895-64774895 on Build GRCh38
rs780689602 Chr.5: 64774832-64774832 on Build GRCh38
rs545690638 Chr.5: 64775005-64775005 on Build GRCh38
rs1029667056 Chr.5: 64774845-64774845 on Build GRCh38
rs762013490 Chr.5: 64774787-64774787 on Build GRCh38

View Full Product Details