Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 75371745-75372258 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL4A3BP
Entrez Gene ID
10087
Gene Name
collagen type IV alpha 3 binding protein
Gene Aliases
CERT, CERTL, GPBP, MRD34, STARD11
UniGene
Hs.270437
Gene Chromosome Location
Chr.5: 75368486-75511981 on Build GRCh38
Transcripts
NM_001130105.1 NM_005713.2 NM_031361.2 XM_006714513.2 XM_011543090.2 XM_011543091.2 XM_017008919.1

SNP Detail

SNP ID Chromosome Location
rs868567471 Chr.5: 75372186-75372186 on Build GRCh38
rs72768356 Chr.5: 75371916-75371916 on Build GRCh38
rs188585129 Chr.5: 75372068-75372068 on Build GRCh38
rs1007354231 Chr.5: 75372226-75372226 on Build GRCh38
rs1045775223 Chr.5: 75372107-75372107 on Build GRCh38
rs887218869 Chr.5: 75371863-75371863 on Build GRCh38
rs1055764925 Chr.5: 75371813-75371813 on Build GRCh38
rs1004760039 Chr.5: 75371870-75371870 on Build GRCh38
rs1003462231 Chr.5: 75371835-75371835 on Build GRCh38
rs967169592 Chr.5: 75371972-75371972 on Build GRCh38
rs1008491078 Chr.5: 75372229-75372229 on Build GRCh38
rs926845212 Chr.5: 75372065-75372065 on Build GRCh38
rs993530844 Chr.5: 75372108-75372108 on Build GRCh38
rs115831918 Chr.5: 75372216-75372216 on Build GRCh38
rs925811398 Chr.5: 75372038-75372038 on Build GRCh38
rs549119423 Chr.5: 75371858-75371858 on Build GRCh38
rs1038551522 Chr.5: 75371883-75371883 on Build GRCh38
rs115785251 Chr.5: 75371906-75371906 on Build GRCh38
rs989937248 Chr.5: 75371989-75371989 on Build GRCh38
rs150560017 Chr.5: 75371899-75371899 on Build GRCh38
rs1038908133 Chr.5: 75372127-75372127 on Build GRCh38
rs997008518 Chr.5: 75372151-75372151 on Build GRCh38
rs898654147 Chr.5: 75372147-75372147 on Build GRCh38
rs915853350 Chr.5: 75372075-75372075 on Build GRCh38
rs1017560489 Chr.5: 75371921-75371921 on Build GRCh38
rs924285878 Chr.5: 75371966-75371966 on Build GRCh38
rs939620154 Chr.5: 75372079-75372079 on Build GRCh38
rs531258334 Chr.5: 75372035-75372035 on Build GRCh38
rs946937881 Chr.5: 75371772-75371773 on Build GRCh38
rs545576529 Chr.5: 75371773-75371773 on Build GRCh38
rs1056368288 Chr.5: 75372091-75372091 on Build GRCh38
rs940119750 Chr.5: 75372119-75372119 on Build GRCh38
rs950016840 Chr.5: 75372083-75372083 on Build GRCh38
rs757075439 Chr.5: 75371815-75371815 on Build GRCh38
rs957484010 Chr.5: 75372041-75372041 on Build GRCh38
rs950065382 Chr.5: 75371817-75371817 on Build GRCh38
rs898701229 Chr.5: 75371893-75371893 on Build GRCh38
rs1049179602 Chr.5: 75372154-75372154 on Build GRCh38
rs571700396 Chr.5: 75371957-75371957 on Build GRCh38
rs955749935 Chr.5: 75371985-75371985 on Build GRCh38
rs550957293 Chr.5: 75372100-75372100 on Build GRCh38
rs192381565 Chr.5: 75372140-75372140 on Build GRCh38
rs531176135 Chr.5: 75372016-75372020 on Build GRCh38
rs1049950427 Chr.5: 75372156-75372156 on Build GRCh38
rs908596767 Chr.5: 75372113-75372113 on Build GRCh38
rs975711004 Chr.5: 75371959-75371959 on Build GRCh38
rs376545316 Chr.5: 75371911-75371911 on Build GRCh38
rs1045645084 Chr.5: 75372043-75372043 on Build GRCh38
rs997114773 Chr.5: 75371895-75371895 on Build GRCh38

View Full Product Details