Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 95767907-95768426 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RHOBTB3
Entrez Gene ID
22836
Gene Name
Rho related BTB domain containing 3
Gene Aliases
-
UniGene
Hs.445030
Gene Chromosome Location
Chr.5: 95713522-95796367 on Build GRCh38
Transcripts
NM_014899.3 XM_011543279.2 XM_017009236.1 XM_017009237.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5404040 c.1240C>T p.L414F Chr.5: 95768124-95768124 on Build GRCh38
1071391 c.1210T>C p.W404R Chr.5: 95768094-95768094 on Build GRCh38
6172375 c.1250C>A p.P417Q Chr.5: 95768134-95768134 on Build GRCh38
1620673 c.1282+5G>A p.? Chr.5: 95768171-95768171 on Build GRCh38
7294508 c.1172T>C p.L391P Chr.5: 95768056-95768056 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769777769 Chr.5: 95768342-95768342 on Build GRCh38
rs749364749 Chr.5: 95768111-95768111 on Build GRCh38
rs780165004 Chr.5: 95768110-95768110 on Build GRCh38
rs887499733 Chr.5: 95768021-95768021 on Build GRCh38
rs762052926 Chr.5: 95767935-95767935 on Build GRCh38
rs768905935 Chr.5: 95768134-95768134 on Build GRCh38
rs953352234 Chr.5: 95768109-95768109 on Build GRCh38
rs920649877 Chr.5: 95768378-95768379 on Build GRCh38
rs370930103 Chr.5: 95768038-95768038 on Build GRCh38
rs753561451 Chr.5: 95768022-95768022 on Build GRCh38
rs368331385 Chr.5: 95768145-95768145 on Build GRCh38
rs748067031 Chr.5: 95768232-95768232 on Build GRCh38
rs949362299 Chr.5: 95768239-95768239 on Build GRCh38
rs375385622 Chr.5: 95768171-95768171 on Build GRCh38
rs763736929 Chr.5: 95768198-95768198 on Build GRCh38
rs970487734 Chr.5: 95768370-95768370 on Build GRCh38
rs772285824 Chr.5: 95768141-95768141 on Build GRCh38
rs201479791 Chr.5: 95768157-95768157 on Build GRCh38
rs929436953 Chr.5: 95767978-95767978 on Build GRCh38
rs1028846729 Chr.5: 95768101-95768101 on Build GRCh38
rs768158493 Chr.5: 95768144-95768144 on Build GRCh38
rs917927114 Chr.5: 95767953-95767953 on Build GRCh38
rs926832582 Chr.5: 95768364-95768364 on Build GRCh38
rs756196984 Chr.5: 95768094-95768094 on Build GRCh38
rs377606917 Chr.5: 95768039-95768039 on Build GRCh38
rs1014159919 Chr.5: 95768240-95768240 on Build GRCh38
rs959908347 Chr.5: 95768234-95768234 on Build GRCh38
rs61733078 Chr.5: 95768084-95768084 on Build GRCh38
rs980796125 Chr.5: 95768317-95768317 on Build GRCh38
rs777794232 Chr.5: 95768089-95768089 on Build GRCh38
rs866886492 Chr.5: 95768117-95768118 on Build GRCh38
rs905483340 Chr.5: 95767991-95767991 on Build GRCh38
rs776977512 Chr.5: 95768110-95768111 on Build GRCh38
rs1045333406 Chr.5: 95767990-95767990 on Build GRCh38
rs931459943 Chr.5: 95767999-95767999 on Build GRCh38
rs768119451 Chr.5: 95767998-95767998 on Build GRCh38
rs901507880 Chr.5: 95768054-95768054 on Build GRCh38
rs899529990 Chr.5: 95768088-95768088 on Build GRCh38
rs1048568816 Chr.5: 95768016-95768016 on Build GRCh38
rs762808666 Chr.5: 95768191-95768191 on Build GRCh38
rs949583154 Chr.5: 95767955-95767955 on Build GRCh38
rs375515640 Chr.5: 95768135-95768135 on Build GRCh38
rs780160264 Chr.5: 95768091-95768091 on Build GRCh38
rs1040711243 Chr.5: 95768087-95768087 on Build GRCh38
rs1022954322 Chr.5: 95768292-95768292 on Build GRCh38
rs199725974 Chr.5: 95768156-95768156 on Build GRCh38
rs369687651 Chr.5: 95768002-95768002 on Build GRCh38
rs200603095 Chr.5: 95768208-95768208 on Build GRCh38
rs760294946 Chr.5: 95768015-95768015 on Build GRCh38
rs764962207 Chr.5: 95768170-95768170 on Build GRCh38
rs973448093 Chr.5: 95768376-95768376 on Build GRCh38
rs771554966 Chr.5: 95768148-95768148 on Build GRCh38
rs56068445 Chr.5: 95768209-95768209 on Build GRCh38
rs1004637035 Chr.5: 95768023-95768023 on Build GRCh38
rs752581957 Chr.5: 95768060-95768060 on Build GRCh38

View Full Product Details