Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 139278697-139279199 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MATR3
Entrez Gene ID
9782
Gene Name
matrin 3
Gene Aliases
ALS21, MPD2, VCPDM
UniGene
Hs.268939
Gene Chromosome Location
Chr.5: 139273752-139331677 on Build GRCh38
Transcripts
NM_001194954.1 NM_001282278.1 NM_199189.2 NR_003141.3 NR_036536.1 NR_002915.1

Gene Symbol
SNHG4
Entrez Gene ID
724102
Gene Name
small nucleolar RNA host gene 4
Gene Aliases
NCRNA00059, U19H
UniGene
Hs.268939
Gene Chromosome Location
Chr.5: 139273752-139283184 on Build GRCh38
Transcripts
NM_001194954.1 NM_001282278.1 NM_199189.2 NR_003141.3 NR_036536.1 NR_002915.1

Gene Symbol
SNORA74A
Entrez Gene ID
26821
Gene Name
small nucleolar RNA, H/ACA box 74A
Gene Aliases
RNU19, U19
UniGene
Hs.684893
Gene Chromosome Location
Chr.5: 139278780-139278979 on Build GRCh38
Transcripts
NM_001194954.1 NM_001282278.1 NM_199189.2 NR_003141.3 NR_036536.1 NR_002915.1

SNP Detail

SNP ID Chromosome Location
rs774830325 Chr.5: 139278815-139278815 on Build GRCh38
rs200783037 Chr.5: 139278922-139278922 on Build GRCh38
rs772625981 Chr.5: 139279170-139279170 on Build GRCh38
rs746292862 Chr.5: 139279106-139279106 on Build GRCh38
rs369875833 Chr.5: 139278891-139278891 on Build GRCh38
rs374727741 Chr.5: 139278807-139278807 on Build GRCh38
rs780839683 Chr.5: 139278852-139278853 on Build GRCh38
rs1004235598 Chr.5: 139278839-139278839 on Build GRCh38
rs138489295 Chr.5: 139278781-139278781 on Build GRCh38
rs748151367 Chr.5: 139278787-139278787 on Build GRCh38
rs963319627 Chr.5: 139278880-139278880 on Build GRCh38
rs977365319 Chr.5: 139278896-139278896 on Build GRCh38
rs746887017 Chr.5: 139278916-139278916 on Build GRCh38
rs776356559 Chr.5: 139278803-139278803 on Build GRCh38
rs755976960 Chr.5: 139278929-139278930 on Build GRCh38
rs141402332 Chr.5: 139279060-139279060 on Build GRCh38
rs776414857 Chr.5: 139278869-139278869 on Build GRCh38
rs1054690325 Chr.5: 139278949-139278949 on Build GRCh38
rs749404686 Chr.5: 139278772-139278772 on Build GRCh38
rs751146415 Chr.5: 139278810-139278812 on Build GRCh38
rs747619505 Chr.5: 139279130-139279130 on Build GRCh38
rs752745872 Chr.5: 139279070-139279070 on Build GRCh38
rs762547988 Chr.5: 139278936-139278936 on Build GRCh38
rs750071501 Chr.5: 139278972-139278972 on Build GRCh38
rs371621649 Chr.5: 139278968-139278968 on Build GRCh38
rs956462534 Chr.5: 139278984-139278984 on Build GRCh38
rs754899989 Chr.5: 139279044-139279044 on Build GRCh38
rs750285543 Chr.5: 139278890-139278890 on Build GRCh38
rs770731715 Chr.5: 139278920-139278920 on Build GRCh38
rs763609314 Chr.5: 139278745-139278745 on Build GRCh38
rs762458094 Chr.5: 139278819-139278819 on Build GRCh38
rs754114812 Chr.5: 139278872-139278872 on Build GRCh38
rs764775079 Chr.5: 139279110-139279110 on Build GRCh38
rs527641003 Chr.5: 139278976-139278976 on Build GRCh38
rs756181852 Chr.5: 139279071-139279071 on Build GRCh38
rs576214782 Chr.5: 139278747-139278747 on Build GRCh38
rs761217079 Chr.5: 139278965-139278965 on Build GRCh38
rs771970098 Chr.5: 139278795-139278795 on Build GRCh38
rs924506473 Chr.5: 139278909-139278909 on Build GRCh38
rs545646962 Chr.5: 139278789-139278789 on Build GRCh38
rs998325217 Chr.5: 139278798-139278798 on Build GRCh38
rs752127962 Chr.5: 139278735-139278735 on Build GRCh38
rs561044139 Chr.5: 139279105-139279105 on Build GRCh38
rs934165385 Chr.5: 139279146-139279146 on Build GRCh38
rs756462888 Chr.5: 139278752-139278752 on Build GRCh38
rs770641799 Chr.5: 139278802-139278802 on Build GRCh38
rs776178573 Chr.5: 139279115-139279115 on Build GRCh38
rs1051248367 Chr.5: 139279160-139279160 on Build GRCh38
rs921492269 Chr.5: 139279140-139279140 on Build GRCh38
rs769374661 Chr.5: 139278812-139278812 on Build GRCh38
rs576243316 Chr.5: 139278738-139278738 on Build GRCh38
rs766595666 Chr.5: 139278851-139278851 on Build GRCh38
rs746797159 Chr.5: 139278801-139278801 on Build GRCh38
rs773870198 Chr.5: 139278954-139278954 on Build GRCh38
rs755118024 Chr.5: 139278873-139278873 on Build GRCh38
rs757002054 Chr.5: 139278851-139278852 on Build GRCh38
rs947007268 Chr.5: 139279083-139279083 on Build GRCh38
rs775122347 Chr.5: 139278931-139278931 on Build GRCh38
rs780319113 Chr.5: 139278758-139278758 on Build GRCh38
rs537108216 Chr.5: 139279101-139279101 on Build GRCh38
rs769307549 Chr.5: 139278925-139278925 on Build GRCh38
rs766096563 Chr.5: 139278996-139278996 on Build GRCh38
rs780005095 Chr.5: 139279011-139279012 on Build GRCh38
rs1051621824 Chr.5: 139278814-139278814 on Build GRCh38
rs777688671 Chr.5: 139278904-139278904 on Build GRCh38
rs778810762 Chr.5: 139278786-139278786 on Build GRCh38
rs768124888 Chr.5: 139278822-139278822 on Build GRCh38
rs975638202 Chr.5: 139279129-139279129 on Build GRCh38
rs751426767 Chr.5: 139279016-139279016 on Build GRCh38
rs866283362 Chr.5: 139278827-139278827 on Build GRCh38
rs182932144 Chr.5: 139279117-139279117 on Build GRCh38
rs773043476 Chr.5: 139278796-139278796 on Build GRCh38
rs761125546 Chr.5: 139278835-139278835 on Build GRCh38
rs772683636 Chr.5: 139279114-139279114 on Build GRCh38
rs750885677 Chr.5: 139278832-139278832 on Build GRCh38
rs771261809 Chr.5: 139279088-139279088 on Build GRCh38
rs768214996 Chr.5: 139278944-139278944 on Build GRCh38
rs189174220 Chr.5: 139278751-139278751 on Build GRCh38
rs191909504 Chr.5: 139278899-139278899 on Build GRCh38
rs1039696718 Chr.5: 139279066-139279066 on Build GRCh38
rs779093065 Chr.5: 139278879-139278879 on Build GRCh38
rs991173993 Chr.5: 139278739-139278739 on Build GRCh38
rs766800308 Chr.5: 139278966-139278966 on Build GRCh38
rs757797235 Chr.5: 139278744-139278744 on Build GRCh38
rs745898033 Chr.5: 139279089-139279089 on Build GRCh38
rs373293227 Chr.5: 139278921-139278921 on Build GRCh38
rs942737350 Chr.5: 139279054-139279054 on Build GRCh38

View Full Product Details