Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 140547576-140548070 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKHD1-EIF4EBP3
Entrez Gene ID
404734
Gene Name
ANKHD1-EIF4EBP3 readthrough
Gene Aliases
MASK-BP3, MASK-BP3ARF
UniGene
Hs.594084
Gene Chromosome Location
Chr.5: 140401814-140549578 on Build GRCh38
Transcripts
NM_020690.5 NM_003732.2

Gene Symbol
EIF4EBP3
Entrez Gene ID
8637
Gene Name
eukaryotic translation initiation factor 4E binding protein 3
Gene Aliases
4E-BP3, 4EBP3
UniGene
Hs.594084
Gene Chromosome Location
Chr.5: 140547666-140549578 on Build GRCh38
Transcripts
NM_020690.5 NM_003732.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4966577 c.43C>T p.Q15* Chr.5: 140547780-140547780 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs117892438 Chr.5: 140547947-140547947 on Build GRCh38
rs543925107 Chr.5: 140547641-140547641 on Build GRCh38
rs971447493 Chr.5: 140547869-140547869 on Build GRCh38
rs958976199 Chr.5: 140547946-140547946 on Build GRCh38
rs779561737 Chr.5: 140547790-140547790 on Build GRCh38
rs1005886719 Chr.5: 140547767-140547767 on Build GRCh38
rs924864764 Chr.5: 140547889-140547889 on Build GRCh38
rs1008149949 Chr.5: 140547719-140547734 on Build GRCh38
rs1047667714 Chr.5: 140547709-140547709 on Build GRCh38
rs748460833 Chr.5: 140547835-140547835 on Build GRCh38
rs937592211 Chr.5: 140547679-140547680 on Build GRCh38
rs1006106334 Chr.5: 140548029-140548029 on Build GRCh38
rs186301886 Chr.5: 140547786-140547786 on Build GRCh38
rs934645879 Chr.5: 140547632-140547632 on Build GRCh38
rs1020892573 Chr.5: 140547621-140547623 on Build GRCh38
rs971499810 Chr.5: 140547634-140547634 on Build GRCh38
rs1024711995 Chr.5: 140548044-140548044 on Build GRCh38
rs778095721 Chr.5: 140547998-140547998 on Build GRCh38
rs1006800770 Chr.5: 140547798-140547798 on Build GRCh38
rs768037170 Chr.5: 140547700-140547700 on Build GRCh38
rs369934971 Chr.5: 140548014-140548014 on Build GRCh38
rs923127529 Chr.5: 140548009-140548009 on Build GRCh38
rs778357350 Chr.5: 140547743-140547743 on Build GRCh38
rs981684955 Chr.5: 140547887-140547887 on Build GRCh38
rs886128648 Chr.5: 140547738-140547738 on Build GRCh38
rs1054511151 Chr.5: 140547633-140547633 on Build GRCh38
rs1016052711 Chr.5: 140547791-140547791 on Build GRCh38
rs200051899 Chr.5: 140547817-140547817 on Build GRCh38
rs990117524 Chr.5: 140547943-140547943 on Build GRCh38
rs990468922 Chr.5: 140547679-140547679 on Build GRCh38
rs1036758296 Chr.5: 140547728-140547728 on Build GRCh38
rs753147013 Chr.5: 140547711-140547711 on Build GRCh38
rs886346445 Chr.5: 140548021-140548021 on Build GRCh38
rs985030466 Chr.5: 140547965-140547965 on Build GRCh38
rs570202628 Chr.5: 140548042-140548042 on Build GRCh38
rs971559251 Chr.5: 140547877-140547877 on Build GRCh38
rs529394797 Chr.5: 140547991-140547991 on Build GRCh38
rs1047714768 Chr.5: 140548018-140548018 on Build GRCh38
rs909536381 Chr.5: 140547714-140547714 on Build GRCh38
rs1037776549 Chr.5: 140548033-140548033 on Build GRCh38
rs1012422156 Chr.5: 140548038-140548038 on Build GRCh38
rs781551674 Chr.5: 140547772-140547772 on Build GRCh38
rs1050906174 Chr.5: 140547750-140547750 on Build GRCh38
rs959388671 Chr.5: 140547800-140547800 on Build GRCh38
rs956075131 Chr.5: 140547916-140547916 on Build GRCh38
rs75475787 Chr.5: 140547830-140547830 on Build GRCh38
rs918417034 Chr.5: 140547731-140547731 on Build GRCh38
rs1039284179 Chr.5: 140547665-140547665 on Build GRCh38
rs532829787 Chr.5: 140547666-140547666 on Build GRCh38
rs35165892 Chr.5: 140547683-140547683 on Build GRCh38
rs749837015 Chr.5: 140547745-140547745 on Build GRCh38
rs901655936 Chr.5: 140547675-140547675 on Build GRCh38
rs1020335138 Chr.5: 140547867-140547867 on Build GRCh38
rs1054879786 Chr.5: 140547816-140547816 on Build GRCh38
rs1050945887 Chr.5: 140548022-140548022 on Build GRCh38
rs116513628 Chr.5: 140547851-140547851 on Build GRCh38
rs568288661 Chr.5: 140547873-140547873 on Build GRCh38
rs943433491 Chr.5: 140547644-140547644 on Build GRCh38
rs573800504 Chr.5: 140547615-140547615 on Build GRCh38
rs914542165 Chr.5: 140547944-140547944 on Build GRCh38
rs894786906 Chr.5: 140548034-140548034 on Build GRCh38
rs1034463114 Chr.5: 140547880-140547880 on Build GRCh38
rs544983486 Chr.5: 140547734-140547734 on Build GRCh38
rs181969062 Chr.5: 140547643-140547643 on Build GRCh38
rs924627011 Chr.5: 140547620-140547620 on Build GRCh38
rs997858005 Chr.5: 140547688-140547688 on Build GRCh38
rs933777214 Chr.5: 140547746-140547746 on Build GRCh38
rs372216475 Chr.5: 140547774-140547774 on Build GRCh38
rs957717773 Chr.5: 140547609-140547609 on Build GRCh38
rs909556624 Chr.5: 140547988-140547988 on Build GRCh38

View Full Product Details