Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 141174756-141175281 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCDHB7
Entrez Gene ID
56129
Gene Name
protocadherin beta 7
Gene Aliases
PCDH-BETA7
UniGene
Hs.203830
Gene Chromosome Location
Chr.5: 141172621-141176376 on Build GRCh38
Transcripts
NM_018940.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7382909 c.2037C>A p.D679E Chr.5: 141174872-141174872 on Build GRCh38
4569762 c.1999T>G p.S667A Chr.5: 141174834-141174834 on Build GRCh38
1495946 c.2304T>A p.I768I Chr.5: 141175139-141175139 on Build GRCh38
3851473 c.2015G>A p.R672Q Chr.5: 141174850-141174850 on Build GRCh38
5796807 c.2078C>T p.A693V Chr.5: 141174913-141174913 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781841171 Chr.5: 141174959-141174959 on Build GRCh38
rs782436379 Chr.5: 141174994-141174995 on Build GRCh38
rs782726150 Chr.5: 141175059-141175059 on Build GRCh38
rs782583722 Chr.5: 141175102-141175102 on Build GRCh38
rs782206091 Chr.5: 141175236-141175236 on Build GRCh38
rs532026348 Chr.5: 141175190-141175190 on Build GRCh38
rs782057144 Chr.5: 141175181-141175181 on Build GRCh38
rs782482240 Chr.5: 141175149-141175149 on Build GRCh38
rs782757345 Chr.5: 141174896-141174896 on Build GRCh38
rs2910314 Chr.5: 141174981-141174981 on Build GRCh38
rs782453182 Chr.5: 141175091-141175091 on Build GRCh38
rs782687028 Chr.5: 141175155-141175155 on Build GRCh38
rs781871908 Chr.5: 141174913-141174913 on Build GRCh38
rs782758323 Chr.5: 141174780-141174780 on Build GRCh38
rs781932710 Chr.5: 141175109-141175109 on Build GRCh38
rs80165947 Chr.5: 141174985-141174985 on Build GRCh38
rs781837484 Chr.5: 141174897-141174897 on Build GRCh38
rs782105017 Chr.5: 141175193-141175193 on Build GRCh38
rs374878755 Chr.5: 141175050-141175050 on Build GRCh38
rs782733977 Chr.5: 141174806-141174806 on Build GRCh38
rs782567006 Chr.5: 141174827-141174827 on Build GRCh38
rs782337386 Chr.5: 141174942-141174942 on Build GRCh38
rs1811236 Chr.5: 141174903-141174903 on Build GRCh38
rs781817792 Chr.5: 141174867-141174867 on Build GRCh38
rs782200287 Chr.5: 141175049-141175049 on Build GRCh38
rs781832555 Chr.5: 141174958-141174958 on Build GRCh38
rs781816876 Chr.5: 141175206-141175206 on Build GRCh38
rs782469503 Chr.5: 141174869-141174869 on Build GRCh38
rs751461245 Chr.5: 141174938-141174938 on Build GRCh38
rs781889077 Chr.5: 141174813-141174813 on Build GRCh38
rs556010646 Chr.5: 141175162-141175162 on Build GRCh38
rs782592274 Chr.5: 141174787-141174787 on Build GRCh38
rs782414303 Chr.5: 141174879-141174879 on Build GRCh38
rs552497579 Chr.5: 141175030-141175030 on Build GRCh38
rs782319816 Chr.5: 141174849-141174849 on Build GRCh38
rs782137017 Chr.5: 141174958-141174958 on Build GRCh38
rs148077207 Chr.5: 141174992-141174992 on Build GRCh38
rs782635712 Chr.5: 141174832-141174832 on Build GRCh38
rs146407243 Chr.5: 141175218-141175218 on Build GRCh38
rs557284400 Chr.5: 141175118-141175118 on Build GRCh38
rs770524511 Chr.5: 141175149-141175150 on Build GRCh38
rs782370226 Chr.5: 141175034-141175034 on Build GRCh38
rs782412716 Chr.5: 141175113-141175113 on Build GRCh38
rs782506638 Chr.5: 141174914-141174914 on Build GRCh38
rs782277575 Chr.5: 141175157-141175157 on Build GRCh38
rs200672237 Chr.5: 141174891-141174891 on Build GRCh38
rs782101940 Chr.5: 141174854-141174854 on Build GRCh38
rs146189351 Chr.5: 141174974-141174974 on Build GRCh38
rs782262598 Chr.5: 141174794-141174794 on Build GRCh38
rs781830000 Chr.5: 141175088-141175088 on Build GRCh38
rs781843762 Chr.5: 141175127-141175127 on Build GRCh38
rs782052131 Chr.5: 141174952-141174952 on Build GRCh38
rs782219472 Chr.5: 141174922-141174922 on Build GRCh38
rs782798616 Chr.5: 141175064-141175064 on Build GRCh38
rs782372428 Chr.5: 141175252-141175252 on Build GRCh38
rs781962988 Chr.5: 141175055-141175055 on Build GRCh38
rs782652788 Chr.5: 141175029-141175029 on Build GRCh38
rs782497916 Chr.5: 141174865-141174865 on Build GRCh38
rs782651484 Chr.5: 141174880-141174880 on Build GRCh38
rs782522165 Chr.5: 141174965-141174965 on Build GRCh38
rs781863227 Chr.5: 141175006-141175006 on Build GRCh38
rs11741863 Chr.5: 141175025-141175025 on Build GRCh38
rs139833816 Chr.5: 141174866-141174866 on Build GRCh38
rs201836983 Chr.5: 141174789-141174789 on Build GRCh38
rs782455599 Chr.5: 141175215-141175215 on Build GRCh38
rs17844474 Chr.5: 141174800-141174800 on Build GRCh38
rs782094357 Chr.5: 141174941-141174941 on Build GRCh38
rs782298217 Chr.5: 141175024-141175024 on Build GRCh38
rs782583150 Chr.5: 141174997-141174998 on Build GRCh38
rs142434001 Chr.5: 141175054-141175054 on Build GRCh38
rs782734599 Chr.5: 141175134-141175134 on Build GRCh38
rs782350099 Chr.5: 141174839-141174839 on Build GRCh38
rs548320013 Chr.5: 141175107-141175107 on Build GRCh38
rs782568457 Chr.5: 141175138-141175138 on Build GRCh38
rs782385262 Chr.5: 141174937-141174937 on Build GRCh38
rs782564883 Chr.5: 141175019-141175019 on Build GRCh38
rs781787452 Chr.5: 141175060-141175060 on Build GRCh38
rs782279641 Chr.5: 141175241-141175241 on Build GRCh38
rs782304667 Chr.5: 141175152-141175152 on Build GRCh38
rs781891969 Chr.5: 141174955-141174955 on Build GRCh38
rs782388523 Chr.5: 141174801-141174801 on Build GRCh38
rs782505189 Chr.5: 141175007-141175007 on Build GRCh38
rs568290690 Chr.5: 141175108-141175108 on Build GRCh38
rs782351245 Chr.5: 141175173-141175173 on Build GRCh38
rs781943105 Chr.5: 141174810-141174810 on Build GRCh38
rs782806667 Chr.5: 141175187-141175187 on Build GRCh38
rs782643775 Chr.5: 141175224-141175224 on Build GRCh38
rs782125129 Chr.5: 141174892-141174892 on Build GRCh38
rs781986819 Chr.5: 141174852-141174852 on Build GRCh38
rs781958963 Chr.5: 141174840-141174840 on Build GRCh38
rs368678683 Chr.5: 141174990-141174990 on Build GRCh38
rs781795378 Chr.5: 141175137-141175137 on Build GRCh38
rs748894525 Chr.5: 141175077-141175077 on Build GRCh38
rs142123060 Chr.5: 141174969-141174969 on Build GRCh38
rs782092408 Chr.5: 141175130-141175130 on Build GRCh38
rs5871767 Chr.5: 141174858-141174858 on Build GRCh38
rs782550160 Chr.5: 141174908-141174908 on Build GRCh38
rs111865664 Chr.5: 141174788-141174788 on Build GRCh38
rs782121921 Chr.5: 141174777-141174777 on Build GRCh38
rs558696629 Chr.5: 141174805-141174805 on Build GRCh38
rs142134051 Chr.5: 141175188-141175188 on Build GRCh38
rs782697359 Chr.5: 141175217-141175217 on Build GRCh38
rs782240512 Chr.5: 141175031-141175031 on Build GRCh38
rs782505611 Chr.5: 141174888-141174889 on Build GRCh38
rs782480763 Chr.5: 141175230-141175230 on Build GRCh38
rs782717515 Chr.5: 141174964-141174964 on Build GRCh38
rs782637685 Chr.5: 141175143-141175143 on Build GRCh38
rs774018894 Chr.5: 141175151-141175151 on Build GRCh38
rs782603723 Chr.5: 141175233-141175233 on Build GRCh38
rs782058168 Chr.5: 141174811-141174811 on Build GRCh38
rs782043784 Chr.5: 141174850-141174850 on Build GRCh38
rs781840659 Chr.5: 141174823-141174823 on Build GRCh38
rs782038638 Chr.5: 141174978-141174978 on Build GRCh38
rs782528648 Chr.5: 141174828-141174828 on Build GRCh38
rs199927796 Chr.5: 141174884-141174884 on Build GRCh38
rs782078108 Chr.5: 141175058-141175058 on Build GRCh38
rs11750810 Chr.5: 141174936-141174936 on Build GRCh38
rs565948427 Chr.5: 141175033-141175033 on Build GRCh38
rs781940026 Chr.5: 141175179-141175179 on Build GRCh38
rs1811237 Chr.5: 141174900-141174900 on Build GRCh38
rs782818479 Chr.5: 141174812-141174812 on Build GRCh38
rs781984836 Chr.5: 141175167-141175167 on Build GRCh38
rs372840693 Chr.5: 141174783-141174783 on Build GRCh38
rs782504225 Chr.5: 141175083-141175083 on Build GRCh38
rs782195827 Chr.5: 141174796-141174796 on Build GRCh38
rs782450304 Chr.5: 141175018-141175018 on Build GRCh38
rs782395805 Chr.5: 141175158-141175158 on Build GRCh38
rs6892507 Chr.5: 141175227-141175227 on Build GRCh38
rs781876695 Chr.5: 141175070-141175070 on Build GRCh38
rs782161386 Chr.5: 141174995-141174995 on Build GRCh38
rs782627462 Chr.5: 141175106-141175106 on Build GRCh38
rs781821710 Chr.5: 141175013-141175013 on Build GRCh38
rs781958717 Chr.5: 141174888-141174888 on Build GRCh38
rs782618323 Chr.5: 141175086-141175086 on Build GRCh38
rs782205232 Chr.5: 141174844-141174844 on Build GRCh38
rs566136085 Chr.5: 141174818-141174818 on Build GRCh38
rs781787105 Chr.5: 141174856-141174856 on Build GRCh38
rs17844479 Chr.5: 141175105-141175105 on Build GRCh38
rs782303923 Chr.5: 141174878-141174878 on Build GRCh38
rs782735782 Chr.5: 141174991-141174991 on Build GRCh38
rs572068556 Chr.5: 141174963-141174963 on Build GRCh38
rs782753147 Chr.5: 141175205-141175205 on Build GRCh38
rs150314773 Chr.5: 141174944-141174944 on Build GRCh38
rs534796001 Chr.5: 141174836-141174836 on Build GRCh38
rs782795589 Chr.5: 141174851-141174851 on Build GRCh38
rs369017861 Chr.5: 141175111-141175111 on Build GRCh38
rs373953146 Chr.5: 141175239-141175239 on Build GRCh38
rs782619147 Chr.5: 141174920-141174920 on Build GRCh38
rs559668200 Chr.5: 141174954-141174954 on Build GRCh38
rs781849487 Chr.5: 141175145-141175145 on Build GRCh38
rs782514292 Chr.5: 141174815-141174815 on Build GRCh38
rs111582678 Chr.5: 141174948-141174948 on Build GRCh38
rs375732369 Chr.5: 141174909-141174909 on Build GRCh38
rs544964950 Chr.5: 141174923-141174923 on Build GRCh38
rs782230641 Chr.5: 141175169-141175169 on Build GRCh38
rs782659022 Chr.5: 141174791-141174791 on Build GRCh38
rs782711863 Chr.5: 141174901-141174901 on Build GRCh38
rs781958096 Chr.5: 141175048-141175048 on Build GRCh38
rs782495579 Chr.5: 141174971-141174971 on Build GRCh38
rs782431968 Chr.5: 141174890-141174890 on Build GRCh38
rs782022941 Chr.5: 141175115-141175115 on Build GRCh38
rs782128416 Chr.5: 141175051-141175051 on Build GRCh38
rs554701227 Chr.5: 141174863-141174863 on Build GRCh38
rs17844475 Chr.5: 141174889-141174889 on Build GRCh38
rs782317015 Chr.5: 141175238-141175238 on Build GRCh38
rs138922913 Chr.5: 141174976-141174976 on Build GRCh38
rs141902328 Chr.5: 141174996-141174996 on Build GRCh38
rs782796386 Chr.5: 141174857-141174857 on Build GRCh38
rs143161042 Chr.5: 141174873-141174873 on Build GRCh38
rs180836755 Chr.5: 141175116-141175116 on Build GRCh38
rs782542413 Chr.5: 141175222-141175222 on Build GRCh38

View Full Product Details