Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 147906347-147906857 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C5orf46
Entrez Gene ID
389336
Gene Name
chromosome 5 open reading frame 46
Gene Aliases
SSSP1
UniGene
Hs.660038
Gene Chromosome Location
Chr.5: 147850463-147906594 on Build GRCh38
Transcripts
NM_206966.2 XM_005268446.3 XM_017009459.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4789668 c.46G>T p.V16F Chr.5: 147906456-147906456 on Build GRCh38
1198899 c.37G>T p.G13* Chr.5: 147906465-147906465 on Build GRCh38
5437812 c.52T>A p.F18I Chr.5: 147906450-147906450 on Build GRCh38
1310824 c.24G>C p.L8L Chr.5: 147906478-147906478 on Build GRCh38
3612139 c.51A>C p.L17F Chr.5: 147906451-147906451 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749511265 Chr.5: 147906413-147906413 on Build GRCh38
rs996722954 Chr.5: 147906694-147906694 on Build GRCh38
rs776341983 Chr.5: 147906527-147906527 on Build GRCh38
rs189402927 Chr.5: 147906637-147906637 on Build GRCh38
rs547955627 Chr.5: 147906559-147906559 on Build GRCh38
rs371115721 Chr.5: 147906736-147906736 on Build GRCh38
rs115301671 Chr.5: 147906574-147906574 on Build GRCh38
rs746111247 Chr.5: 147906521-147906521 on Build GRCh38
rs769959748 Chr.5: 147906401-147906401 on Build GRCh38
rs760143397 Chr.5: 147906464-147906464 on Build GRCh38
rs758407329 Chr.5: 147906433-147906433 on Build GRCh38
rs7722926 Chr.5: 147906450-147906450 on Build GRCh38
rs933317535 Chr.5: 147906781-147906781 on Build GRCh38
rs376580082 Chr.5: 147906514-147906514 on Build GRCh38
rs1052011359 Chr.5: 147906571-147906571 on Build GRCh38
rs1011348428 Chr.5: 147906683-147906683 on Build GRCh38
rs764706954 Chr.5: 147906476-147906476 on Build GRCh38
rs137948247 Chr.5: 147906798-147906798 on Build GRCh38
rs556531650 Chr.5: 147906706-147906706 on Build GRCh38
rs141343367 Chr.5: 147906437-147906437 on Build GRCh38
rs1038188817 Chr.5: 147906689-147906689 on Build GRCh38
rs773143524 Chr.5: 147906537-147906537 on Build GRCh38
rs566347493 Chr.5: 147906561-147906561 on Build GRCh38
rs774445059 Chr.5: 147906429-147906429 on Build GRCh38
rs755325643 Chr.5: 147906493-147906493 on Build GRCh38
rs767808813 Chr.5: 147906494-147906494 on Build GRCh38
rs139879345 Chr.5: 147906483-147906483 on Build GRCh38
rs1032329305 Chr.5: 147906524-147906524 on Build GRCh38
rs943628534 Chr.5: 147906812-147906812 on Build GRCh38
rs778045316 Chr.5: 147906403-147906403 on Build GRCh38
rs756745156 Chr.5: 147906504-147906504 on Build GRCh38
rs774698813 Chr.5: 147906525-147906537 on Build GRCh38
rs956266281 Chr.5: 147906773-147906773 on Build GRCh38
rs370032483 Chr.5: 147906439-147906439 on Build GRCh38
rs551386373 Chr.5: 147906643-147906643 on Build GRCh38
rs893960347 Chr.5: 147906816-147906816 on Build GRCh38
rs908425644 Chr.5: 147906661-147906661 on Build GRCh38
rs374550828 Chr.5: 147906744-147906744 on Build GRCh38
rs374415209 Chr.5: 147906545-147906545 on Build GRCh38
rs375716588 Chr.5: 147906498-147906498 on Build GRCh38
rs898347512 Chr.5: 147906693-147906693 on Build GRCh38
rs771350207 Chr.5: 147906414-147906414 on Build GRCh38
rs768470750 Chr.5: 147906522-147906522 on Build GRCh38
rs766410332 Chr.5: 147906445-147906445 on Build GRCh38
rs779151431 Chr.5: 147906516-147906516 on Build GRCh38
rs137968854 Chr.5: 147906702-147906704 on Build GRCh38
rs765836979 Chr.5: 147906551-147906551 on Build GRCh38
rs372380037 Chr.5: 147906482-147906482 on Build GRCh38
rs1053934217 Chr.5: 147906815-147906815 on Build GRCh38
rs972106821 Chr.5: 147906763-147906763 on Build GRCh38
rs748070485 Chr.5: 147906529-147906529 on Build GRCh38
rs981547701 Chr.5: 147906740-147906740 on Build GRCh38
rs919373094 Chr.5: 147906778-147906778 on Build GRCh38
rs890909775 Chr.5: 147906747-147906747 on Build GRCh38
rs115395632 Chr.5: 147906490-147906490 on Build GRCh38
rs1020069699 Chr.5: 147906695-147906695 on Build GRCh38
rs761929225 Chr.5: 147906567-147906567 on Build GRCh38
rs61742197 Chr.5: 147906512-147906512 on Build GRCh38
rs114801379 Chr.5: 147906810-147906810 on Build GRCh38
rs746765598 Chr.5: 147906431-147906431 on Build GRCh38
rs989779108 Chr.5: 147906377-147906377 on Build GRCh38
rs769649038 Chr.5: 147906536-147906536 on Build GRCh38
rs909063884 Chr.5: 147906757-147906757 on Build GRCh38
rs892860517 Chr.5: 147906679-147906679 on Build GRCh38
rs753155181 Chr.5: 147906557-147906557 on Build GRCh38
rs2250145 Chr.5: 147906491-147906491 on Build GRCh38
rs942098008 Chr.5: 147906762-147906762 on Build GRCh38
rs574808994 Chr.5: 147906707-147906707 on Build GRCh38
rs748630808 Chr.5: 147906391-147906391 on Build GRCh38
rs958876390 Chr.5: 147906746-147906746 on Build GRCh38
rs1153088 Chr.5: 147906754-147906754 on Build GRCh38
rs185635303 Chr.5: 147906597-147906597 on Build GRCh38
rs905071104 Chr.5: 147906784-147906784 on Build GRCh38
rs1047757904 Chr.5: 147906651-147906651 on Build GRCh38

View Full Product Details