Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 151033523-151034019 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TNIP1
Entrez Gene ID
10318
Gene Name
TNFAIP3 interacting protein 1
Gene Aliases
ABIN-1, NAF1, VAN, nip40-1
UniGene
Hs.355141
Gene Chromosome Location
Chr.5: 151029943-151087660 on Build GRCh38
Transcripts
NM_001252385.1 NM_001252386.1 NM_001252390.1 NM_001252391.1 NM_001252392.1 NM_001252393.1 NM_001258454.1 NM_001258455.1 NM_001258456.1 NM_006058.4 XM_005268355.1 XM_006714751.1 XM_006714752.2 XM_017008945.1 XM_017008946.1 XM_017008947.1 XM_017008948.1 XM_017008949.1 XM_017008950.1 XM_017008951.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3683798 c.1740G>C p.E580D Chr.5: 151033647-151033647 on Build GRCh38
5867896 c.1750C>T p.P584S Chr.5: 151033637-151033637 on Build GRCh38
4955333 c.1746G>A p.P582P Chr.5: 151033641-151033641 on Build GRCh38
4747982 c.1721delC p.P574fs*41 Chr.5: 151033666-151033666 on Build GRCh38
4747983 c.1721delC p.P574fs*41 Chr.5: 151033666-151033666 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752544824 Chr.5: 151033689-151033689 on Build GRCh38
rs751048169 Chr.5: 151033842-151033842 on Build GRCh38
rs763948793 Chr.5: 151033780-151033780 on Build GRCh38
rs924833995 Chr.5: 151033856-151033856 on Build GRCh38
rs62382331 Chr.5: 151033641-151033641 on Build GRCh38
rs563130838 Chr.5: 151033973-151033973 on Build GRCh38
rs999807138 Chr.5: 151033966-151033966 on Build GRCh38
rs371379662 Chr.5: 151033850-151033850 on Build GRCh38
rs775119195 Chr.5: 151033959-151033959 on Build GRCh38
rs769812772 Chr.5: 151033901-151033901 on Build GRCh38
rs139724991 Chr.5: 151033676-151033676 on Build GRCh38
rs144751861 Chr.5: 151033751-151033751 on Build GRCh38
rs888695021 Chr.5: 151033639-151033639 on Build GRCh38
rs751492571 Chr.5: 151033566-151033566 on Build GRCh38
rs762760836 Chr.5: 151033682-151033682 on Build GRCh38
rs1032710289 Chr.5: 151033742-151033742 on Build GRCh38
rs903328105 Chr.5: 151033570-151033570 on Build GRCh38
rs770392543 Chr.5: 151033610-151033610 on Build GRCh38
rs777390870 Chr.5: 151033581-151033581 on Build GRCh38
rs769246621 Chr.5: 151033737-151033737 on Build GRCh38
rs903260365 Chr.5: 151033922-151033922 on Build GRCh38
rs745753422 Chr.5: 151033894-151033894 on Build GRCh38
rs550789494 Chr.5: 151033857-151033857 on Build GRCh38
rs747829265 Chr.5: 151033576-151033576 on Build GRCh38
rs763825554 Chr.5: 151033685-151033686 on Build GRCh38
rs755914682 Chr.5: 151033577-151033577 on Build GRCh38
rs577337766 Chr.5: 151033873-151033873 on Build GRCh38
rs777580735 Chr.5: 151033654-151033654 on Build GRCh38
rs200926279 Chr.5: 151033623-151033623 on Build GRCh38
rs758021000 Chr.5: 151033838-151033838 on Build GRCh38
rs776965065 Chr.5: 151033627-151033627 on Build GRCh38
rs2233305 Chr.5: 151033845-151033845 on Build GRCh38
rs150564405 Chr.5: 151033661-151033661 on Build GRCh38
rs758508125 Chr.5: 151033634-151033634 on Build GRCh38
rs775014523 Chr.5: 151033665-151033666 on Build GRCh38
rs753587294 Chr.5: 151033702-151033702 on Build GRCh38
rs776174642 Chr.5: 151033600-151033601 on Build GRCh38
rs781222825 Chr.5: 151033574-151033574 on Build GRCh38
rs770400306 Chr.5: 151033596-151033597 on Build GRCh38
rs2233307 Chr.5: 151033569-151033569 on Build GRCh38
rs745511990 Chr.5: 151033622-151033622 on Build GRCh38
rs962560416 Chr.5: 151033760-151033760 on Build GRCh38
rs377591817 Chr.5: 151033708-151033708 on Build GRCh38
rs762176055 Chr.5: 151033628-151033628 on Build GRCh38
rs201482038 Chr.5: 151033713-151033713 on Build GRCh38
rs779604622 Chr.5: 151033840-151033840 on Build GRCh38
rs769120189 Chr.5: 151033667-151033667 on Build GRCh38
rs772415850 Chr.5: 151033622-151033622 on Build GRCh38
rs2233306 Chr.5: 151033729-151033729 on Build GRCh38
rs746540532 Chr.5: 151033736-151033736 on Build GRCh38
rs374743804 Chr.5: 151033655-151033655 on Build GRCh38
rs766547205 Chr.5: 151033687-151033687 on Build GRCh38
rs537648496 Chr.5: 151033642-151033642 on Build GRCh38
rs143590032 Chr.5: 151033658-151033658 on Build GRCh38
rs1003680695 Chr.5: 151033727-151033727 on Build GRCh38
rs754175269 Chr.5: 151033548-151033548 on Build GRCh38
rs1044753505 Chr.5: 151033906-151033906 on Build GRCh38
rs764624153 Chr.5: 151033624-151033624 on Build GRCh38
rs372807299 Chr.5: 151033630-151033630 on Build GRCh38
rs1051287240 Chr.5: 151033555-151033555 on Build GRCh38
rs745403030 Chr.5: 151033621-151033621 on Build GRCh38
rs753833491 Chr.5: 151033786-151033786 on Build GRCh38
rs117226070 Chr.5: 151033926-151033926 on Build GRCh38
rs757412381 Chr.5: 151033552-151033552 on Build GRCh38
rs139496041 Chr.5: 151033752-151033752 on Build GRCh38
rs747562444 Chr.5: 151033666-151033666 on Build GRCh38
rs764036761 Chr.5: 151033695-151033695 on Build GRCh38
rs773778622 Chr.5: 151033617-151033617 on Build GRCh38
rs368609279 Chr.5: 151033565-151033565 on Build GRCh38
rs893599855 Chr.5: 151033990-151033990 on Build GRCh38
rs374059963 Chr.5: 151033817-151033817 on Build GRCh38
rs987272580 Chr.5: 151033853-151033853 on Build GRCh38
rs563081133 Chr.5: 151033668-151033668 on Build GRCh38
rs144215516 Chr.5: 151033769-151033769 on Build GRCh38
rs750431294 Chr.5: 151033631-151033631 on Build GRCh38
rs561087579 Chr.5: 151033648-151033648 on Build GRCh38
rs760651668 Chr.5: 151033694-151033694 on Build GRCh38
rs146625793 Chr.5: 151033772-151033772 on Build GRCh38
rs904953852 Chr.5: 151033722-151033722 on Build GRCh38
rs930141592 Chr.5: 151033569-151033570 on Build GRCh38
rs537463860 Chr.5: 151033657-151033657 on Build GRCh38
rs200826999 Chr.5: 151033788-151033788 on Build GRCh38
rs778420111 Chr.5: 151033710-151033710 on Build GRCh38
rs893395964 Chr.5: 151033897-151033897 on Build GRCh38
rs572940734 Chr.5: 151033816-151033816 on Build GRCh38
rs60602178 Chr.5: 151033967-151033967 on Build GRCh38
rs759641663 Chr.5: 151033777-151033777 on Build GRCh38
rs781766260 Chr.5: 151033738-151033738 on Build GRCh38
rs779667955 Chr.5: 151033732-151033732 on Build GRCh38
rs765311858 Chr.5: 151033669-151033669 on Build GRCh38
rs749006420 Chr.5: 151033590-151033590 on Build GRCh38
rs755898310 Chr.5: 151033646-151033646 on Build GRCh38
rs763274749 Chr.5: 151033754-151033754 on Build GRCh38
rs998238204 Chr.5: 151033664-151033664 on Build GRCh38
rs933975957 Chr.5: 151033903-151033903 on Build GRCh38
rs531026678 Chr.5: 151033728-151033728 on Build GRCh38
rs769417545 Chr.5: 151033650-151033651 on Build GRCh38
rs11558276 Chr.5: 151033652-151033652 on Build GRCh38
rs1006080595 Chr.5: 151033655-151033656 on Build GRCh38
rs934926050 Chr.5: 151033599-151033599 on Build GRCh38

View Full Product Details