Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 157162633-157163135 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM71B
Entrez Gene ID
153745
Gene Name
family with sequence similarity 71 member B
Gene Aliases
-
UniGene
Hs.666099
Gene Chromosome Location
Chr.5: 157162332-157166268 on Build GRCh38
Transcripts
NM_130899.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1435521 c.1458C>T p.S486S Chr.5: 157162807-157162807 on Build GRCh38
351177 c.1623C>T p.L541L Chr.5: 157162642-157162642 on Build GRCh38
1064905 c.1485C>A p.S495S Chr.5: 157162780-157162780 on Build GRCh38
3994144 c.1258C>G p.P420A Chr.5: 157163007-157163007 on Build GRCh38
311078 c.1242C>A p.S414R Chr.5: 157163023-157163023 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs115938677 Chr.5: 157162926-157162926 on Build GRCh38
rs781591264 Chr.5: 157163029-157163029 on Build GRCh38
rs779402784 Chr.5: 157162858-157162858 on Build GRCh38
rs372740796 Chr.5: 157162803-157162803 on Build GRCh38
rs145275929 Chr.5: 157162685-157162685 on Build GRCh38
rs147642455 Chr.5: 157162717-157162717 on Build GRCh38
rs751459650 Chr.5: 157162885-157162885 on Build GRCh38
rs778909934 Chr.5: 157162694-157162694 on Build GRCh38
rs753856277 Chr.5: 157162689-157162689 on Build GRCh38
rs771992939 Chr.5: 157163043-157163043 on Build GRCh38
rs775886064 Chr.5: 157162862-157162862 on Build GRCh38
rs780510911 Chr.5: 157162938-157162952 on Build GRCh38
rs764772861 Chr.5: 157162988-157162988 on Build GRCh38
rs752061588 Chr.5: 157163108-157163108 on Build GRCh38
rs375898947 Chr.5: 157162963-157162963 on Build GRCh38
rs866067615 Chr.5: 157163110-157163110 on Build GRCh38
rs780817844 Chr.5: 157162785-157162785 on Build GRCh38
rs763835991 Chr.5: 157163062-157163062 on Build GRCh38
rs868213784 Chr.5: 157162781-157162781 on Build GRCh38
rs140911228 Chr.5: 157162890-157162890 on Build GRCh38
rs559552263 Chr.5: 157162775-157162775 on Build GRCh38
rs529901585 Chr.5: 157162898-157162898 on Build GRCh38
rs145584571 Chr.5: 157162804-157162804 on Build GRCh38
rs201052186 Chr.5: 157162860-157162860 on Build GRCh38
rs370122169 Chr.5: 157162738-157162738 on Build GRCh38
rs749843966 Chr.5: 157162855-157162855 on Build GRCh38
rs764710657 Chr.5: 157163067-157163067 on Build GRCh38
rs374886824 Chr.5: 157162786-157162786 on Build GRCh38
rs138283603 Chr.5: 157163061-157163061 on Build GRCh38
rs753479388 Chr.5: 157162955-157162955 on Build GRCh38
rs375482078 Chr.5: 157162801-157162801 on Build GRCh38
rs768903129 Chr.5: 157162868-157162868 on Build GRCh38
rs534409939 Chr.5: 157162769-157162769 on Build GRCh38
rs1020550018 Chr.5: 157163018-157163018 on Build GRCh38
rs369862230 Chr.5: 157162894-157162894 on Build GRCh38
rs200703166 Chr.5: 157163034-157163034 on Build GRCh38
rs751677405 Chr.5: 157162668-157162668 on Build GRCh38
rs201158406 Chr.5: 157162919-157162919 on Build GRCh38
rs950130332 Chr.5: 157162739-157162739 on Build GRCh38
rs749941931 Chr.5: 157163081-157163081 on Build GRCh38
rs201677910 Chr.5: 157162874-157162874 on Build GRCh38
rs780628441 Chr.5: 157162998-157162998 on Build GRCh38
rs754625757 Chr.5: 157162851-157162851 on Build GRCh38
rs975055330 Chr.5: 157162846-157162846 on Build GRCh38
rs937291183 Chr.5: 157162975-157162975 on Build GRCh38
rs771331137 Chr.5: 157162857-157162857 on Build GRCh38
rs367950062 Chr.5: 157162796-157162796 on Build GRCh38
rs767349540 Chr.5: 157162937-157162937 on Build GRCh38
rs754739653 Chr.5: 157162838-157162838 on Build GRCh38
rs141593979 Chr.5: 157162995-157162995 on Build GRCh38
rs758595119 Chr.5: 157162982-157162983 on Build GRCh38
rs763324021 Chr.5: 157162744-157162744 on Build GRCh38
rs766607619 Chr.5: 157162753-157162753 on Build GRCh38
rs374433668 Chr.5: 157162749-157162749 on Build GRCh38
rs368629528 Chr.5: 157162849-157162849 on Build GRCh38
rs964885906 Chr.5: 157163085-157163085 on Build GRCh38
rs966410657 Chr.5: 157162806-157162806 on Build GRCh38
rs766597102 Chr.5: 157162660-157162660 on Build GRCh38
rs35758261 Chr.5: 157162837-157162837 on Build GRCh38
rs1027918459 Chr.5: 157163087-157163087 on Build GRCh38
rs893412817 Chr.5: 157162968-157162968 on Build GRCh38
rs769240870 Chr.5: 157162790-157162790 on Build GRCh38
rs775977622 Chr.5: 157162718-157162718 on Build GRCh38
rs1033306473 Chr.5: 157162787-157162787 on Build GRCh38
rs752706329 Chr.5: 157162761-157162761 on Build GRCh38
rs745546229 Chr.5: 157162692-157162692 on Build GRCh38
rs765964858 Chr.5: 157162772-157162772 on Build GRCh38
rs150872087 Chr.5: 157163053-157163053 on Build GRCh38
rs756195281 Chr.5: 157162766-157162766 on Build GRCh38
rs766229012 Chr.5: 157162879-157162879 on Build GRCh38
rs758729717 Chr.5: 157162900-157162900 on Build GRCh38
rs149262421 Chr.5: 157163086-157163086 on Build GRCh38
rs755742422 Chr.5: 157162893-157162893 on Build GRCh38
rs752804834 Chr.5: 157162776-157162777 on Build GRCh38
rs148949971 Chr.5: 157162827-157162827 on Build GRCh38
rs769872026 Chr.5: 157162924-157162924 on Build GRCh38
rs765278429 Chr.5: 157162767-157162767 on Build GRCh38
rs752390382 Chr.5: 157162892-157162892 on Build GRCh38
rs745851698 Chr.5: 157163042-157163042 on Build GRCh38
rs35703155 Chr.5: 157162978-157162979 on Build GRCh38
rs780725075 Chr.5: 157162847-157162847 on Build GRCh38
rs372173223 Chr.5: 157162797-157162797 on Build GRCh38
rs930692996 Chr.5: 157162661-157162661 on Build GRCh38
rs767685435 Chr.5: 157162758-157162758 on Build GRCh38
rs745662390 Chr.5: 157162703-157162703 on Build GRCh38
rs748364201 Chr.5: 157163033-157163033 on Build GRCh38
rs759273697 Chr.5: 157162887-157162887 on Build GRCh38
rs767004419 Chr.5: 157163099-157163099 on Build GRCh38
rs758298028 Chr.5: 157162774-157162774 on Build GRCh38
rs778276804 Chr.5: 157162791-157162791 on Build GRCh38
rs368867239 Chr.5: 157162873-157162873 on Build GRCh38
rs774666300 Chr.5: 157162754-157162754 on Build GRCh38
rs531234661 Chr.5: 157163063-157163063 on Build GRCh38
rs775252520 Chr.5: 157162944-157162944 on Build GRCh38
rs886544549 Chr.5: 157162736-157162736 on Build GRCh38
rs746344491 Chr.5: 157162899-157162899 on Build GRCh38
rs200655776 Chr.5: 157163064-157163064 on Build GRCh38
rs768991205 Chr.5: 157162910-157162910 on Build GRCh38
rs748193511 Chr.5: 157162921-157162921 on Build GRCh38
rs755333214 Chr.5: 157163015-157163015 on Build GRCh38
rs762773856 Chr.5: 157162828-157162828 on Build GRCh38
rs779678953 Chr.5: 157162776-157162776 on Build GRCh38
rs777235522 Chr.5: 157162852-157162852 on Build GRCh38
rs201705403 Chr.5: 157162863-157162863 on Build GRCh38
rs759647623 Chr.5: 157162756-157162756 on Build GRCh38
rs758210021 Chr.5: 157162714-157162714 on Build GRCh38
rs747380312 Chr.5: 157163012-157163012 on Build GRCh38
rs747288663 Chr.5: 157162906-157162906 on Build GRCh38
rs1055104938 Chr.5: 157162949-157162949 on Build GRCh38
rs548135799 Chr.5: 157163071-157163071 on Build GRCh38
rs763232495 Chr.5: 157162654-157162654 on Build GRCh38
rs754569329 Chr.5: 157162783-157162783 on Build GRCh38
rs748708135 Chr.5: 157162724-157162724 on Build GRCh38
rs763011132 Chr.5: 157162877-157162877 on Build GRCh38
rs766250626 Chr.5: 157162831-157162831 on Build GRCh38
rs144873852 Chr.5: 157162925-157162925 on Build GRCh38
rs764229512 Chr.5: 157162821-157162821 on Build GRCh38
rs149717280 Chr.5: 157162798-157162798 on Build GRCh38
rs184587958 Chr.5: 157162904-157162904 on Build GRCh38
rs372413919 Chr.5: 157162950-157162950 on Build GRCh38
rs779776498 Chr.5: 157162715-157162715 on Build GRCh38
rs755110939 Chr.5: 157162683-157162683 on Build GRCh38
rs201767660 Chr.5: 157162974-157162974 on Build GRCh38
rs758956625 Chr.5: 157162996-157162996 on Build GRCh38
rs749935001 Chr.5: 157162768-157162768 on Build GRCh38
rs561046616 Chr.5: 157162895-157162895 on Build GRCh38

View Full Product Details