Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 157803496-157804011 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLINT1
Entrez Gene ID
9685
Gene Name
clathrin interactor 1
Gene Aliases
CLINT, ENTH, EPN4, EPNR
UniGene
Hs.644000
Gene Chromosome Location
Chr.5: 157785743-157859175 on Build GRCh38
Transcripts
NM_001195555.1 NM_001195556.1 NM_014666.3 XM_011534701.2 XM_017010087.1 XM_017010088.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7361532 c.937G>C p.D313H Chr.5: 157803671-157803671 on Build GRCh38
6513549 c.903C>T p.S301S Chr.5: 157803705-157803705 on Build GRCh38
6193152 c.949C>T p.Q317* Chr.5: 157803659-157803659 on Build GRCh38
6900667 c.971G>A p.G324D Chr.5: 157803691-157803691 on Build GRCh38
6380914 c.1005G>T p.Q335H Chr.5: 157803657-157803657 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377420226 Chr.5: 157803636-157803636 on Build GRCh38
rs572222486 Chr.5: 157803639-157803639 on Build GRCh38
rs916096488 Chr.5: 157803625-157803625 on Build GRCh38
rs747594174 Chr.5: 157803695-157803695 on Build GRCh38
rs762616691 Chr.5: 157803635-157803635 on Build GRCh38
rs948122526 Chr.5: 157803592-157803592 on Build GRCh38
rs935372254 Chr.5: 157803914-157803914 on Build GRCh38
rs761934597 Chr.5: 157803749-157803749 on Build GRCh38
rs759510048 Chr.5: 157803654-157803654 on Build GRCh38
rs751937348 Chr.5: 157803675-157803675 on Build GRCh38
rs374407849 Chr.5: 157803969-157803969 on Build GRCh38
rs538558785 Chr.5: 157803762-157803762 on Build GRCh38
rs960139906 Chr.5: 157803978-157803978 on Build GRCh38
rs1016870068 Chr.5: 157803852-157803853 on Build GRCh38
rs1026020267 Chr.5: 157803798-157803798 on Build GRCh38
rs1023493027 Chr.5: 157803852-157803852 on Build GRCh38
rs777358337 Chr.5: 157803709-157803709 on Build GRCh38
rs745620751 Chr.5: 157803612-157803613 on Build GRCh38
rs992038809 Chr.5: 157803838-157803838 on Build GRCh38
rs1050611333 Chr.5: 157803945-157803945 on Build GRCh38
rs979542176 Chr.5: 157803894-157803894 on Build GRCh38
rs578101637 Chr.5: 157803826-157803826 on Build GRCh38
rs371124871 Chr.5: 157803735-157803735 on Build GRCh38
rs574205216 Chr.5: 157803940-157803940 on Build GRCh38
rs765384333 Chr.5: 157803750-157803750 on Build GRCh38
rs554094686 Chr.5: 157803565-157803565 on Build GRCh38
rs186195219 Chr.5: 157803941-157803941 on Build GRCh38
rs769379925 Chr.5: 157803632-157803632 on Build GRCh38
rs750695619 Chr.5: 157803670-157803670 on Build GRCh38
rs774178976 Chr.5: 157803718-157803718 on Build GRCh38
rs181660288 Chr.5: 157803772-157803772 on Build GRCh38
rs770552517 Chr.5: 157803716-157803716 on Build GRCh38
rs925290368 Chr.5: 157803638-157803638 on Build GRCh38
rs758599125 Chr.5: 157803671-157803671 on Build GRCh38
rs767617456 Chr.5: 157803663-157803663 on Build GRCh38
rs991989443 Chr.5: 157803568-157803568 on Build GRCh38
rs761988038 Chr.5: 157803667-157803667 on Build GRCh38
rs563801532 Chr.5: 157803875-157803875 on Build GRCh38
rs772067911 Chr.5: 157803745-157803745 on Build GRCh38
rs919345953 Chr.5: 157803573-157803573 on Build GRCh38
rs189499790 Chr.5: 157803942-157803942 on Build GRCh38
rs368857753 Chr.5: 157803834-157803834 on Build GRCh38
rs925374865 Chr.5: 157803912-157803912 on Build GRCh38
rs942189668 Chr.5: 157803970-157803970 on Build GRCh38
rs776790372 Chr.5: 157803665-157803665 on Build GRCh38
rs765515647 Chr.5: 157803669-157803669 on Build GRCh38
rs755656419 Chr.5: 157803698-157803698 on Build GRCh38
rs772856506 Chr.5: 157803633-157803633 on Build GRCh38
rs747718418 Chr.5: 157803602-157803602 on Build GRCh38
rs889657256 Chr.5: 157803778-157803778 on Build GRCh38
rs746961576 Chr.5: 157803922-157803922 on Build GRCh38
rs188926451 Chr.5: 157803757-157803757 on Build GRCh38
rs889294012 Chr.5: 157803959-157803959 on Build GRCh38
rs938827526 Chr.5: 157803589-157803589 on Build GRCh38
rs993026729 Chr.5: 157803598-157803598 on Build GRCh38
rs756107983 Chr.5: 157803751-157803752 on Build GRCh38
rs916384984 Chr.5: 157803590-157803590 on Build GRCh38
rs960846090 Chr.5: 157803530-157803530 on Build GRCh38
rs775599754 Chr.5: 157803746-157803746 on Build GRCh38
rs902092826 Chr.5: 157803682-157803682 on Build GRCh38
rs748936606 Chr.5: 157803713-157803713 on Build GRCh38
rs766737026 Chr.5: 157803672-157803672 on Build GRCh38
rs2270810 Chr.5: 157803524-157803524 on Build GRCh38
rs933526917 Chr.5: 157803687-157803687 on Build GRCh38
rs779807549 Chr.5: 157803755-157803758 on Build GRCh38
rs552907130 Chr.5: 157803685-157803685 on Build GRCh38

View Full Product Details