Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 163467460-163467959 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HMMR
Entrez Gene ID
3161
Gene Name
hyaluronan mediated motility receptor
Gene Aliases
CD168, IHABP, RHAMM
UniGene
Hs.740467
Gene Chromosome Location
Chr.5: 163460511-163491945 on Build GRCh38
Transcripts
NM_001142556.1 NM_001142557.1 NM_012484.2 NM_012485.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5795529 c.264G>C p.E88D Chr.5: 163467742-163467742 on Build GRCh38
1065368 c.247G>T p.D83Y Chr.5: 163467725-163467725 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs755740711 Chr.5: 163467756-163467756 on Build GRCh38
rs146702035 Chr.5: 163467560-163467560 on Build GRCh38
rs561052191 Chr.5: 163467685-163467685 on Build GRCh38
rs202024983 Chr.5: 163467689-163467689 on Build GRCh38
rs148145643 Chr.5: 163467713-163467713 on Build GRCh38
rs753736591 Chr.5: 163467794-163467794 on Build GRCh38
rs1040636650 Chr.5: 163467518-163467518 on Build GRCh38
rs768701385 Chr.5: 163467662-163467662 on Build GRCh38
rs370065820 Chr.5: 163467561-163467561 on Build GRCh38
rs960335142 Chr.5: 163467830-163467830 on Build GRCh38
rs777436705 Chr.5: 163467786-163467786 on Build GRCh38
rs188930251 Chr.5: 163467577-163467577 on Build GRCh38
rs929668298 Chr.5: 163467919-163467919 on Build GRCh38
rs373753736 Chr.5: 163467676-163467676 on Build GRCh38
rs1030014896 Chr.5: 163467630-163467630 on Build GRCh38
rs946021726 Chr.5: 163467849-163467849 on Build GRCh38
rs1047132308 Chr.5: 163467920-163467920 on Build GRCh38
rs944838002 Chr.5: 163467874-163467874 on Build GRCh38
rs774271874 Chr.5: 163467651-163467651 on Build GRCh38
rs372361681 Chr.5: 163467657-163467657 on Build GRCh38
rs535040611 Chr.5: 163467784-163467784 on Build GRCh38
rs754580127 Chr.5: 163467723-163467723 on Build GRCh38
rs767100503 Chr.5: 163467727-163467727 on Build GRCh38
rs988517256 Chr.5: 163467751-163467751 on Build GRCh38
rs772838272 Chr.5: 163467788-163467791 on Build GRCh38
rs888114693 Chr.5: 163467937-163467937 on Build GRCh38
rs942727718 Chr.5: 163467890-163467890 on Build GRCh38
rs978879566 Chr.5: 163467860-163467860 on Build GRCh38
rs551749048 Chr.5: 163467881-163467881 on Build GRCh38
rs915335596 Chr.5: 163467787-163467787 on Build GRCh38
rs6881930 Chr.5: 163467840-163467840 on Build GRCh38
rs888161787 Chr.5: 163467554-163467554 on Build GRCh38
rs994217290 Chr.5: 163467539-163467539 on Build GRCh38
rs767059677 Chr.5: 163467711-163467711 on Build GRCh38
rs1004620304 Chr.5: 163467663-163467663 on Build GRCh38
rs763372273 Chr.5: 163467704-163467704 on Build GRCh38
rs913256119 Chr.5: 163467844-163467844 on Build GRCh38
rs775479048 Chr.5: 163467705-163467705 on Build GRCh38
rs539700455 Chr.5: 163467855-163467855 on Build GRCh38
rs200952082 Chr.5: 163467795-163467795 on Build GRCh38
rs919475946 Chr.5: 163467900-163467900 on Build GRCh38
rs963554865 Chr.5: 163467740-163467740 on Build GRCh38
rs376642365 Chr.5: 163467717-163467717 on Build GRCh38
rs747794669 Chr.5: 163467783-163467784 on Build GRCh38
rs369485598 Chr.5: 163467687-163467687 on Build GRCh38
rs778762622 Chr.5: 163467767-163467767 on Build GRCh38
rs762058216 Chr.5: 163467682-163467682 on Build GRCh38
rs149681564 Chr.5: 163467550-163467550 on Build GRCh38
rs776879767 Chr.5: 163467667-163467667 on Build GRCh38
rs980243861 Chr.5: 163467780-163467780 on Build GRCh38
rs748532197 Chr.5: 163467576-163467576 on Build GRCh38
rs900865743 Chr.5: 163467532-163467532 on Build GRCh38
rs986207136 Chr.5: 163467805-163467805 on Build GRCh38
rs926029085 Chr.5: 163467877-163467877 on Build GRCh38
rs2287777 Chr.5: 163467648-163467648 on Build GRCh38
rs752321701 Chr.5: 163467754-163467754 on Build GRCh38
rs533661620 Chr.5: 163467527-163467527 on Build GRCh38
rs1023462335 Chr.5: 163467569-163467569 on Build GRCh38
rs772045545 Chr.5: 163467788-163467793 on Build GRCh38
rs768433654 Chr.5: 163467681-163467681 on Build GRCh38

View Full Product Details