Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 179023694-179024187 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF879
Entrez Gene ID
345462
Gene Name
zinc finger protein 879
Gene Aliases
DKFZp686E2433
UniGene
Hs.445740
Gene Chromosome Location
Chr.5: 179023759-179034387 on Build GRCh38
Transcripts
NM_001136116.1 XM_005265908.3 XM_011534550.2 XM_011534551.2

SNP Detail

SNP ID Chromosome Location
rs981636171 Chr.5: 179024153-179024153 on Build GRCh38
rs991238781 Chr.5: 179023780-179023780 on Build GRCh38
rs760872669 Chr.5: 179023784-179023784 on Build GRCh38
rs771072901 Chr.5: 179023845-179023845 on Build GRCh38
rs920819918 Chr.5: 179024017-179024017 on Build GRCh38
rs1018802304 Chr.5: 179023847-179023847 on Build GRCh38
rs957261980 Chr.5: 179024107-179024107 on Build GRCh38
rs1012715838 Chr.5: 179024123-179024123 on Build GRCh38
rs889219530 Chr.5: 179024032-179024032 on Build GRCh38
rs930803209 Chr.5: 179024018-179024018 on Build GRCh38
rs906797846 Chr.5: 179023811-179023811 on Build GRCh38
rs987437497 Chr.5: 179023724-179023724 on Build GRCh38
rs904124618 Chr.5: 179023763-179023763 on Build GRCh38
rs372059481 Chr.5: 179023929-179023929 on Build GRCh38
rs573702563 Chr.5: 179023889-179023889 on Build GRCh38
rs991242376 Chr.5: 179023907-179023907 on Build GRCh38
rs6859767 Chr.5: 179024106-179024106 on Build GRCh38
rs555317229 Chr.5: 179023887-179023887 on Build GRCh38
rs1023355395 Chr.5: 179024142-179024142 on Build GRCh38
rs1037053340 Chr.5: 179024013-179024013 on Build GRCh38
rs1008311202 Chr.5: 179023840-179023840 on Build GRCh38
rs146907764 Chr.5: 179023946-179023946 on Build GRCh38
rs1008922079 Chr.5: 179023856-179023856 on Build GRCh38
rs6859576 Chr.5: 179023994-179023994 on Build GRCh38
rs539690455 Chr.5: 179023791-179023791 on Build GRCh38
rs534754633 Chr.5: 179023939-179023939 on Build GRCh38
rs765493753 Chr.5: 179024005-179024005 on Build GRCh38
rs899366494 Chr.5: 179023832-179023832 on Build GRCh38
rs1052614448 Chr.5: 179023833-179023833 on Build GRCh38
rs971184175 Chr.5: 179024140-179024140 on Build GRCh38
rs935830691 Chr.5: 179023774-179023774 on Build GRCh38
rs908226726 Chr.5: 179023973-179023973 on Build GRCh38
rs1019336182 Chr.5: 179023864-179023864 on Build GRCh38
rs544909201 Chr.5: 179024070-179024070 on Build GRCh38
rs1043982152 Chr.5: 179023743-179023743 on Build GRCh38
rs1036596563 Chr.5: 179023821-179023821 on Build GRCh38
rs552030919 Chr.5: 179023825-179023825 on Build GRCh38
rs1022781382 Chr.5: 179024132-179024132 on Build GRCh38
rs565565536 Chr.5: 179023725-179023725 on Build GRCh38
rs531073372 Chr.5: 179023751-179023751 on Build GRCh38
rs1009015474 Chr.5: 179024039-179024039 on Build GRCh38
rs967679223 Chr.5: 179023759-179023759 on Build GRCh38
rs960553663 Chr.5: 179024119-179024119 on Build GRCh38
rs977766757 Chr.5: 179023882-179023882 on Build GRCh38
rs935574807 Chr.5: 179023960-179023960 on Build GRCh38
rs889168705 Chr.5: 179023853-179023853 on Build GRCh38
rs1050563021 Chr.5: 179024022-179024022 on Build GRCh38
rs1015283426 Chr.5: 179024116-179024116 on Build GRCh38
rs998860184 Chr.5: 179024105-179024105 on Build GRCh38
rs977296181 Chr.5: 179023760-179023760 on Build GRCh38
rs566266513 Chr.5: 179023777-179023777 on Build GRCh38
rs143802265 Chr.5: 179023772-179023772 on Build GRCh38
rs981142823 Chr.5: 179023962-179023962 on Build GRCh38
rs1045349053 Chr.5: 179023799-179023799 on Build GRCh38
rs940967176 Chr.5: 179023813-179023813 on Build GRCh38
rs575419924 Chr.5: 179024152-179024152 on Build GRCh38
rs1040442764 Chr.5: 179024047-179024047 on Build GRCh38
rs528143877 Chr.5: 179023739-179023739 on Build GRCh38
rs750737399 Chr.5: 179024084-179024084 on Build GRCh38
rs537768067 Chr.5: 179023872-179023872 on Build GRCh38
rs906523811 Chr.5: 179024035-179024035 on Build GRCh38
rs570290107 Chr.5: 179023836-179023836 on Build GRCh38
rs1032926633 Chr.5: 179023883-179023883 on Build GRCh38
rs867882449 Chr.5: 179024056-179024056 on Build GRCh38
rs138318314 Chr.5: 179023714-179023714 on Build GRCh38
rs907697290 Chr.5: 179023882-179023883 on Build GRCh38
rs34383116 Chr.5: 179023761-179023762 on Build GRCh38
rs939698775 Chr.5: 179023984-179023984 on Build GRCh38

View Full Product Details