Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 179798736-179799275 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MGAT4B
Entrez Gene ID
11282
Gene Name
mannosyl (alpha-1,3-)-glycoprotein beta-1,4-N-acetylglucosaminyltransferase, isozyme B
Gene Aliases
GNT-IV, GNT-IVB
UniGene
Hs.567419
Gene Chromosome Location
Chr.5: 179797597-179806952 on Build GRCh38
Transcripts
NM_014275.4 NM_054013.3 XM_017008998.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5868299 c.1387C>T p.R463W Chr.5: 179798929-179798929 on Build GRCh38
5204924 c.1114C>T p.H372Y Chr.5: 179799238-179799238 on Build GRCh38
5180448 c.1206G>T p.E402D Chr.5: 179799065-179799065 on Build GRCh38
6895903 c.1228G>T p.E410* Chr.5: 179799088-179799088 on Build GRCh38
3854242 c.1130A>G p.K377R Chr.5: 179799267-179799267 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761863989 Chr.5: 179799253-179799253 on Build GRCh38
rs577469008 Chr.5: 179798897-179798897 on Build GRCh38
rs376972309 Chr.5: 179798913-179798913 on Build GRCh38
rs374162408 Chr.5: 179799239-179799239 on Build GRCh38
rs549597971 Chr.5: 179799173-179799173 on Build GRCh38
rs374437340 Chr.5: 179798989-179798989 on Build GRCh38
rs765751203 Chr.5: 179798829-179798829 on Build GRCh38
rs779862747 Chr.5: 179799186-179799186 on Build GRCh38
rs774214600 Chr.5: 179798966-179798966 on Build GRCh38
rs752411779 Chr.5: 179799061-179799061 on Build GRCh38
rs752777524 Chr.5: 179799111-179799111 on Build GRCh38
rs143317140 Chr.5: 179798830-179798830 on Build GRCh38
rs769637690 Chr.5: 179799206-179799206 on Build GRCh38
rs1042249469 Chr.5: 179798987-179798987 on Build GRCh38
rs556662797 Chr.5: 179798960-179798960 on Build GRCh38
rs776255201 Chr.5: 179799190-179799190 on Build GRCh38
rs114009346 Chr.5: 179799152-179799152 on Build GRCh38
rs950488950 Chr.5: 179798859-179798859 on Build GRCh38
rs776181503 Chr.5: 179799155-179799155 on Build GRCh38
rs113507366 Chr.5: 179799047-179799047 on Build GRCh38
rs779094646 Chr.5: 179798909-179798909 on Build GRCh38
rs866010274 Chr.5: 179798982-179798982 on Build GRCh38
rs752365931 Chr.5: 179798977-179798977 on Build GRCh38
rs1009470449 Chr.5: 179799078-179799078 on Build GRCh38
rs370110578 Chr.5: 179798955-179798955 on Build GRCh38
rs759982479 Chr.5: 179799216-179799216 on Build GRCh38
rs759417044 Chr.5: 179799056-179799056 on Build GRCh38
rs191964745 Chr.5: 179799227-179799227 on Build GRCh38
rs1041668342 Chr.5: 179798908-179798908 on Build GRCh38
rs547106742 Chr.5: 179798814-179798814 on Build GRCh38
rs981771027 Chr.5: 179798861-179798861 on Build GRCh38
rs890447632 Chr.5: 179799062-179799062 on Build GRCh38
rs1052218991 Chr.5: 179798803-179798803 on Build GRCh38
rs752137181 Chr.5: 179799093-179799093 on Build GRCh38
rs758223404 Chr.5: 179798812-179798812 on Build GRCh38
rs915196198 Chr.5: 179798963-179798963 on Build GRCh38
rs187369760 Chr.5: 179798912-179798912 on Build GRCh38
rs894309481 Chr.5: 179799204-179799204 on Build GRCh38
rs370254588 Chr.5: 179799187-179799187 on Build GRCh38
rs764183299 Chr.5: 179799172-179799172 on Build GRCh38
rs781571069 Chr.5: 179798945-179798945 on Build GRCh38
rs758135903 Chr.5: 179799185-179799185 on Build GRCh38
rs767797305 Chr.5: 179798844-179798844 on Build GRCh38
rs766318263 Chr.5: 179798894-179798894 on Build GRCh38
rs41285561 Chr.5: 179798978-179798978 on Build GRCh38
rs1024767006 Chr.5: 179798769-179798769 on Build GRCh38
rs1024360336 Chr.5: 179798976-179798976 on Build GRCh38
rs766971194 Chr.5: 179798975-179798975 on Build GRCh38
rs769950703 Chr.5: 179799013-179799013 on Build GRCh38
rs778042352 Chr.5: 179799030-179799030 on Build GRCh38
rs899427281 Chr.5: 179799101-179799101 on Build GRCh38
rs373142990 Chr.5: 179799160-179799160 on Build GRCh38
rs149772768 Chr.5: 179798929-179798929 on Build GRCh38
rs781374590 Chr.5: 179799007-179799007 on Build GRCh38
rs977893173 Chr.5: 179799231-179799231 on Build GRCh38
rs199906848 Chr.5: 179799074-179799074 on Build GRCh38
rs747580349 Chr.5: 179799195-179799195 on Build GRCh38
rs780258686 Chr.5: 179798935-179798935 on Build GRCh38
rs983004168 Chr.5: 179798789-179798789 on Build GRCh38
rs374499619 Chr.5: 179799181-179799181 on Build GRCh38
rs756160967 Chr.5: 179799125-179799125 on Build GRCh38
rs761479850 Chr.5: 179799159-179799159 on Build GRCh38
rs767951345 Chr.5: 179799222-179799222 on Build GRCh38
rs750703932 Chr.5: 179798869-179798869 on Build GRCh38
rs759050954 Chr.5: 179798969-179798969 on Build GRCh38
rs371137796 Chr.5: 179799168-179799168 on Build GRCh38
rs775350978 Chr.5: 179799163-179799174 on Build GRCh38
rs368749618 Chr.5: 179799098-179799098 on Build GRCh38
rs769654245 Chr.5: 179798957-179798957 on Build GRCh38
rs779479635 Chr.5: 179799064-179799064 on Build GRCh38
rs761290023 Chr.5: 179799230-179799230 on Build GRCh38
rs535824668 Chr.5: 179798843-179798843 on Build GRCh38
rs994973301 Chr.5: 179798833-179798833 on Build GRCh38
rs768151436 Chr.5: 179799163-179799163 on Build GRCh38
rs748405884 Chr.5: 179799215-179799215 on Build GRCh38
rs372988890 Chr.5: 179799126-179799126 on Build GRCh38
rs772060257 Chr.5: 179799149-179799149 on Build GRCh38
rs903077757 Chr.5: 179798910-179798910 on Build GRCh38
rs766811234 Chr.5: 179799089-179799089 on Build GRCh38
rs756442794 Chr.5: 179798914-179798914 on Build GRCh38
rs757508104 Chr.5: 179799180-179799180 on Build GRCh38
rs760231271 Chr.5: 179799072-179799072 on Build GRCh38
rs755592209 Chr.5: 179799094-179799094 on Build GRCh38
rs370596784 Chr.5: 179799154-179799154 on Build GRCh38
rs745626425 Chr.5: 179799137-179799137 on Build GRCh38
rs778726361 Chr.5: 179799133-179799133 on Build GRCh38
rs559304447 Chr.5: 179798893-179798893 on Build GRCh38
rs950942084 Chr.5: 179799131-179799131 on Build GRCh38
rs774302363 Chr.5: 179799052-179799052 on Build GRCh38
rs753180944 Chr.5: 179799166-179799166 on Build GRCh38
rs748343126 Chr.5: 179798950-179798950 on Build GRCh38
rs763234378 Chr.5: 179799075-179799075 on Build GRCh38
rs944318936 Chr.5: 179798860-179798860 on Build GRCh38
rs867392773 Chr.5: 179799008-179799008 on Build GRCh38
rs921431686 Chr.5: 179798836-179798836 on Build GRCh38
rs375317939 Chr.5: 179798882-179798882 on Build GRCh38
rs752278018 Chr.5: 179798903-179798903 on Build GRCh38
rs780722671 Chr.5: 179798981-179798981 on Build GRCh38
rs749049937 Chr.5: 179799044-179799044 on Build GRCh38
rs971385498 Chr.5: 179798774-179798774 on Build GRCh38
rs367786665 Chr.5: 179799183-179799183 on Build GRCh38
rs931756093 Chr.5: 179798840-179798840 on Build GRCh38
rs142557156 Chr.5: 179799248-179799248 on Build GRCh38
rs998829142 Chr.5: 179798794-179798794 on Build GRCh38
rs1008206168 Chr.5: 179798819-179798819 on Build GRCh38
rs934756400 Chr.5: 179799023-179799023 on Build GRCh38
rs750774332 Chr.5: 179798911-179798911 on Build GRCh38
rs1012723809 Chr.5: 179799224-179799224 on Build GRCh38
rs376673003 Chr.5: 179799060-179799060 on Build GRCh38
rs775396214 Chr.5: 179799151-179799151 on Build GRCh38
rs748216524 Chr.5: 179799009-179799009 on Build GRCh38
rs911615587 Chr.5: 179799145-179799145 on Build GRCh38
rs562457 Chr.5: 179798768-179798768 on Build GRCh38
rs749485704 Chr.5: 179799128-179799128 on Build GRCh38
rs146867088 Chr.5: 179798961-179798961 on Build GRCh38
rs146600579 Chr.5: 179799099-179799099 on Build GRCh38
rs202150332 Chr.5: 179799161-179799161 on Build GRCh38
rs41285559 Chr.5: 179798811-179798811 on Build GRCh38
rs1043155633 Chr.5: 179798767-179798767 on Build GRCh38
rs749437560 Chr.5: 179798962-179798962 on Build GRCh38
rs555504893 Chr.5: 179799129-179799129 on Build GRCh38

View Full Product Details