Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 151047374-151047899 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BNIPL
Entrez Gene ID
149428
Gene Name
BCL2 interacting protein like
Gene Aliases
BNIP-S, BNIP-Salpha, BNIP-Sbeta, BNIPL-1, BNIPL-2, BNIPL1, BNIPL2, BNIPS, PP753
UniGene
Hs.591473
Gene Chromosome Location
Chr.1: 151036553-151047600 on Build GRCh38
Transcripts
NM_001159642.1 NM_138278.3 XM_011509236.1 XM_017000420.1 NM_017860.3

Gene Symbol
C1orf56
Entrez Gene ID
54964
Gene Name
chromosome 1 open reading frame 56
Gene Aliases
MENT
UniGene
Hs.549171
Gene Chromosome Location
Chr.1: 151047783-151051420 on Build GRCh38
Transcripts
NM_001159642.1 NM_138278.3 XM_011509236.1 XM_017000420.1 NM_017860.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5782694 c.15C>T p.A5A Chr.1: 151047862-151047862 on Build GRCh38
3369272 c.33C>T p.V11V Chr.1: 151047880-151047880 on Build GRCh38
6540829 c.13G>T p.A5S Chr.1: 151047860-151047860 on Build GRCh38
1295018 c.21G>A p.A7A Chr.1: 151047868-151047868 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs748071990 Chr.1: 151047835-151047835 on Build GRCh38
rs116330355 Chr.1: 151047491-151047491 on Build GRCh38
rs1000588042 Chr.1: 151047699-151047699 on Build GRCh38
rs963362664 Chr.1: 151047746-151047746 on Build GRCh38
rs889473368 Chr.1: 151047509-151047509 on Build GRCh38
rs758097746 Chr.1: 151047805-151047805 on Build GRCh38
rs960514054 Chr.1: 151047666-151047666 on Build GRCh38
rs587636906 Chr.1: 151047684-151047684 on Build GRCh38
rs1036525186 Chr.1: 151047440-151047440 on Build GRCh38
rs765420649 Chr.1: 151047679-151047679 on Build GRCh38
rs968168525 Chr.1: 151047601-151047601 on Build GRCh38
rs587685535 Chr.1: 151047588-151047588 on Build GRCh38
rs983502889 Chr.1: 151047537-151047537 on Build GRCh38
rs752918257 Chr.1: 151047683-151047683 on Build GRCh38
rs977832904 Chr.1: 151047621-151047621 on Build GRCh38
rs754448024 Chr.1: 151047830-151047830 on Build GRCh38
rs903500564 Chr.1: 151047858-151047858 on Build GRCh38
rs1006566232 Chr.1: 151047802-151047802 on Build GRCh38
rs587600278 Chr.1: 151047800-151047800 on Build GRCh38
rs587641203 Chr.1: 151047729-151047729 on Build GRCh38
rs751113549 Chr.1: 151047829-151047829 on Build GRCh38
rs997749658 Chr.1: 151047468-151047468 on Build GRCh38
rs587765989 Chr.1: 151047630-151047630 on Build GRCh38
rs587630379 Chr.1: 151047545-151047545 on Build GRCh38
rs779763776 Chr.1: 151047827-151047827 on Build GRCh38
rs12066560 Chr.1: 151047718-151047718 on Build GRCh38
rs891085687 Chr.1: 151047681-151047681 on Build GRCh38
rs777514994 Chr.1: 151047846-151047846 on Build GRCh38
rs768061314 Chr.1: 151047723-151047723 on Build GRCh38
rs115698912 Chr.1: 151047744-151047744 on Build GRCh38
rs759572470 Chr.1: 151047844-151047844 on Build GRCh38
rs756431825 Chr.1: 151047838-151047838 on Build GRCh38
rs1012454222 Chr.1: 151047727-151047727 on Build GRCh38
rs907935648 Chr.1: 151047556-151047556 on Build GRCh38
rs1048366 Chr.1: 151047572-151047572 on Build GRCh38
rs185988506 Chr.1: 151047450-151047450 on Build GRCh38
rs376195501 Chr.1: 151047593-151047593 on Build GRCh38
rs749562753 Chr.1: 151047850-151047850 on Build GRCh38
rs948079776 Chr.1: 151047701-151047701 on Build GRCh38
rs1050655289 Chr.1: 151047768-151047768 on Build GRCh38
rs940813196 Chr.1: 151047730-151047730 on Build GRCh38
rs1030862383 Chr.1: 151047638-151047638 on Build GRCh38
rs1049615711 Chr.1: 151047862-151047862 on Build GRCh38
rs1042084243 Chr.1: 151047687-151047687 on Build GRCh38
rs1008300887 Chr.1: 151047685-151047685 on Build GRCh38
rs769509897 Chr.1: 151047837-151047837 on Build GRCh38
rs781009141 Chr.1: 151047832-151047832 on Build GRCh38
rs587711835 Chr.1: 151047690-151047690 on Build GRCh38
rs760991831 Chr.1: 151047452-151047452 on Build GRCh38
rs776697474 Chr.1: 151047761-151047761 on Build GRCh38
rs587698460 Chr.1: 151047453-151047453 on Build GRCh38
rs902049727 Chr.1: 151047758-151047758 on Build GRCh38
rs587749424 Chr.1: 151047694-151047694 on Build GRCh38
rs771128428 Chr.1: 151047869-151047869 on Build GRCh38
rs918175797 Chr.1: 151047461-151047461 on Build GRCh38
rs763520740 Chr.1: 151047725-151047726 on Build GRCh38

View Full Product Details