Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 7394686-7395183 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RIOK1
Entrez Gene ID
83732
Gene Name
RIO kinase 1
Gene Aliases
AD034, RRP10, bA288G3.1
UniGene
Hs.437474
Gene Chromosome Location
Chr.6: 7389816-7418037 on Build GRCh38
Transcripts
NM_031480.2 XM_005249446.3 XM_011514933.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
80456 c.348T>C p.F116F Chr.6: 7395124-7395124 on Build GRCh38
6107775 c.322C>G p.P108A Chr.6: 7395098-7395098 on Build GRCh38
1081507 c.344A>C p.K115T Chr.6: 7395120-7395120 on Build GRCh38
223046 c.367+1G>A p.? Chr.6: 7395144-7395144 on Build GRCh38
484340 c.305C>T p.S102L Chr.6: 7395081-7395081 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777086697 Chr.6: 7394724-7394724 on Build GRCh38
rs375949170 Chr.6: 7395014-7395014 on Build GRCh38
rs553166454 Chr.6: 7395077-7395077 on Build GRCh38
rs990897676 Chr.6: 7394851-7394851 on Build GRCh38
rs200484094 Chr.6: 7395107-7395107 on Build GRCh38
rs889943194 Chr.6: 7395070-7395070 on Build GRCh38
rs368392943 Chr.6: 7395011-7395011 on Build GRCh38
rs114381343 Chr.6: 7395089-7395089 on Build GRCh38
rs949328486 Chr.6: 7394905-7394905 on Build GRCh38
rs142247752 Chr.6: 7395140-7395140 on Build GRCh38
rs924291343 Chr.6: 7394783-7394788 on Build GRCh38
rs971653507 Chr.6: 7394970-7394970 on Build GRCh38
rs35248924 Chr.6: 7394893-7394893 on Build GRCh38
rs368061988 Chr.6: 7394852-7394853 on Build GRCh38
rs769061872 Chr.6: 7395071-7395071 on Build GRCh38
rs985007121 Chr.6: 7394769-7394769 on Build GRCh38
rs188124300 Chr.6: 7394968-7394968 on Build GRCh38
rs569460415 Chr.6: 7394765-7394765 on Build GRCh38
rs1006077796 Chr.6: 7394892-7394893 on Build GRCh38
rs918047304 Chr.6: 7394881-7394881 on Build GRCh38
rs538389109 Chr.6: 7394829-7394832 on Build GRCh38
rs371718113 Chr.6: 7395059-7395059 on Build GRCh38
rs375154367 Chr.6: 7395160-7395160 on Build GRCh38
rs776773785 Chr.6: 7395075-7395075 on Build GRCh38
rs554618381 Chr.6: 7394819-7394819 on Build GRCh38
rs375314972 Chr.6: 7395116-7395116 on Build GRCh38
rs757823265 Chr.6: 7394956-7394956 on Build GRCh38
rs1026100852 Chr.6: 7394874-7394874 on Build GRCh38
rs746273739 Chr.6: 7395023-7395023 on Build GRCh38
rs777390484 Chr.6: 7395029-7395029 on Build GRCh38
rs991577783 Chr.6: 7394936-7394936 on Build GRCh38
rs371946445 Chr.6: 7394803-7394803 on Build GRCh38
rs901722564 Chr.6: 7394852-7394853 on Build GRCh38
rs887119596 Chr.6: 7395142-7395142 on Build GRCh38
rs185514944 Chr.6: 7394812-7394812 on Build GRCh38
rs577329182 Chr.6: 7395048-7395048 on Build GRCh38
rs772728649 Chr.6: 7395040-7395040 on Build GRCh38
rs369600325 Chr.6: 7395143-7395143 on Build GRCh38
rs748772188 Chr.6: 7395039-7395039 on Build GRCh38
rs935787859 Chr.6: 7394797-7394797 on Build GRCh38
rs773311605 Chr.6: 7395095-7395095 on Build GRCh38
rs140246108 Chr.6: 7394965-7394965 on Build GRCh38
rs762844607 Chr.6: 7394987-7394987 on Build GRCh38
rs751373578 Chr.6: 7395006-7395006 on Build GRCh38
rs755669377 Chr.6: 7394713-7394713 on Build GRCh38
rs965681268 Chr.6: 7394716-7394716 on Build GRCh38
rs367708123 Chr.6: 7395021-7395021 on Build GRCh38
rs935669211 Chr.6: 7395019-7395019 on Build GRCh38
rs943157599 Chr.6: 7394836-7394836 on Build GRCh38
rs12527405 Chr.6: 7394996-7394996 on Build GRCh38
rs958882542 Chr.6: 7394923-7394923 on Build GRCh38
rs917476130 Chr.6: 7394733-7394733 on Build GRCh38
rs866875419 Chr.6: 7395060-7395060 on Build GRCh38
rs866982961 Chr.6: 7394760-7394760 on Build GRCh38
rs55698032 Chr.6: 7395117-7395117 on Build GRCh38
rs766312760 Chr.6: 7395114-7395114 on Build GRCh38
rs998194918 Chr.6: 7394853-7394853 on Build GRCh38

View Full Product Details