Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 18138877-18139378 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TPMT
Entrez Gene ID
7172
Gene Name
thiopurine S-methyltransferase
Gene Aliases
TPMTD
UniGene
Hs.444319
Gene Chromosome Location
Chr.6: 18128311-18155165 on Build GRCh38
Transcripts
NM_000367.3 XM_011514839.2 XM_017011241.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6806885 c.461C>T p.A154V Chr.6: 18138996-18138996 on Build GRCh38
5908704 c.478C>T p.P160S Chr.6: 18138979-18138979 on Build GRCh38
3761722 c.474C>T p.I158I Chr.6: 18138983-18138983 on Build GRCh38
4986704 c.460G>A p.A154T Chr.6: 18138997-18138997 on Build GRCh38
6535870 c.480A>C p.P160P Chr.6: 18138977-18138977 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs371416001 Chr.6: 18139143-18139143 on Build GRCh38
rs55698609 Chr.6: 18139219-18139219 on Build GRCh38
rs756585774 Chr.6: 18138950-18138950 on Build GRCh38
rs775254609 Chr.6: 18139035-18139035 on Build GRCh38
rs771163159 Chr.6: 18139127-18139127 on Build GRCh38
rs764218923 Chr.6: 18138912-18138912 on Build GRCh38
rs759199430 Chr.6: 18139004-18139004 on Build GRCh38
rs953244525 Chr.6: 18139127-18139127 on Build GRCh38
rs892264381 Chr.6: 18138902-18138902 on Build GRCh38
rs780544641 Chr.6: 18138958-18138958 on Build GRCh38
rs1015135138 Chr.6: 18139045-18139045 on Build GRCh38
rs999504545 Chr.6: 18139042-18139042 on Build GRCh38
rs760971282 Chr.6: 18139077-18139078 on Build GRCh38
rs923485672 Chr.6: 18139123-18139123 on Build GRCh38
rs902457648 Chr.6: 18139251-18139251 on Build GRCh38
rs1052328735 Chr.6: 18139172-18139172 on Build GRCh38
rs188681338 Chr.6: 18139167-18139167 on Build GRCh38
rs759632871 Chr.6: 18139327-18139327 on Build GRCh38
rs369077559 Chr.6: 18138985-18138985 on Build GRCh38
rs1055892392 Chr.6: 18139350-18139350 on Build GRCh38
rs970885400 Chr.6: 18139129-18139129 on Build GRCh38
rs763954681 Chr.6: 18139040-18139040 on Build GRCh38
rs569219705 Chr.6: 18139233-18139233 on Build GRCh38
rs750823453 Chr.6: 18138941-18138941 on Build GRCh38
rs1052129421 Chr.6: 18139231-18139231 on Build GRCh38
rs773446298 Chr.6: 18139047-18139048 on Build GRCh38
rs750869528 Chr.6: 18139154-18139154 on Build GRCh38
rs140542140 Chr.6: 18139161-18139161 on Build GRCh38
rs1012344905 Chr.6: 18139274-18139274 on Build GRCh38
rs1046640888 Chr.6: 18139228-18139228 on Build GRCh38
rs767249628 Chr.6: 18139100-18139100 on Build GRCh38
rs577776972 Chr.6: 18138913-18138913 on Build GRCh38
rs530367205 Chr.6: 18139254-18139254 on Build GRCh38
rs2842934 Chr.6: 18138983-18138983 on Build GRCh38
rs1002017719 Chr.6: 18139292-18139292 on Build GRCh38
rs962163466 Chr.6: 18139074-18139074 on Build GRCh38
rs376140800 Chr.6: 18138921-18138921 on Build GRCh38
rs563724950 Chr.6: 18139205-18139205 on Build GRCh38
rs544066731 Chr.6: 18139193-18139193 on Build GRCh38
rs777339242 Chr.6: 18138979-18138979 on Build GRCh38
rs756573222 Chr.6: 18139155-18139155 on Build GRCh38
rs1800460 Chr.6: 18138997-18138997 on Build GRCh38
rs1023843087 Chr.6: 18139126-18139126 on Build GRCh38
rs528468301 Chr.6: 18139089-18139089 on Build GRCh38
rs976236762 Chr.6: 18139092-18139092 on Build GRCh38
rs539964543 Chr.6: 18139275-18139275 on Build GRCh38
rs769630417 Chr.6: 18139029-18139029 on Build GRCh38
rs747392577 Chr.6: 18139069-18139070 on Build GRCh38
rs773721497 Chr.6: 18139008-18139008 on Build GRCh38
rs761750303 Chr.6: 18139073-18139073 on Build GRCh38
rs893803623 Chr.6: 18139261-18139261 on Build GRCh38
rs34072670 Chr.6: 18139262-18139263 on Build GRCh38
rs112339338 Chr.6: 18138970-18138970 on Build GRCh38
rs982772377 Chr.6: 18139134-18139134 on Build GRCh38
rs144041067 Chr.6: 18138969-18138969 on Build GRCh38
rs16880276 Chr.6: 18139034-18139034 on Build GRCh38
rs767812417 Chr.6: 18139079-18139079 on Build GRCh38
rs72552737 Chr.6: 18139027-18139027 on Build GRCh38
rs933145584 Chr.6: 18139230-18139230 on Build GRCh38
rs781214285 Chr.6: 18138932-18138932 on Build GRCh38
rs1038727797 Chr.6: 18139212-18139212 on Build GRCh38
rs56019966 Chr.6: 18139041-18139041 on Build GRCh38
rs1012335019 Chr.6: 18139103-18139103 on Build GRCh38
rs144401881 Chr.6: 18139210-18139210 on Build GRCh38
rs941769105 Chr.6: 18138935-18138935 on Build GRCh38
rs772191669 Chr.6: 18138942-18138942 on Build GRCh38
rs762947310 Chr.6: 18139038-18139038 on Build GRCh38

View Full Product Details