Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 18148717-18149214 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TPMT
Entrez Gene ID
7172
Gene Name
thiopurine S-methyltransferase
Gene Aliases
TPMTD
UniGene
Hs.444319
Gene Chromosome Location
Chr.6: 18128311-18155165 on Build GRCh38
Transcripts
NM_000367.3 XM_011514839.2 XM_017011241.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3860667 c.82G>C p.E28Q Chr.6: 18149046-18149046 on Build GRCh38
1442420 c.105C>T p.N35N Chr.6: 18149023-18149023 on Build GRCh38
3357979 c.36G>A p.E12E Chr.6: 18149092-18149092 on Build GRCh38
4160403 c.62A>C p.N21T Chr.6: 18149066-18149066 on Build GRCh38
1567911 c.89A>G p.Q30R Chr.6: 18149039-18149039 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs16880307 Chr.6: 18149058-18149058 on Build GRCh38
rs763783722 Chr.6: 18148997-18148997 on Build GRCh38
rs759836180 Chr.6: 18149032-18149033 on Build GRCh38
rs543679347 Chr.6: 18149176-18149176 on Build GRCh38
rs545549817 Chr.6: 18148967-18148967 on Build GRCh38
rs541573301 Chr.6: 18148871-18148871 on Build GRCh38
rs772597033 Chr.6: 18148958-18148958 on Build GRCh38
rs369346510 Chr.6: 18149163-18149163 on Build GRCh38
rs779065769 Chr.6: 18149065-18149065 on Build GRCh38
rs977305863 Chr.6: 18148915-18148915 on Build GRCh38
rs143125661 Chr.6: 18149120-18149120 on Build GRCh38
rs747281890 Chr.6: 18149167-18149167 on Build GRCh38
rs917706425 Chr.6: 18149139-18149139 on Build GRCh38
rs201965126 Chr.6: 18148952-18148952 on Build GRCh38
rs72552742 Chr.6: 18149045-18149045 on Build GRCh38
rs3931660 Chr.6: 18148874-18148874 on Build GRCh38
rs201529425 Chr.6: 18148978-18148978 on Build GRCh38
rs762702707 Chr.6: 18148991-18148991 on Build GRCh38
rs948295693 Chr.6: 18149084-18149084 on Build GRCh38
rs762507236 Chr.6: 18149087-18149087 on Build GRCh38
rs761059662 Chr.6: 18148970-18148970 on Build GRCh38
rs367678410 Chr.6: 18149023-18149023 on Build GRCh38
rs975745626 Chr.6: 18148829-18148829 on Build GRCh38
rs757932402 Chr.6: 18148933-18148933 on Build GRCh38
rs757081801 Chr.6: 18149006-18149006 on Build GRCh38
rs771278148 Chr.6: 18149085-18149085 on Build GRCh38
rs531775781 Chr.6: 18148885-18148885 on Build GRCh38
rs747615886 Chr.6: 18149081-18149081 on Build GRCh38
rs768458136 Chr.6: 18149092-18149092 on Build GRCh38
rs200695400 Chr.6: 18149009-18149009 on Build GRCh38
rs776967691 Chr.6: 18149032-18149036 on Build GRCh38
rs773433252 Chr.6: 18149080-18149080 on Build GRCh38
rs751967904 Chr.6: 18149078-18149078 on Build GRCh38
rs750181764 Chr.6: 18149119-18149119 on Build GRCh38
rs931453225 Chr.6: 18149103-18149103 on Build GRCh38
rs749266740 Chr.6: 18149056-18149056 on Build GRCh38
rs963918757 Chr.6: 18148795-18148795 on Build GRCh38
rs113886055 Chr.6: 18148955-18148955 on Build GRCh38
rs750424422 Chr.6: 18149022-18149022 on Build GRCh38
rs767187141 Chr.6: 18149014-18149014 on Build GRCh38
rs200780293 Chr.6: 18149098-18149098 on Build GRCh38
rs9333569 Chr.6: 18149127-18149127 on Build GRCh38
rs1037040111 Chr.6: 18148846-18148846 on Build GRCh38
rs781515455 Chr.6: 18149172-18149172 on Build GRCh38
rs372490350 Chr.6: 18149149-18149149 on Build GRCh38
rs758611974 Chr.6: 18149160-18149160 on Build GRCh38
rs574354573 Chr.6: 18148817-18148817 on Build GRCh38
rs772470049 Chr.6: 18149079-18149079 on Build GRCh38
rs748335791 Chr.6: 18149074-18149074 on Build GRCh38
rs755089146 Chr.6: 18149150-18149150 on Build GRCh38
rs267607275 Chr.6: 18149126-18149126 on Build GRCh38
rs111674665 Chr.6: 18149086-18149086 on Build GRCh38
rs764858046 Chr.6: 18148973-18148973 on Build GRCh38
rs764615038 Chr.6: 18148801-18148801 on Build GRCh38
rs755215484 Chr.6: 18149057-18149057 on Build GRCh38
rs771494830 Chr.6: 18149168-18149168 on Build GRCh38
rs202034437 Chr.6: 18149175-18149175 on Build GRCh38
rs72552741 Chr.6: 18149031-18149031 on Build GRCh38
rs200148021 Chr.6: 18148999-18148999 on Build GRCh38
rs376768623 Chr.6: 18149157-18149157 on Build GRCh38
rs1019331826 Chr.6: 18148782-18148782 on Build GRCh38
rs995621858 Chr.6: 18148872-18148872 on Build GRCh38
rs914385368 Chr.6: 18148855-18148855 on Build GRCh38
rs780247688 Chr.6: 18149033-18149033 on Build GRCh38
rs879204243 Chr.6: 18148884-18148884 on Build GRCh38

View Full Product Details