Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 24666592-24667093 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACOT13
Entrez Gene ID
55856
Gene Name
acyl-CoA thioesterase 13
Gene Aliases
HT012, PNAS-27, THEM2
UniGene
Hs.731605
Gene Chromosome Location
Chr.6: 24667035-24705069 on Build GRCh38
Transcripts
NM_001160094.1 NM_018473.3 NM_016614.2

Gene Symbol
TDP2
Entrez Gene ID
51567
Gene Name
tyrosyl-DNA phosphodiesterase 2
Gene Aliases
AD022, EAP2, EAPII, TTRAP, dJ30M3.3, hTDP2
UniGene
Hs.403010
Gene Chromosome Location
Chr.6: 24649977-24666887 on Build GRCh38
Transcripts
NM_001160094.1 NM_018473.3 NM_016614.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6566429 c.151G>A p.E51K Chr.6: 24666802-24666802 on Build GRCh38
1567904 c.85T>A p.C29S Chr.6: 24666778-24666778 on Build GRCh38
3829765 c.156G>A p.W52* Chr.6: 24666707-24666707 on Build GRCh38
5975912 c.111C>T p.S37S Chr.6: 24666752-24666752 on Build GRCh38
3719281 c.131C>T p.A44V Chr.6: 24666822-24666822 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751092306 Chr.6: 24666884-24666884 on Build GRCh38
rs775826087 Chr.6: 24666617-24666620 on Build GRCh38
rs368460491 Chr.6: 24666752-24666752 on Build GRCh38
rs1048331198 Chr.6: 24666882-24666882 on Build GRCh38
rs376227773 Chr.6: 24667065-24667065 on Build GRCh38
rs199980848 Chr.6: 24666866-24666866 on Build GRCh38
rs777517528 Chr.6: 24666855-24666855 on Build GRCh38
rs773899351 Chr.6: 24666983-24666983 on Build GRCh38
rs200485971 Chr.6: 24666747-24666747 on Build GRCh38
rs756749615 Chr.6: 24666949-24666949 on Build GRCh38
rs1015603888 Chr.6: 24667016-24667016 on Build GRCh38
rs67657725 Chr.6: 24666978-24666979 on Build GRCh38
rs764713488 Chr.6: 24666786-24666786 on Build GRCh38
rs771628314 Chr.6: 24666978-24666978 on Build GRCh38
rs904667491 Chr.6: 24666836-24666836 on Build GRCh38
rs201132552 Chr.6: 24666889-24666889 on Build GRCh38
rs778549935 Chr.6: 24666806-24666806 on Build GRCh38
rs763659985 Chr.6: 24666883-24666883 on Build GRCh38
rs577053435 Chr.6: 24666999-24666999 on Build GRCh38
rs747517187 Chr.6: 24666975-24666975 on Build GRCh38
rs778744134 Chr.6: 24666850-24666850 on Build GRCh38
rs753545235 Chr.6: 24666793-24666793 on Build GRCh38
rs779031310 Chr.6: 24666962-24666962 on Build GRCh38
rs769972820 Chr.6: 24666862-24666862 on Build GRCh38
rs781147591 Chr.6: 24666891-24666891 on Build GRCh38
rs772663070 Chr.6: 24666931-24666931 on Build GRCh38
rs17249848 Chr.6: 24667028-24667060 on Build GRCh38
rs576461785 Chr.6: 24667053-24667053 on Build GRCh38
rs750374601 Chr.6: 24666957-24666957 on Build GRCh38
rs553824268 Chr.6: 24666970-24666970 on Build GRCh38
rs1033685038 Chr.6: 24666953-24666953 on Build GRCh38
rs542343971 Chr.6: 24667057-24667057 on Build GRCh38
rs746736116 Chr.6: 24666829-24666829 on Build GRCh38
rs1050300465 Chr.6: 24666734-24666734 on Build GRCh38
rs1004014515 Chr.6: 24666937-24666937 on Build GRCh38
rs760554150 Chr.6: 24666766-24666766 on Build GRCh38
rs762368465 Chr.6: 24666868-24666868 on Build GRCh38
rs140404711 Chr.6: 24666839-24666839 on Build GRCh38
rs753218291 Chr.6: 24666708-24666708 on Build GRCh38
rs749593177 Chr.6: 24666773-24666773 on Build GRCh38
rs62400776 Chr.6: 24666971-24666971 on Build GRCh38
rs369863568 Chr.6: 24666871-24666871 on Build GRCh38
rs748475191 Chr.6: 24666620-24666620 on Build GRCh38
rs186475291 Chr.6: 24667064-24667064 on Build GRCh38
rs760326906 Chr.6: 24666845-24666846 on Build GRCh38
rs779514628 Chr.6: 24666791-24666791 on Build GRCh38
rs764930902 Chr.6: 24666872-24666872 on Build GRCh38
rs768818848 Chr.6: 24666913-24666913 on Build GRCh38
rs374879671 Chr.6: 24666743-24666743 on Build GRCh38
rs386406527 Chr.6: 24666976-24666977 on Build GRCh38
rs200802722 Chr.6: 24666730-24666730 on Build GRCh38
rs762016194 Chr.6: 24666845-24666845 on Build GRCh38
rs201203124 Chr.6: 24666652-24666652 on Build GRCh38
rs761642526 Chr.6: 24666661-24666661 on Build GRCh38
rs777794075 Chr.6: 24666824-24666824 on Build GRCh38
rs17243486 Chr.6: 24667069-24667069 on Build GRCh38
rs533990165 Chr.6: 24666965-24666965 on Build GRCh38
rs965158841 Chr.6: 24667019-24667019 on Build GRCh38
rs772007601 Chr.6: 24666972-24666972 on Build GRCh38
rs773676234 Chr.6: 24666870-24666870 on Build GRCh38
rs766343145 Chr.6: 24667002-24667002 on Build GRCh38
rs779185834 Chr.6: 24666800-24666800 on Build GRCh38
rs758789376 Chr.6: 24666622-24666622 on Build GRCh38
rs796178595 Chr.6: 24666805-24666805 on Build GRCh38
rs200941577 Chr.6: 24666977-24666977 on Build GRCh38
rs921001990 Chr.6: 24667032-24667032 on Build GRCh38
rs773122729 Chr.6: 24666672-24666672 on Build GRCh38
rs3835233 Chr.6: 24666979-24666980 on Build GRCh38
rs770777577 Chr.6: 24666830-24666830 on Build GRCh38
rs749881140 Chr.6: 24666799-24666825 on Build GRCh38
rs774850927 Chr.6: 24666636-24666636 on Build GRCh38
rs755927062 Chr.6: 24666896-24666896 on Build GRCh38
rs749279037 Chr.6: 24666863-24666863 on Build GRCh38
rs906322996 Chr.6: 24667060-24667060 on Build GRCh38
rs151213624 Chr.6: 24666788-24666788 on Build GRCh38
rs777205836 Chr.6: 24666939-24666939 on Build GRCh38
rs761059957 Chr.6: 24666938-24666938 on Build GRCh38
rs545908975 Chr.6: 24667044-24667044 on Build GRCh38
rs749520354 Chr.6: 24666902-24666902 on Build GRCh38
rs770844602 Chr.6: 24666859-24666859 on Build GRCh38
rs747466730 Chr.6: 24666722-24666722 on Build GRCh38
rs1045822817 Chr.6: 24667041-24667064 on Build GRCh38
rs767396098 Chr.6: 24666952-24666952 on Build GRCh38
rs752212750 Chr.6: 24666851-24666851 on Build GRCh38
rs763706463 Chr.6: 24666946-24666946 on Build GRCh38
rs141964303 Chr.6: 24666688-24666688 on Build GRCh38
rs768684552 Chr.6: 24666646-24666646 on Build GRCh38
rs35306604 Chr.6: 24666702-24666702 on Build GRCh38
rs17243164 Chr.6: 24666977-24666978 on Build GRCh38
rs776938646 Chr.6: 24666771-24666771 on Build GRCh38
rs748758001 Chr.6: 24666742-24666742 on Build GRCh38
rs556619847 Chr.6: 24667050-24667050 on Build GRCh38
rs888243785 Chr.6: 24666782-24666782 on Build GRCh38
rs745712158 Chr.6: 24666861-24666861 on Build GRCh38
rs199594700 Chr.6: 24666799-24666799 on Build GRCh38
rs376618846 Chr.6: 24666666-24666666 on Build GRCh38
rs758697778 Chr.6: 24666719-24666719 on Build GRCh38
rs776653065 Chr.6: 24666831-24666831 on Build GRCh38
rs767005683 Chr.6: 24666772-24666773 on Build GRCh38
rs534169765 Chr.6: 24666809-24666809 on Build GRCh38
rs780181755 Chr.6: 24666900-24666900 on Build GRCh38
rs1036886409 Chr.6: 24667059-24667059 on Build GRCh38
rs202243869 Chr.6: 24666879-24666879 on Build GRCh38
rs773774995 Chr.6: 24667045-24667045 on Build GRCh38
rs748879090 Chr.6: 24666637-24666637 on Build GRCh38
rs780767326 Chr.6: 24666633-24666633 on Build GRCh38
rs753826333 Chr.6: 24666697-24666697 on Build GRCh38
rs574259087 Chr.6: 24666634-24666634 on Build GRCh38
rs766669494 Chr.6: 24666692-24666692 on Build GRCh38
rs866636503 Chr.6: 24666927-24666927 on Build GRCh38
rs61760186 Chr.6: 24666846-24666846 on Build GRCh38
rs755450845 Chr.6: 24666614-24666614 on Build GRCh38
rs142424556 Chr.6: 24666854-24666854 on Build GRCh38
rs773876097 Chr.6: 24666932-24666932 on Build GRCh38
rs752608318 Chr.6: 24666758-24666758 on Build GRCh38
rs146127613 Chr.6: 24666781-24666781 on Build GRCh38
rs771868751 Chr.6: 24666822-24666822 on Build GRCh38
rs976873894 Chr.6: 24667030-24667030 on Build GRCh38
rs201104912 Chr.6: 24666857-24666857 on Build GRCh38
rs761256693 Chr.6: 24666992-24666992 on Build GRCh38
rs777034962 Chr.6: 24666979-24666979 on Build GRCh38
rs1001662438 Chr.6: 24666837-24666837 on Build GRCh38
rs779796863 Chr.6: 24666733-24666733 on Build GRCh38
rs763890425 Chr.6: 24666844-24666844 on Build GRCh38
rs890380136 Chr.6: 24666754-24666754 on Build GRCh38
rs779433262 Chr.6: 24666617-24666617 on Build GRCh38
rs752415197 Chr.6: 24666873-24666873 on Build GRCh38
rs554204280 Chr.6: 24666916-24666916 on Build GRCh38
rs765978658 Chr.6: 24666858-24666859 on Build GRCh38
rs762658629 Chr.6: 24666940-24666940 on Build GRCh38
rs761457468 Chr.6: 24666768-24666769 on Build GRCh38
rs376289525 Chr.6: 24666869-24666869 on Build GRCh38
rs966934739 Chr.6: 24666984-24666984 on Build GRCh38
rs78727527 Chr.6: 24666989-24666989 on Build GRCh38
rs777996628 Chr.6: 24666720-24666720 on Build GRCh38
rs770061117 Chr.6: 24666833-24666833 on Build GRCh38
rs765307607 Chr.6: 24666780-24666780 on Build GRCh38
rs749085711 Chr.6: 24666961-24666961 on Build GRCh38
rs780955346 Chr.6: 24666860-24666860 on Build GRCh38
rs755575770 Chr.6: 24667063-24667063 on Build GRCh38
rs373305535 Chr.6: 24666958-24666958 on Build GRCh38
rs765688086 Chr.6: 24666706-24666706 on Build GRCh38
rs748213796 Chr.6: 24666921-24666921 on Build GRCh38
rs770516444 Chr.6: 24666644-24666644 on Build GRCh38
rs772190723 Chr.6: 24666755-24666755 on Build GRCh38
rs764469672 Chr.6: 24666841-24666841 on Build GRCh38
rs768315412 Chr.6: 24666981-24666981 on Build GRCh38
rs371861850 Chr.6: 24666894-24666894 on Build GRCh38
rs759859886 Chr.6: 24666980-24666980 on Build GRCh38

View Full Product Details