Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 24697792-24698297 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACOT13
Entrez Gene ID
55856
Gene Name
acyl-CoA thioesterase 13
Gene Aliases
HT012, PNAS-27, THEM2
UniGene
Hs.731605
Gene Chromosome Location
Chr.6: 24667035-24705069 on Build GRCh38
Transcripts
NM_001160094.1 NM_018473.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7200320 c.163G>C p.G55R Chr.6: 24698033-24698033 on Build GRCh38
6847677 c.61G>A p.V21I Chr.6: 24697931-24697931 on Build GRCh38
7200319 c.232G>C p.G78R Chr.6: 24698033-24698033 on Build GRCh38
6847676 c.130G>A p.V44I Chr.6: 24697931-24697931 on Build GRCh38
3761729 c.186G>A p.T62T Chr.6: 24697987-24697987 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767820339 Chr.6: 24698006-24698006 on Build GRCh38
rs755344631 Chr.6: 24697931-24697931 on Build GRCh38
rs373077414 Chr.6: 24698074-24698074 on Build GRCh38
rs572706204 Chr.6: 24697933-24697935 on Build GRCh38
rs369757449 Chr.6: 24697973-24697973 on Build GRCh38
rs780932293 Chr.6: 24698058-24698058 on Build GRCh38
rs749605795 Chr.6: 24697912-24697912 on Build GRCh38
rs138986814 Chr.6: 24698042-24698042 on Build GRCh38
rs761698522 Chr.6: 24698091-24698091 on Build GRCh38
rs942961704 Chr.6: 24697867-24697867 on Build GRCh38
rs756277636 Chr.6: 24697937-24697937 on Build GRCh38
rs766809760 Chr.6: 24698025-24698025 on Build GRCh38
rs753401228 Chr.6: 24697866-24697867 on Build GRCh38
rs879175157 Chr.6: 24697901-24697901 on Build GRCh38
rs1017083065 Chr.6: 24698130-24698130 on Build GRCh38
rs377106918 Chr.6: 24698026-24698026 on Build GRCh38
rs936098645 Chr.6: 24698196-24698196 on Build GRCh38
rs951838503 Chr.6: 24698110-24698110 on Build GRCh38
rs199826558 Chr.6: 24698157-24698157 on Build GRCh38
rs576617412 Chr.6: 24697864-24697865 on Build GRCh38
rs774254597 Chr.6: 24698075-24698075 on Build GRCh38
rs186922814 Chr.6: 24698232-24698232 on Build GRCh38
rs922569505 Chr.6: 24698221-24698221 on Build GRCh38
rs934636538 Chr.6: 24697927-24697927 on Build GRCh38
rs200470807 Chr.6: 24697991-24697991 on Build GRCh38
rs142012282 Chr.6: 24697986-24697986 on Build GRCh38
rs754714185 Chr.6: 24697855-24697855 on Build GRCh38
rs767492012 Chr.6: 24697868-24697868 on Build GRCh38
rs964586975 Chr.6: 24698154-24698154 on Build GRCh38
rs554819611 Chr.6: 24698176-24698176 on Build GRCh38
rs757296889 Chr.6: 24697860-24697860 on Build GRCh38
rs143667676 Chr.6: 24697959-24697959 on Build GRCh38
rs1054971783 Chr.6: 24698220-24698220 on Build GRCh38
rs148153546 Chr.6: 24697970-24697970 on Build GRCh38
rs933953280 Chr.6: 24698169-24698169 on Build GRCh38
rs1050354505 Chr.6: 24697985-24697985 on Build GRCh38
rs894340527 Chr.6: 24698000-24698000 on Build GRCh38
rs376640796 Chr.6: 24698153-24698153 on Build GRCh38
rs764953927 Chr.6: 24697890-24697891 on Build GRCh38
rs200235265 Chr.6: 24697932-24697932 on Build GRCh38
rs369051288 Chr.6: 24698156-24698156 on Build GRCh38
rs370510266 Chr.6: 24698067-24698067 on Build GRCh38
rs752291562 Chr.6: 24697874-24697875 on Build GRCh38
rs376495400 Chr.6: 24697956-24697956 on Build GRCh38
rs12193165 Chr.6: 24698272-24698272 on Build GRCh38
rs924641530 Chr.6: 24698189-24698189 on Build GRCh38
rs753156463 Chr.6: 24698039-24698039 on Build GRCh38
rs772136221 Chr.6: 24698094-24698094 on Build GRCh38
rs545035137 Chr.6: 24697969-24697969 on Build GRCh38
rs906841084 Chr.6: 24698032-24698032 on Build GRCh38
rs951609627 Chr.6: 24697837-24697837 on Build GRCh38
rs752469354 Chr.6: 24697904-24697904 on Build GRCh38
rs773497095 Chr.6: 24698099-24698099 on Build GRCh38
rs1025983538 Chr.6: 24698086-24698086 on Build GRCh38
rs760130636 Chr.6: 24697865-24697866 on Build GRCh38
rs758439505 Chr.6: 24697928-24697928 on Build GRCh38
rs7765904 Chr.6: 24697987-24697987 on Build GRCh38
rs757377688 Chr.6: 24698069-24698070 on Build GRCh38
rs780641771 Chr.6: 24697909-24697909 on Build GRCh38
rs780070370 Chr.6: 24698068-24698068 on Build GRCh38
rs141508229 Chr.6: 24697950-24697950 on Build GRCh38
rs984985970 Chr.6: 24698124-24698124 on Build GRCh38
rs750562644 Chr.6: 24697874-24697874 on Build GRCh38
rs992726077 Chr.6: 24698103-24698103 on Build GRCh38
rs988804113 Chr.6: 24698215-24698215 on Build GRCh38
rs977399843 Chr.6: 24698170-24698170 on Build GRCh38
rs769582127 Chr.6: 24697972-24697972 on Build GRCh38
rs979269613 Chr.6: 24698135-24698135 on Build GRCh38
rs1012817519 Chr.6: 24698001-24698001 on Build GRCh38
rs758095038 Chr.6: 24698179-24698179 on Build GRCh38
rs181894670 Chr.6: 24698142-24698142 on Build GRCh38
rs555097121 Chr.6: 24698191-24698191 on Build GRCh38
rs756545800 Chr.6: 24697883-24697883 on Build GRCh38
rs528877421 Chr.6: 24698068-24698069 on Build GRCh38
rs779233647 Chr.6: 24698034-24698034 on Build GRCh38
rs566849273 Chr.6: 24697994-24697994 on Build GRCh38
rs796467407 Chr.6: 24697865-24697866 on Build GRCh38
rs760831059 Chr.6: 24698020-24698020 on Build GRCh38
rs910074333 Chr.6: 24697856-24697856 on Build GRCh38
rs1049985068 Chr.6: 24698261-24698261 on Build GRCh38
rs750429212 Chr.6: 24698017-24698017 on Build GRCh38
rs1015992324 Chr.6: 24698033-24698033 on Build GRCh38
rs768118089 Chr.6: 24697862-24697862 on Build GRCh38

View Full Product Details