Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 153438110-153438615 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
S100A7L2
Entrez Gene ID
645922
Gene Name
S100 calcium binding protein A7 like 2
Gene Aliases
S100a7b
UniGene
Hs.647713
Gene Chromosome Location
Chr.1: 153436995-153440027 on Build GRCh38
Transcripts
NM_001045479.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
896350 c.73G>T p.V25F Chr.1: 153438290-153438290 on Build GRCh38
284756 c.40G>T p.V14F Chr.1: 153438290-153438290 on Build GRCh38
4526795 c.141G>A p.M47I Chr.1: 153438222-153438222 on Build GRCh38
6271367 c.89C>A p.P30Q Chr.1: 153438241-153438241 on Build GRCh38
4441563 c.77C>T p.A26V Chr.1: 153438286-153438286 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs943172702 Chr.1: 153438319-153438319 on Build GRCh38
rs115100684 Chr.1: 153438227-153438227 on Build GRCh38
rs140012440 Chr.1: 153438278-153438278 on Build GRCh38
rs112403134 Chr.1: 153438288-153438288 on Build GRCh38
rs775614251 Chr.1: 153438514-153438514 on Build GRCh38
rs967256232 Chr.1: 153438511-153438511 on Build GRCh38
rs749322202 Chr.1: 153438297-153438297 on Build GRCh38
rs752092277 Chr.1: 153438285-153438285 on Build GRCh38
rs542193205 Chr.1: 153438203-153438203 on Build GRCh38
rs765976310 Chr.1: 153438362-153438362 on Build GRCh38
rs755339990 Chr.1: 153438202-153438202 on Build GRCh38
rs1008607128 Chr.1: 153438494-153438494 on Build GRCh38
rs10888561 Chr.1: 153438346-153438346 on Build GRCh38
rs116156022 Chr.1: 153438322-153438322 on Build GRCh38
rs766028222 Chr.1: 153438283-153438283 on Build GRCh38
rs74115439 Chr.1: 153438160-153438160 on Build GRCh38
rs1002372993 Chr.1: 153438532-153438532 on Build GRCh38
rs905616068 Chr.1: 153438476-153438476 on Build GRCh38
rs935964641 Chr.1: 153438370-153438370 on Build GRCh38
rs762745610 Chr.1: 153438359-153438359 on Build GRCh38
rs756300211 Chr.1: 153438292-153438292 on Build GRCh38
rs995125472 Chr.1: 153438545-153438545 on Build GRCh38
rs866066252 Chr.1: 153438179-153438179 on Build GRCh38
rs754034724 Chr.1: 153438394-153438394 on Build GRCh38
rs774776530 Chr.1: 153438266-153438266 on Build GRCh38
rs565264295 Chr.1: 153438268-153438268 on Build GRCh38
rs763924500 Chr.1: 153438238-153438238 on Build GRCh38
rs749773542 Chr.1: 153438163-153438163 on Build GRCh38
rs61742168 Chr.1: 153438301-153438301 on Build GRCh38
rs772970078 Chr.1: 153438353-153438353 on Build GRCh38
rs139863738 Chr.1: 153438207-153438207 on Build GRCh38
rs753056328 Chr.1: 153438390-153438390 on Build GRCh38
rs56106046 Chr.1: 153438382-153438382 on Build GRCh38
rs55672410 Chr.1: 153438239-153438239 on Build GRCh38
rs1020506777 Chr.1: 153438500-153438500 on Build GRCh38
rs767921885 Chr.1: 153438373-153438373 on Build GRCh38
rs749962040 Chr.1: 153438170-153438170 on Build GRCh38
rs890161682 Chr.1: 153438464-153438464 on Build GRCh38
rs764814686 Chr.1: 153438273-153438273 on Build GRCh38
rs1024314156 Chr.1: 153438579-153438579 on Build GRCh38
rs779010377 Chr.1: 153438317-153438317 on Build GRCh38
rs778147727 Chr.1: 153438393-153438393 on Build GRCh38
rs562529596 Chr.1: 153438541-153438541 on Build GRCh38
rs796760878 Chr.1: 153438275-153438275 on Build GRCh38
rs749908678 Chr.1: 153438277-153438277 on Build GRCh38
rs369177408 Chr.1: 153438286-153438286 on Build GRCh38
rs776433707 Chr.1: 153438328-153438328 on Build GRCh38
rs1027000606 Chr.1: 153438556-153438556 on Build GRCh38
rs769896747 Chr.1: 153438138-153438138 on Build GRCh38
rs745612821 Chr.1: 153438358-153438358 on Build GRCh38
rs538593753 Chr.1: 153438195-153438195 on Build GRCh38
rs763860135 Chr.1: 153438557-153438557 on Build GRCh38
rs991486408 Chr.1: 153438559-153438559 on Build GRCh38
rs773222063 Chr.1: 153438144-153438144 on Build GRCh38
rs760020482 Chr.1: 153438372-153438372 on Build GRCh38
rs780729086 Chr.1: 153438193-153438193 on Build GRCh38
rs145850471 Chr.1: 153438436-153438436 on Build GRCh38
rs188609987 Chr.1: 153438272-153438272 on Build GRCh38
rs117909437 Chr.1: 153438137-153438137 on Build GRCh38
rs61743588 Chr.1: 153438206-153438206 on Build GRCh38
rs970858737 Chr.1: 153438175-153438175 on Build GRCh38
rs199975815 Chr.1: 153438187-153438187 on Build GRCh38
rs764801146 Chr.1: 153438169-153438169 on Build GRCh38
rs867714831 Chr.1: 153438314-153438314 on Build GRCh38
rs781614358 Chr.1: 153438287-153438287 on Build GRCh38
rs746949636 Chr.1: 153438321-153438321 on Build GRCh38
rs147282626 Chr.1: 153438149-153438149 on Build GRCh38
rs911989113 Chr.1: 153438318-153438318 on Build GRCh38
rs574523317 Chr.1: 153438590-153438590 on Build GRCh38
rs768869936 Chr.1: 153438542-153438542 on Build GRCh38
rs571172133 Chr.1: 153438166-153438166 on Build GRCh38
rs1000459791 Chr.1: 153438517-153438517 on Build GRCh38
rs1024081505 Chr.1: 153438173-153438173 on Build GRCh38
rs760643509 Chr.1: 153438234-153438234 on Build GRCh38
rs866720700 Chr.1: 153438260-153438260 on Build GRCh38
rs749383032 Chr.1: 153438224-153438224 on Build GRCh38
rs987440903 Chr.1: 153438589-153438589 on Build GRCh38
rs760482040 Chr.1: 153438158-153438158 on Build GRCh38
rs758921339 Chr.1: 153438191-153438191 on Build GRCh38
rs751180548 Chr.1: 153438368-153438368 on Build GRCh38
rs558708581 Chr.1: 153438440-153438440 on Build GRCh38
rs756462105 Chr.1: 153438392-153438392 on Build GRCh38
rs377062054 Chr.1: 153438148-153438148 on Build GRCh38
rs775433023 Chr.1: 153438228-153438228 on Build GRCh38

View Full Product Details