Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 63679823-63680298 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PHF3
Entrez Gene ID
23469
Gene Name
PHD finger protein 3
Gene Aliases
-
UniGene
Hs.348921
Gene Chromosome Location
Chr.6: 63635802-63715522 on Build GRCh38
Transcripts
NM_001290259.1 NM_001290260.1 NM_015153.3 XM_005248701.3 XM_005248702.3 XM_005248703.3 XM_006715426.3 XM_011535648.2 XM_017010626.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7383740 c.253C>T p.L85F Chr.6: 63680008-63680008 on Build GRCh38
3777799 c.310G>A p.E104K Chr.6: 63680065-63680065 on Build GRCh38
211895 c.280G>T p.V94F Chr.6: 63680035-63680035 on Build GRCh38
4714007 c.301A>G p.T101A Chr.6: 63680056-63680056 on Build GRCh38
7203995 c.394T>G p.L132V Chr.6: 63680149-63680149 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs565834322 Chr.6: 63679996-63679996 on Build GRCh38
rs753791596 Chr.6: 63680080-63680080 on Build GRCh38
rs752850735 Chr.6: 63679983-63679983 on Build GRCh38
rs372565137 Chr.6: 63680077-63680077 on Build GRCh38
rs765683811 Chr.6: 63680091-63680091 on Build GRCh38
rs893080172 Chr.6: 63680144-63680144 on Build GRCh38
rs767811891 Chr.6: 63679961-63679961 on Build GRCh38
rs199924633 Chr.6: 63680047-63680047 on Build GRCh38
rs966192621 Chr.6: 63679867-63679867 on Build GRCh38
rs756613408 Chr.6: 63679894-63679894 on Build GRCh38
rs113209495 Chr.6: 63680249-63680249 on Build GRCh38
rs755661900 Chr.6: 63680120-63680120 on Build GRCh38
rs779252012 Chr.6: 63679992-63679992 on Build GRCh38
rs762792385 Chr.6: 63679937-63679937 on Build GRCh38
rs771019080 Chr.6: 63680133-63680133 on Build GRCh38
rs946720616 Chr.6: 63680269-63680269 on Build GRCh38
rs200825087 Chr.6: 63680014-63680014 on Build GRCh38
rs755824915 Chr.6: 63680204-63680204 on Build GRCh38
rs938253250 Chr.6: 63679939-63679939 on Build GRCh38
rs775135145 Chr.6: 63680147-63680147 on Build GRCh38
rs777484913 Chr.6: 63679858-63679859 on Build GRCh38
rs776793128 Chr.6: 63680180-63680180 on Build GRCh38
rs773237156 Chr.6: 63679932-63679932 on Build GRCh38
rs114299749 Chr.6: 63679958-63679958 on Build GRCh38
rs767398831 Chr.6: 63680063-63680063 on Build GRCh38
rs764050302 Chr.6: 63679988-63679988 on Build GRCh38
rs776150089 Chr.6: 63679915-63679915 on Build GRCh38
rs758797993 Chr.6: 63680098-63680098 on Build GRCh38
rs766250928 Chr.6: 63679950-63679950 on Build GRCh38
rs777343593 Chr.6: 63680126-63680126 on Build GRCh38
rs770929256 Chr.6: 63680051-63680051 on Build GRCh38
rs748710464 Chr.6: 63680131-63680131 on Build GRCh38
rs200623505 Chr.6: 63679998-63679998 on Build GRCh38
rs1032830708 Chr.6: 63680071-63680071 on Build GRCh38
rs368894996 Chr.6: 63680013-63680013 on Build GRCh38
rs374757388 Chr.6: 63679951-63679951 on Build GRCh38
rs761846545 Chr.6: 63680181-63680181 on Build GRCh38
rs573232742 Chr.6: 63680064-63680064 on Build GRCh38
rs746189779 Chr.6: 63680008-63680008 on Build GRCh38
rs776145885 Chr.6: 63680192-63680193 on Build GRCh38
rs756267185 Chr.6: 63679987-63679987 on Build GRCh38
rs912140290 Chr.6: 63680185-63680185 on Build GRCh38
rs749735870 Chr.6: 63679898-63679898 on Build GRCh38
rs747889572 Chr.6: 63680112-63680112 on Build GRCh38
rs778294095 Chr.6: 63679895-63679895 on Build GRCh38
rs139169906 Chr.6: 63680067-63680067 on Build GRCh38
rs1007477831 Chr.6: 63680244-63680244 on Build GRCh38
rs915276611 Chr.6: 63680259-63680259 on Build GRCh38
rs745971726 Chr.6: 63680109-63680111 on Build GRCh38
rs369713410 Chr.6: 63680208-63680208 on Build GRCh38
rs760967733 Chr.6: 63680165-63680165 on Build GRCh38
rs373123753 Chr.6: 63680055-63680055 on Build GRCh38
rs761262914 Chr.6: 63679919-63679919 on Build GRCh38
rs943675462 Chr.6: 63680193-63680193 on Build GRCh38
rs780389096 Chr.6: 63680109-63680109 on Build GRCh38
rs368685403 Chr.6: 63680146-63680146 on Build GRCh38
rs371765931 Chr.6: 63680173-63680173 on Build GRCh38
rs763433944 Chr.6: 63680198-63680198 on Build GRCh38
rs752762643 Chr.6: 63679889-63679889 on Build GRCh38
rs779246050 Chr.6: 63679906-63679906 on Build GRCh38
rs950846062 Chr.6: 63680095-63680095 on Build GRCh38
rs765270327 Chr.6: 63680187-63680187 on Build GRCh38
rs769160520 Chr.6: 63679921-63679921 on Build GRCh38
rs774483403 Chr.6: 63680139-63680139 on Build GRCh38
rs781438616 Chr.6: 63679999-63680001 on Build GRCh38
rs113423383 Chr.6: 63679888-63679888 on Build GRCh38
rs754436055 Chr.6: 63679989-63679989 on Build GRCh38
rs768228799 Chr.6: 63679913-63679913 on Build GRCh38
rs745768942 Chr.6: 63680142-63680142 on Build GRCh38
rs757812520 Chr.6: 63680087-63680087 on Build GRCh38
rs757464030 Chr.6: 63679978-63679978 on Build GRCh38
rs74760146 Chr.6: 63680247-63680247 on Build GRCh38
rs375655162 Chr.6: 63680274-63680274 on Build GRCh38
rs746794283 Chr.6: 63679977-63679978 on Build GRCh38
rs750992001 Chr.6: 63680196-63680196 on Build GRCh38
rs774322145 Chr.6: 63680054-63680054 on Build GRCh38
rs751802156 Chr.6: 63680200-63680200 on Build GRCh38
rs201838701 Chr.6: 63680199-63680199 on Build GRCh38
rs758551301 Chr.6: 63680105-63680105 on Build GRCh38
rs764295347 Chr.6: 63680069-63680069 on Build GRCh38
rs772580867 Chr.6: 63680045-63680045 on Build GRCh38
rs750907046 Chr.6: 63680093-63680093 on Build GRCh38
rs771368510 Chr.6: 63679905-63679905 on Build GRCh38
rs769941007 Chr.6: 63680188-63680189 on Build GRCh38

View Full Product Details