Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 83239300-83239802 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ME1
Entrez Gene ID
4199
Gene Name
malic enzyme 1
Gene Aliases
HUMNDME, MES
UniGene
Hs.21160
Gene Chromosome Location
Chr.6: 83210389-83431219 on Build GRCh38
Transcripts
NM_002395.5 XM_011535836.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1082064 c.883C>A p.Q295K Chr.6: 83239568-83239568 on Build GRCh38
5820855 c.857G>A p.R286Q Chr.6: 83239594-83239594 on Build GRCh38
7336927 c.815G>A p.G272E Chr.6: 83239636-83239636 on Build GRCh38
4510638 c.843C>T p.L281L Chr.6: 83239608-83239608 on Build GRCh38
5618763 c.909A>G p.G303G Chr.6: 83239542-83239542 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs568329512 Chr.6: 83239490-83239490 on Build GRCh38
rs970339043 Chr.6: 83239336-83239336 on Build GRCh38
rs1012352641 Chr.6: 83239653-83239653 on Build GRCh38
rs1035347428 Chr.6: 83239365-83239365 on Build GRCh38
rs532466286 Chr.6: 83239342-83239342 on Build GRCh38
rs1039299124 Chr.6: 83239377-83239377 on Build GRCh38
rs771564214 Chr.6: 83239574-83239574 on Build GRCh38
rs755839453 Chr.6: 83239639-83239639 on Build GRCh38
rs879075341 Chr.6: 83239396-83239396 on Build GRCh38
rs943590981 Chr.6: 83239333-83239333 on Build GRCh38
rs112829172 Chr.6: 83239331-83239331 on Build GRCh38
rs576075956 Chr.6: 83239604-83239604 on Build GRCh38
rs559980514 Chr.6: 83239357-83239357 on Build GRCh38
rs988539521 Chr.6: 83239424-83239424 on Build GRCh38
rs752555482 Chr.6: 83239625-83239625 on Build GRCh38
rs548309183 Chr.6: 83239481-83239481 on Build GRCh38
rs192318688 Chr.6: 83239358-83239358 on Build GRCh38
rs373050038 Chr.6: 83239687-83239687 on Build GRCh38
rs1044587675 Chr.6: 83239679-83239679 on Build GRCh38
rs769229515 Chr.6: 83239584-83239584 on Build GRCh38
rs955730718 Chr.6: 83239394-83239394 on Build GRCh38
rs35077452 Chr.6: 83239590-83239591 on Build GRCh38
rs764129992 Chr.6: 83239656-83239656 on Build GRCh38
rs533873402 Chr.6: 83239505-83239505 on Build GRCh38
rs888503722 Chr.6: 83239621-83239621 on Build GRCh38
rs9444054 Chr.6: 83239778-83239778 on Build GRCh38
rs775926006 Chr.6: 83239575-83239575 on Build GRCh38
rs753099067 Chr.6: 83239688-83239688 on Build GRCh38
rs756884032 Chr.6: 83239673-83239673 on Build GRCh38
rs9689848 Chr.6: 83239705-83239705 on Build GRCh38
rs745501186 Chr.6: 83239571-83239571 on Build GRCh38
rs769082392 Chr.6: 83239346-83239346 on Build GRCh38
rs766448099 Chr.6: 83239612-83239612 on Build GRCh38
rs531703700 Chr.6: 83239477-83239477 on Build GRCh38
rs776724282 Chr.6: 83239515-83239515 on Build GRCh38
rs778578317 Chr.6: 83239675-83239675 on Build GRCh38
rs1180230 Chr.6: 83239542-83239542 on Build GRCh38
rs777100717 Chr.6: 83239588-83239588 on Build GRCh38
rs746728608 Chr.6: 83239761-83239761 on Build GRCh38
rs372257465 Chr.6: 83239557-83239557 on Build GRCh38
rs764441041 Chr.6: 83239511-83239511 on Build GRCh38
rs188352843 Chr.6: 83239721-83239721 on Build GRCh38
rs753820374 Chr.6: 83239660-83239660 on Build GRCh38
rs150412840 Chr.6: 83239459-83239459 on Build GRCh38
rs2273135 Chr.6: 83239525-83239525 on Build GRCh38
rs747297978 Chr.6: 83239576-83239576 on Build GRCh38
rs745377867 Chr.6: 83239545-83239545 on Build GRCh38
rs369555353 Chr.6: 83239620-83239620 on Build GRCh38
rs756792249 Chr.6: 83239564-83239564 on Build GRCh38
rs774994131 Chr.6: 83239709-83239709 on Build GRCh38
rs2230901 Chr.6: 83239566-83239566 on Build GRCh38
rs1002799310 Chr.6: 83239337-83239338 on Build GRCh38
rs758100516 Chr.6: 83239684-83239684 on Build GRCh38
rs367778451 Chr.6: 83239594-83239594 on Build GRCh38
rs539655823 Chr.6: 83239549-83239549 on Build GRCh38
rs376254621 Chr.6: 83239599-83239599 on Build GRCh38
rs756085813 Chr.6: 83239554-83239554 on Build GRCh38
rs757806681 Chr.6: 83239458-83239458 on Build GRCh38
rs368485997 Chr.6: 83239519-83239519 on Build GRCh38
rs765564249 Chr.6: 83239498-83239498 on Build GRCh38
rs745309128 Chr.6: 83239681-83239681 on Build GRCh38
rs374822298 Chr.6: 83239558-83239558 on Build GRCh38

View Full Product Details