Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 116584506-116584906 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RWDD1
Entrez Gene ID
51389
Gene Name
RWD domain containing 1
Gene Aliases
CGI-24, PTD013
UniGene
Hs.532164
Gene Chromosome Location
Chr.6: 116571367-116593601 on Build GRCh38
Transcripts
NM_001007464.2 NM_015952.3 NM_016104.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4889690 c.187G>A p.E63K Chr.6: 116584774-116584774 on Build GRCh38
5979253 c.166T>C p.Y56H Chr.6: 116584753-116584753 on Build GRCh38
7321078 c.213C>T p.S71S Chr.6: 116584800-116584800 on Build GRCh38
739556 c.262G>C p.A88P Chr.6: 116584849-116584849 on Build GRCh38
3619687 c.145C>T p.Q49* Chr.6: 116584732-116584732 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749253769 Chr.6: 116584690-116584691 on Build GRCh38
rs144557301 Chr.6: 116584772-116584772 on Build GRCh38
rs200538980 Chr.6: 116584788-116584788 on Build GRCh38
rs776697305 Chr.6: 116584874-116584874 on Build GRCh38
rs536468127 Chr.6: 116584671-116584671 on Build GRCh38
rs886218492 Chr.6: 116584601-116584601 on Build GRCh38
rs139759428 Chr.6: 116584544-116584544 on Build GRCh38
rs779162602 Chr.6: 116584823-116584823 on Build GRCh38
rs369171229 Chr.6: 116584827-116584827 on Build GRCh38
rs761710819 Chr.6: 116584730-116584730 on Build GRCh38
rs930696310 Chr.6: 116584670-116584670 on Build GRCh38
rs372495277 Chr.6: 116584710-116584710 on Build GRCh38
rs576266834 Chr.6: 116584753-116584753 on Build GRCh38
rs1028573539 Chr.6: 116584691-116584691 on Build GRCh38
rs754219454 Chr.6: 116584575-116584575 on Build GRCh38
rs565866473 Chr.6: 116584649-116584649 on Build GRCh38
rs369498026 Chr.6: 116584724-116584724 on Build GRCh38
rs778381800 Chr.6: 116584792-116584792 on Build GRCh38
rs138012483 Chr.6: 116584832-116584832 on Build GRCh38
rs760568012 Chr.6: 116584709-116584709 on Build GRCh38
rs755255477 Chr.6: 116584759-116584759 on Build GRCh38
rs901403241 Chr.6: 116584644-116584644 on Build GRCh38
rs767662635 Chr.6: 116584768-116584768 on Build GRCh38
rs961170536 Chr.6: 116584558-116584558 on Build GRCh38
rs1043483648 Chr.6: 116584839-116584839 on Build GRCh38
rs920665362 Chr.6: 116584635-116584635 on Build GRCh38
rs756663419 Chr.6: 116584779-116584779 on Build GRCh38
rs1000231788 Chr.6: 116584729-116584729 on Build GRCh38
rs554336313 Chr.6: 116584639-116584639 on Build GRCh38
rs774935546 Chr.6: 116584844-116584844 on Build GRCh38
rs1056614791 Chr.6: 116584821-116584821 on Build GRCh38
rs376426771 Chr.6: 116584714-116584714 on Build GRCh38
rs774990586 Chr.6: 116584696-116584696 on Build GRCh38
rs559036012 Chr.6: 116584856-116584856 on Build GRCh38
rs201461238 Chr.6: 116584755-116584755 on Build GRCh38
rs768420550 Chr.6: 116584833-116584834 on Build GRCh38
rs771516596 Chr.6: 116584689-116584689 on Build GRCh38
rs768448993 Chr.6: 116584863-116584863 on Build GRCh38
rs146150369 Chr.6: 116584810-116584810 on Build GRCh38
rs945531556 Chr.6: 116584828-116584828 on Build GRCh38
rs745365193 Chr.6: 116584686-116584686 on Build GRCh38
rs779501059 Chr.6: 116584682-116584683 on Build GRCh38
rs554800049 Chr.6: 116584674-116584674 on Build GRCh38
rs757694143 Chr.6: 116584815-116584815 on Build GRCh38

View Full Product Details