Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 118154502-118155001 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC35F1
Entrez Gene ID
222553
Gene Name
solute carrier family 35 member F1
Gene Aliases
C6orf169, dJ230I3.1
UniGene
Hs.654841
Gene Chromosome Location
Chr.6: 117907226-118317676 on Build GRCh38
Transcripts
NM_001029858.3 XM_005266865.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2148957 c.237_239delGAC p.L79_T80>F Chr.6: 118154508-118154510 on Build GRCh38
4449848 c.349+1G>A p.? Chr.6: 118154621-118154621 on Build GRCh38
6781755 c.283T>C p.F95L Chr.6: 118154554-118154554 on Build GRCh38
1754459 c.268G>A p.A90T Chr.6: 118154539-118154539 on Build GRCh38
7268001 c.349G>T p.G117* Chr.6: 118154620-118154620 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746046282 Chr.6: 118154552-118154552 on Build GRCh38
rs765930142 Chr.6: 118154642-118154642 on Build GRCh38
rs762617944 Chr.6: 118154639-118154639 on Build GRCh38
rs17079793 Chr.6: 118154693-118154693 on Build GRCh38
rs886780587 Chr.6: 118154630-118154630 on Build GRCh38
rs369676363 Chr.6: 118154842-118154842 on Build GRCh38
rs1033934583 Chr.6: 118154953-118154953 on Build GRCh38
rs754996434 Chr.6: 118154525-118154525 on Build GRCh38
rs553090270 Chr.6: 118154758-118154758 on Build GRCh38
rs181683911 Chr.6: 118154911-118154911 on Build GRCh38
rs773031487 Chr.6: 118154638-118154638 on Build GRCh38
rs747028312 Chr.6: 118154598-118154598 on Build GRCh38
rs188952375 Chr.6: 118154760-118154760 on Build GRCh38
rs1040138344 Chr.6: 118154823-118154823 on Build GRCh38
rs142724842 Chr.6: 118154877-118154877 on Build GRCh38
rs564179595 Chr.6: 118154966-118154966 on Build GRCh38
rs558953428 Chr.6: 118154689-118154689 on Build GRCh38
rs779108571 Chr.6: 118154544-118154544 on Build GRCh38
rs758425765 Chr.6: 118154556-118154556 on Build GRCh38
rs116164312 Chr.6: 118154694-118154694 on Build GRCh38
rs114778047 Chr.6: 118154660-118154660 on Build GRCh38
rs752561766 Chr.6: 118154662-118154662 on Build GRCh38
rs777263061 Chr.6: 118154610-118154610 on Build GRCh38
rs755324516 Chr.6: 118154664-118154665 on Build GRCh38
rs999353420 Chr.6: 118154882-118154882 on Build GRCh38
rs541848786 Chr.6: 118154812-118154812 on Build GRCh38
rs205966 Chr.6: 118154635-118154635 on Build GRCh38
rs778024039 Chr.6: 118154722-118154722 on Build GRCh38
rs892175862 Chr.6: 118154837-118154837 on Build GRCh38
rs755931244 Chr.6: 118154668-118154668 on Build GRCh38
rs943529018 Chr.6: 118154570-118154570 on Build GRCh38
rs770218171 Chr.6: 118154636-118154636 on Build GRCh38
rs529939260 Chr.6: 118154526-118154526 on Build GRCh38
rs756699037 Chr.6: 118154542-118154542 on Build GRCh38
rs780230170 Chr.6: 118154587-118154587 on Build GRCh38
rs1009219671 Chr.6: 118154937-118154937 on Build GRCh38
rs1024597694 Chr.6: 118154755-118154755 on Build GRCh38
rs376082648 Chr.6: 118154543-118154543 on Build GRCh38
rs185770023 Chr.6: 118154659-118154659 on Build GRCh38
rs758596898 Chr.6: 118154641-118154641 on Build GRCh38
rs140354760 Chr.6: 118154845-118154845 on Build GRCh38
rs752558083 Chr.6: 118154539-118154539 on Build GRCh38
rs773955160 Chr.6: 118154647-118154647 on Build GRCh38
rs1017603006 Chr.6: 118154951-118154951 on Build GRCh38
rs1022241287 Chr.6: 118154945-118154945 on Build GRCh38
rs960496817 Chr.6: 118154961-118154961 on Build GRCh38
rs754335489 Chr.6: 118154665-118154665 on Build GRCh38
rs546100068 Chr.6: 118154921-118154921 on Build GRCh38
rs766805968 Chr.6: 118154644-118154645 on Build GRCh38
rs147940941 Chr.6: 118154538-118154538 on Build GRCh38
rs976683206 Chr.6: 118154583-118154583 on Build GRCh38
rs1031718683 Chr.6: 118154891-118154891 on Build GRCh38
rs769270401 Chr.6: 118154603-118154603 on Build GRCh38
rs990119474 Chr.6: 118154920-118154920 on Build GRCh38

View Full Product Details