Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 154507062-154507488 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TDRD10
Entrez Gene ID
126668
Gene Name
tudor domain containing 10
Gene Aliases
-
UniGene
Hs.387671
Gene Chromosome Location
Chr.1: 154502219-154548147 on Build GRCh38
Transcripts
NM_001098475.1 NM_182499.3 XM_011509152.1 XM_011509153.1 XM_011509154.1 XM_011509155.1 XM_011509156.1 XM_011509157.1 XM_011509162.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
896615 c.22C>T p.P8S Chr.1: 154507260-154507260 on Build GRCh38
4599952 c.35A>C p.D12A Chr.1: 154507273-154507273 on Build GRCh38
3399790 c.42G>C p.L14L Chr.1: 154507280-154507280 on Build GRCh38
5716146 c.24C>T p.P8P Chr.1: 154507262-154507262 on Build GRCh38
7259156 c.21delC p.Q9fs*46 Chr.1: 154507259-154507259 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs7524349 Chr.1: 154507458-154507458 on Build GRCh38
rs111557274 Chr.1: 154507182-154507182 on Build GRCh38
rs868805517 Chr.1: 154507317-154507317 on Build GRCh38
rs752385441 Chr.1: 154507306-154507306 on Build GRCh38
rs775942160 Chr.1: 154507346-154507346 on Build GRCh38
rs762623633 Chr.1: 154507100-154507100 on Build GRCh38
rs774833353 Chr.1: 154507208-154507208 on Build GRCh38
rs779443554 Chr.1: 154507330-154507330 on Build GRCh38
rs368614573 Chr.1: 154507099-154507099 on Build GRCh38
rs944661572 Chr.1: 154507123-154507123 on Build GRCh38
rs376965293 Chr.1: 154507195-154507195 on Build GRCh38
rs934546739 Chr.1: 154507183-154507183 on Build GRCh38
rs747273917 Chr.1: 154507358-154507358 on Build GRCh38
rs1027026368 Chr.1: 154507398-154507398 on Build GRCh38
rs201533177 Chr.1: 154507205-154507205 on Build GRCh38
rs554594311 Chr.1: 154507320-154507320 on Build GRCh38
rs772400955 Chr.1: 154507216-154507216 on Build GRCh38
rs35092505 Chr.1: 154507182-154507183 on Build GRCh38
rs370913635 Chr.1: 154507233-154507233 on Build GRCh38
rs149167908 Chr.1: 154507387-154507387 on Build GRCh38
rs756745836 Chr.1: 154507321-154507321 on Build GRCh38
rs140425454 Chr.1: 154507180-154507181 on Build GRCh38
rs751522169 Chr.1: 154507275-154507275 on Build GRCh38
rs1003059084 Chr.1: 154507257-154507257 on Build GRCh38
rs767452173 Chr.1: 154507303-154507303 on Build GRCh38
rs753943316 Chr.1: 154507406-154507406 on Build GRCh38
rs911700497 Chr.1: 154507102-154507102 on Build GRCh38
rs1024502542 Chr.1: 154507273-154507273 on Build GRCh38
rs764169226 Chr.1: 154507224-154507224 on Build GRCh38
rs532097724 Chr.1: 154507141-154507141 on Build GRCh38
rs775611605 Chr.1: 154507217-154507217 on Build GRCh38
rs1052825810 Chr.1: 154507187-154507187 on Build GRCh38
rs1039380122 Chr.1: 154507171-154507171 on Build GRCh38
rs373250153 Chr.1: 154507342-154507342 on Build GRCh38
rs920346721 Chr.1: 154507094-154507094 on Build GRCh38
rs748688060 Chr.1: 154507198-154507198 on Build GRCh38
rs761154815 Chr.1: 154507279-154507282 on Build GRCh38
rs1010726875 Chr.1: 154507244-154507244 on Build GRCh38
rs1054459544 Chr.1: 154507204-154507204 on Build GRCh38
rs757151932 Chr.1: 154507280-154507280 on Build GRCh38
rs78947330 Chr.1: 154507181-154507181 on Build GRCh38
rs962895971 Chr.1: 154507391-154507391 on Build GRCh38
rs766484327 Chr.1: 154507236-154507236 on Build GRCh38
rs920251864 Chr.1: 154507414-154507414 on Build GRCh38
rs6664608 Chr.1: 154507194-154507194 on Build GRCh38
rs755804330 Chr.1: 154507307-154507307 on Build GRCh38
rs777223802 Chr.1: 154507318-154507318 on Build GRCh38
rs1001682984 Chr.1: 154507322-154507322 on Build GRCh38
rs772525090 Chr.1: 154507343-154507343 on Build GRCh38
rs10654618 Chr.1: 154507181-154507182 on Build GRCh38
rs759948700 Chr.1: 154507215-154507215 on Build GRCh38
rs781058614 Chr.1: 154507289-154507289 on Build GRCh38
rs552227009 Chr.1: 154507097-154507097 on Build GRCh38

View Full Product Details