Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 154544610-154545124 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TDRD10
Entrez Gene ID
126668
Gene Name
tudor domain containing 10
Gene Aliases
-
UniGene
Hs.387671
Gene Chromosome Location
Chr.1: 154502219-154548147 on Build GRCh38
Transcripts
NM_001098475.1 NM_182499.3 XM_006711156.2 XM_011509152.1 XM_011509153.1 XM_011509154.1 XM_011509155.1 XM_011509156.1 XM_011509157.1 XM_011509158.2 XM_011509159.2 XM_011509160.2 XM_011509161.1 XM_017000255.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4022819 c.904G>A p.D302N Chr.1: 154544901-154544901 on Build GRCh38
5727853 c.923C>T p.P308L Chr.1: 154544920-154544920 on Build GRCh38
377105 c.847G>A p.D283N Chr.1: 154544844-154544844 on Build GRCh38
6533896 c.883C>G p.L295V Chr.1: 154544880-154544880 on Build GRCh38
6480005 c.842T>A p.F281Y Chr.1: 154544839-154544839 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs952534836 Chr.1: 154545091-154545091 on Build GRCh38
rs187309512 Chr.1: 154544703-154544703 on Build GRCh38
rs6681207 Chr.1: 154544741-154544741 on Build GRCh38
rs768688702 Chr.1: 154545000-154545000 on Build GRCh38
rs565242590 Chr.1: 154545040-154545040 on Build GRCh38
rs764503176 Chr.1: 154544826-154544827 on Build GRCh38
rs754055846 Chr.1: 154544823-154544823 on Build GRCh38
rs200586203 Chr.1: 154544905-154544905 on Build GRCh38
rs199789514 Chr.1: 154544749-154544749 on Build GRCh38
rs944068596 Chr.1: 154544689-154544689 on Build GRCh38
rs753177917 Chr.1: 154544784-154544784 on Build GRCh38
rs776981966 Chr.1: 154544838-154544841 on Build GRCh38
rs748993679 Chr.1: 154544898-154544898 on Build GRCh38
rs781098173 Chr.1: 154544890-154544890 on Build GRCh38
rs371839272 Chr.1: 154544976-154544976 on Build GRCh38
rs1042658644 Chr.1: 154544836-154544836 on Build GRCh38
rs201425979 Chr.1: 154544895-154544895 on Build GRCh38
rs776109690 Chr.1: 154544768-154544769 on Build GRCh38
rs142034665 Chr.1: 154544921-154544921 on Build GRCh38
rs765444138 Chr.1: 154544969-154544969 on Build GRCh38
rs1056639889 Chr.1: 154544893-154544893 on Build GRCh38
rs376302248 Chr.1: 154545033-154545033 on Build GRCh38
rs768067970 Chr.1: 154544782-154544782 on Build GRCh38
rs781147506 Chr.1: 154544984-154544984 on Build GRCh38
rs752715022 Chr.1: 154544994-154544994 on Build GRCh38
rs369240196 Chr.1: 154544915-154544915 on Build GRCh38
rs887887273 Chr.1: 154545034-154545034 on Build GRCh38
rs551229256 Chr.1: 154545095-154545095 on Build GRCh38
rs773951849 Chr.1: 154544770-154544770 on Build GRCh38
rs1006690456 Chr.1: 154545101-154545101 on Build GRCh38
rs922287761 Chr.1: 154544633-154544633 on Build GRCh38
rs866123484 Chr.1: 154544936-154544936 on Build GRCh38
rs376156637 Chr.1: 154544773-154544773 on Build GRCh38
rs762550870 Chr.1: 154544958-154544962 on Build GRCh38
rs3811449 Chr.1: 154545028-154545028 on Build GRCh38
rs774000552 Chr.1: 154544901-154544901 on Build GRCh38
rs371525896 Chr.1: 154544897-154544897 on Build GRCh38
rs149740279 Chr.1: 154544756-154544756 on Build GRCh38
rs552932483 Chr.1: 154544709-154544709 on Build GRCh38
rs760135550 Chr.1: 154544778-154544778 on Build GRCh38
rs754978467 Chr.1: 154544885-154544885 on Build GRCh38
rs770651464 Chr.1: 154544900-154544900 on Build GRCh38
rs371217983 Chr.1: 154544785-154544785 on Build GRCh38
rs373715394 Chr.1: 154544944-154544944 on Build GRCh38
rs776117309 Chr.1: 154544954-154544954 on Build GRCh38
rs906755090 Chr.1: 154544883-154544883 on Build GRCh38
rs201929665 Chr.1: 154544755-154544755 on Build GRCh38
rs199739020 Chr.1: 154544884-154544884 on Build GRCh38
rs1041403538 Chr.1: 154544739-154544739 on Build GRCh38
rs762106736 Chr.1: 154544966-154544966 on Build GRCh38
rs532416947 Chr.1: 154545042-154545042 on Build GRCh38
rs141772431 Chr.1: 154544853-154544853 on Build GRCh38
rs764547030 Chr.1: 154544959-154544959 on Build GRCh38
rs778983751 Chr.1: 154544880-154544880 on Build GRCh38
rs760946336 Chr.1: 154544769-154544772 on Build GRCh38
rs764271763 Chr.1: 154544822-154544822 on Build GRCh38
rs200680670 Chr.1: 154544964-154544964 on Build GRCh38
rs765947024 Chr.1: 154544762-154544762 on Build GRCh38
rs769601109 Chr.1: 154544896-154544896 on Build GRCh38
rs908424568 Chr.1: 154544653-154544653 on Build GRCh38
rs575187113 Chr.1: 154544828-154544828 on Build GRCh38
rs367857151 Chr.1: 154544974-154544974 on Build GRCh38
rs988177450 Chr.1: 154544727-154544727 on Build GRCh38
rs745469529 Chr.1: 154544744-154544744 on Build GRCh38

View Full Product Details