Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 155010132-155010625 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZBTB7B
Entrez Gene ID
51043
Gene Name
zinc finger and BTB domain containing 7B
Gene Aliases
CKROX, THPOK, ZBTB15, ZFP-67, ZFP67, ZNF857B, c-KROX, hcKROX
UniGene
Hs.729279
Gene Chromosome Location
Chr.1: 155002628-155018525 on Build GRCh38
Transcripts
NM_001252406.2 NM_001256455.1 NR_045515.1 NR_046206.1 NR_049765.1 XM_006711349.2 XM_006711353.1 XM_006711354.1 XM_006711356.3 XM_006711357.1 XM_006711358.1 XM_006711359.2 XM_011509598.2 XM_011509599.2 XM_017001399.1

SNP Detail

SNP ID Chromosome Location
rs913782283 Chr.1: 155010450-155010450 on Build GRCh38
rs974795733 Chr.1: 155010466-155010466 on Build GRCh38
rs920583567 Chr.1: 155010602-155010602 on Build GRCh38
rs913729846 Chr.1: 155010156-155010156 on Build GRCh38
rs996933939 Chr.1: 155010409-155010409 on Build GRCh38
rs1016254332 Chr.1: 155010402-155010402 on Build GRCh38
rs970976377 Chr.1: 155010491-155010491 on Build GRCh38
rs949511029 Chr.1: 155010175-155010175 on Build GRCh38
rs148500443 Chr.1: 155010572-155010572 on Build GRCh38
rs904223758 Chr.1: 155010289-155010289 on Build GRCh38
rs540470056 Chr.1: 155010596-155010596 on Build GRCh38
rs938025200 Chr.1: 155010266-155010266 on Build GRCh38
rs146295378 Chr.1: 155010214-155010214 on Build GRCh38
rs1031949800 Chr.1: 155010422-155010422 on Build GRCh38
rs571697089 Chr.1: 155010246-155010246 on Build GRCh38
rs899441066 Chr.1: 155010325-155010325 on Build GRCh38
rs1047956323 Chr.1: 155010341-155010341 on Build GRCh38
rs758464160 Chr.1: 155010375-155010375 on Build GRCh38
rs933523296 Chr.1: 155010199-155010199 on Build GRCh38
rs1019788492 Chr.1: 155010376-155010376 on Build GRCh38
rs890623331 Chr.1: 155010355-155010355 on Build GRCh38
rs1001219330 Chr.1: 155010351-155010351 on Build GRCh38
rs750458151 Chr.1: 155010319-155010319 on Build GRCh38
rs913494823 Chr.1: 155010227-155010227 on Build GRCh38
rs957976945 Chr.1: 155010425-155010425 on Build GRCh38
rs945678713 Chr.1: 155010262-155010262 on Build GRCh38
rs1038138694 Chr.1: 155010575-155010575 on Build GRCh38
rs1037685565 Chr.1: 155010298-155010298 on Build GRCh38
rs536216310 Chr.1: 155010506-155010506 on Build GRCh38
rs963799330 Chr.1: 155010384-155010384 on Build GRCh38
rs767633420 Chr.1: 155010318-155010318 on Build GRCh38
rs374058863 Chr.1: 155010379-155010379 on Build GRCh38
rs1046921344 Chr.1: 155010209-155010209 on Build GRCh38
rs556970459 Chr.1: 155010383-155010383 on Build GRCh38
rs112344141 Chr.1: 155010560-155010560 on Build GRCh38
rs993739932 Chr.1: 155010340-155010340 on Build GRCh38
rs1002784206 Chr.1: 155010483-155010483 on Build GRCh38
rs538737447 Chr.1: 155010258-155010258 on Build GRCh38
rs371106985 Chr.1: 155010295-155010295 on Build GRCh38
rs926720442 Chr.1: 155010525-155010525 on Build GRCh38
rs568888521 Chr.1: 155010437-155010437 on Build GRCh38
rs963401390 Chr.1: 155010462-155010462 on Build GRCh38
rs922077257 Chr.1: 155010180-155010180 on Build GRCh38
rs988016374 Chr.1: 155010434-155010434 on Build GRCh38

View Full Product Details