Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 155912903-155913438 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA0907
Entrez Gene ID
22889
Gene Name
KIAA0907
Gene Aliases
BLOM7
UniGene
Hs.24656
Gene Chromosome Location
Chr.1: 155913043-155934442 on Build GRCh38
Transcripts
NM_014949.3

SNP Detail

SNP ID Chromosome Location
rs559036349 Chr.1: 155912941-155912941 on Build GRCh38
rs888054081 Chr.1: 155913129-155913129 on Build GRCh38
rs914443114 Chr.1: 155913393-155913394 on Build GRCh38
rs756687145 Chr.1: 155913357-155913358 on Build GRCh38
rs1026247897 Chr.1: 155913069-155913069 on Build GRCh38
rs949639764 Chr.1: 155913399-155913399 on Build GRCh38
rs963906626 Chr.1: 155913254-155913254 on Build GRCh38
rs975619828 Chr.1: 155913277-155913277 on Build GRCh38
rs899219759 Chr.1: 155913032-155913032 on Build GRCh38
rs57810844 Chr.1: 155912969-155912969 on Build GRCh38
rs747241214 Chr.1: 155913034-155913034 on Build GRCh38
rs976164394 Chr.1: 155913345-155913345 on Build GRCh38
rs1019907247 Chr.1: 155913238-155913238 on Build GRCh38
rs564079590 Chr.1: 155912959-155912959 on Build GRCh38
rs1037782087 Chr.1: 155913030-155913030 on Build GRCh38
rs957751911 Chr.1: 155913326-155913326 on Build GRCh38
rs41264985 Chr.1: 155913307-155913307 on Build GRCh38
rs529516575 Chr.1: 155913228-155913228 on Build GRCh38
rs982382338 Chr.1: 155913412-155913412 on Build GRCh38
rs1044080263 Chr.1: 155912961-155912961 on Build GRCh38
rs965098967 Chr.1: 155913322-155913322 on Build GRCh38
rs947017899 Chr.1: 155912933-155912933 on Build GRCh38
rs913557756 Chr.1: 155913364-155913364 on Build GRCh38
rs987820070 Chr.1: 155913351-155913351 on Build GRCh38
rs575669584 Chr.1: 155912965-155912965 on Build GRCh38
rs754793805 Chr.1: 155912980-155912980 on Build GRCh38
rs905340112 Chr.1: 155912997-155912997 on Build GRCh38
rs1017594337 Chr.1: 155913313-155913313 on Build GRCh38
rs76903692 Chr.1: 155913384-155913384 on Build GRCh38
rs988660459 Chr.1: 155913387-155913387 on Build GRCh38
rs568161866 Chr.1: 155913061-155913061 on Build GRCh38
rs993635679 Chr.1: 155913029-155913030 on Build GRCh38
rs375049161 Chr.1: 155913320-155913320 on Build GRCh38
rs534301604 Chr.1: 155913012-155913012 on Build GRCh38
rs887037568 Chr.1: 155913170-155913170 on Build GRCh38

View Full Product Details