Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 158192541-158193039 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GTF2H5
Entrez Gene ID
404672
Gene Name
general transcription factor IIH subunit 5
Gene Aliases
C6orf175, TFB5, TFIIH, TGF2H5, TTD, TTD-A, TTD3, TTDA, bA120J8.2
UniGene
Hs.356224
Gene Chromosome Location
Chr.6: 158168316-158199344 on Build GRCh38
Transcripts
NM_207118.2 XM_017010862.1

SNP Detail

SNP ID Chromosome Location
rs912657971 Chr.6: 158192937-158192937 on Build GRCh38
rs114283221 Chr.6: 158192751-158192751 on Build GRCh38
rs553727575 Chr.6: 158192632-158192632 on Build GRCh38
rs954568311 Chr.6: 158192924-158192924 on Build GRCh38
rs1055306261 Chr.6: 158192644-158192644 on Build GRCh38
rs779363088 Chr.6: 158192906-158192906 on Build GRCh38
rs1011646119 Chr.6: 158192686-158192686 on Build GRCh38
rs907430256 Chr.6: 158192592-158192592 on Build GRCh38
rs557693022 Chr.6: 158192769-158192769 on Build GRCh38
rs553573764 Chr.6: 158192740-158192740 on Build GRCh38
rs4425618 Chr.6: 158192571-158192571 on Build GRCh38
rs1053555849 Chr.6: 158192993-158192993 on Build GRCh38
rs983546459 Chr.6: 158192785-158192785 on Build GRCh38
rs185453197 Chr.6: 158192600-158192600 on Build GRCh38
rs964544778 Chr.6: 158192748-158192748 on Build GRCh38
rs148725344 Chr.6: 158192948-158192948 on Build GRCh38
rs1011705834 Chr.6: 158193001-158193001 on Build GRCh38
rs142282567 Chr.6: 158192967-158192967 on Build GRCh38
rs544915277 Chr.6: 158192801-158192801 on Build GRCh38
rs575947789 Chr.6: 158192780-158192780 on Build GRCh38
rs190258294 Chr.6: 158192684-158192684 on Build GRCh38
rs545593901 Chr.6: 158192759-158192759 on Build GRCh38
rs1028779934 Chr.6: 158192848-158192848 on Build GRCh38
rs941810493 Chr.6: 158192809-158192809 on Build GRCh38
rs531642881 Chr.6: 158192997-158192997 on Build GRCh38
rs145493528 Chr.6: 158192633-158192633 on Build GRCh38
rs542469058 Chr.6: 158192897-158192897 on Build GRCh38
rs567913573 Chr.6: 158192708-158192708 on Build GRCh38
rs956341287 Chr.6: 158192679-158192679 on Build GRCh38
rs144279837 Chr.6: 158192823-158192823 on Build GRCh38
rs963634939 Chr.6: 158192831-158192831 on Build GRCh38
rs886079118 Chr.6: 158192754-158192754 on Build GRCh38
rs945371638 Chr.6: 158192944-158192944 on Build GRCh38
rs750009596 Chr.6: 158192905-158192906 on Build GRCh38
rs527663296 Chr.6: 158192567-158192567 on Build GRCh38
rs561888186 Chr.6: 158192962-158192970 on Build GRCh38
rs986832483 Chr.6: 158192931-158192931 on Build GRCh38
rs1042614750 Chr.6: 158192584-158192584 on Build GRCh38
rs916290691 Chr.6: 158192872-158192872 on Build GRCh38
rs867616208 Chr.6: 158192768-158192768 on Build GRCh38
rs560541924 Chr.6: 158192936-158192936 on Build GRCh38
rs974665842 Chr.6: 158192847-158192847 on Build GRCh38
rs9457163 Chr.6: 158192878-158192878 on Build GRCh38
rs907522382 Chr.6: 158192803-158192803 on Build GRCh38
rs945396747 Chr.6: 158192583-158192583 on Build GRCh38
rs895348549 Chr.6: 158192720-158192720 on Build GRCh38
rs535308712 Chr.6: 158192727-158192727 on Build GRCh38
rs542220539 Chr.6: 158192954-158192954 on Build GRCh38
rs765393457 Chr.6: 158192693-158192693 on Build GRCh38
rs10625768 Chr.6: 158192914-158192915 on Build GRCh38

View Full Product Details