Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 727555-728090 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAAF5
Entrez Gene ID
54919
Gene Name
dynein axonemal assembly factor 5
Gene Aliases
CILD18, HEATR2
UniGene
Hs.535896
Gene Chromosome Location
Chr.7: 726701-786479 on Build GRCh38
Transcripts
NM_017802.3 NR_075098.1 NM_001164758.1 NM_001164759.1

Gene Symbol
PRKAR1B
Entrez Gene ID
5575
Gene Name
protein kinase cAMP-dependent type I regulatory subunit beta
Gene Aliases
PRKAR1
UniGene
Hs.520851
Gene Chromosome Location
Chr.7: 549185-727676 on Build GRCh38
Transcripts
NM_017802.3 NR_075098.1 NM_001164758.1 NM_001164759.1

SNP Detail

SNP ID Chromosome Location
rs989413566 Chr.7: 728062-728071 on Build GRCh38
rs978618357 Chr.7: 728043-728043 on Build GRCh38
rs185688276 Chr.7: 727808-727808 on Build GRCh38
rs1041140286 Chr.7: 727741-727741 on Build GRCh38
rs948234667 Chr.7: 727900-727900 on Build GRCh38
rs905621386 Chr.7: 727913-727913 on Build GRCh38
rs546318847 Chr.7: 727740-727740 on Build GRCh38
rs373622656 Chr.7: 727901-727901 on Build GRCh38
rs1026060782 Chr.7: 727982-727982 on Build GRCh38
rs1022742585 Chr.7: 727625-727625 on Build GRCh38
rs1042564544 Chr.7: 727881-727881 on Build GRCh38
rs963194969 Chr.7: 727768-727768 on Build GRCh38
rs981411966 Chr.7: 727694-727694 on Build GRCh38
rs771834298 Chr.7: 728048-728048 on Build GRCh38
rs987832566 Chr.7: 727811-727811 on Build GRCh38
rs964706129 Chr.7: 727836-727836 on Build GRCh38
rs150764929 Chr.7: 727689-727689 on Build GRCh38
rs1002970211 Chr.7: 727643-727643 on Build GRCh38
rs915205273 Chr.7: 728021-728021 on Build GRCh38
rs1008713136 Chr.7: 727991-727991 on Build GRCh38
rs1022653968 Chr.7: 727635-727635 on Build GRCh38
rs373402051 Chr.7: 727916-727917 on Build GRCh38
rs78843602 Chr.7: 727621-727621 on Build GRCh38
rs969571364 Chr.7: 727691-727691 on Build GRCh38
rs925575761 Chr.7: 727696-727696 on Build GRCh38
rs13238725 Chr.7: 727678-727678 on Build GRCh38
rs963098841 Chr.7: 727764-727764 on Build GRCh38
rs913931558 Chr.7: 727708-727708 on Build GRCh38
rs950699630 Chr.7: 727664-727664 on Build GRCh38
rs958249393 Chr.7: 727701-727701 on Build GRCh38
rs1016144651 Chr.7: 727677-727677 on Build GRCh38
rs566229013 Chr.7: 727774-727774 on Build GRCh38
rs902774168 Chr.7: 727596-727596 on Build GRCh38
rs1004166428 Chr.7: 727601-727601 on Build GRCh38
rs944118754 Chr.7: 727715-727715 on Build GRCh38
rs1026532320 Chr.7: 727797-727797 on Build GRCh38
rs537764929 Chr.7: 727899-727899 on Build GRCh38
rs939536322 Chr.7: 728049-728049 on Build GRCh38
rs868387831 Chr.7: 727763-727763 on Build GRCh38
rs974692117 Chr.7: 727843-727843 on Build GRCh38
rs376428945 Chr.7: 727789-727789 on Build GRCh38
rs569002944 Chr.7: 727897-727897 on Build GRCh38
rs1018836246 Chr.7: 727995-727995 on Build GRCh38
rs577980695 Chr.7: 727958-727958 on Build GRCh38
rs190923136 Chr.7: 727917-727917 on Build GRCh38
rs915513546 Chr.7: 728066-728066 on Build GRCh38
rs994955438 Chr.7: 727965-727965 on Build GRCh38
rs950642133 Chr.7: 727990-727990 on Build GRCh38
rs538688992 Chr.7: 728023-728023 on Build GRCh38
rs532928032 Chr.7: 727697-727697 on Build GRCh38
rs924146035 Chr.7: 727891-727891 on Build GRCh38
rs956721476 Chr.7: 728052-728052 on Build GRCh38
rs911805364 Chr.7: 727831-727831 on Build GRCh38
rs1005237029 Chr.7: 727969-727969 on Build GRCh38
rs974724600 Chr.7: 727999-727999 on Build GRCh38
rs905589201 Chr.7: 727641-727641 on Build GRCh38
rs1057149706 Chr.7: 727896-727896 on Build GRCh38
rs1012573498 Chr.7: 727624-727624 on Build GRCh38
rs778578134 Chr.7: 727980-727980 on Build GRCh38
rs1035322190 Chr.7: 727645-727645 on Build GRCh38
rs4724904 Chr.7: 727850-727850 on Build GRCh38
rs571123282 Chr.7: 727904-727905 on Build GRCh38
rs939529246 Chr.7: 727892-727892 on Build GRCh38
rs964570923 Chr.7: 727998-727998 on Build GRCh38
rs994692075 Chr.7: 727770-727770 on Build GRCh38
rs922030448 Chr.7: 727776-727776 on Build GRCh38
rs1057234100 Chr.7: 727935-727935 on Build GRCh38
rs1025137320 Chr.7: 727687-727687 on Build GRCh38
rs988162548 Chr.7: 727703-727703 on Build GRCh38
rs1056716297 Chr.7: 727614-727614 on Build GRCh38
rs946769994 Chr.7: 727864-727864 on Build GRCh38
rs1045184931 Chr.7: 727906-727906 on Build GRCh38

View Full Product Details