Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 27155281-27155786 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HOXA-AS3
Entrez Gene ID
100133311
Gene Name
HOXA cluster antisense RNA 3
Gene Aliases
HOXA6as
UniGene
Hs.610216
Gene Chromosome Location
Chr.7: 27140364-27155928 on Build GRCh38
Transcripts
NR_038831.1 NR_038832.1 NM_006896.3

Gene Symbol
HOXA7
Entrez Gene ID
3204
Gene Name
homeobox A7
Gene Aliases
ANTP, HOX1, HOX1.1, HOX1A
UniGene
Hs.610216
Gene Chromosome Location
Chr.7: 27153716-27156677 on Build GRCh38
Transcripts
NR_038831.1 NR_038832.1 NM_006896.3

SNP Detail

SNP ID Chromosome Location
rs767174687 Chr.7: 27155370-27155370 on Build GRCh38
rs997245871 Chr.7: 27155500-27155500 on Build GRCh38
rs1023656268 Chr.7: 27155703-27155703 on Build GRCh38
rs17472091 Chr.7: 27155466-27155466 on Build GRCh38
rs1052828500 Chr.7: 27155666-27155666 on Build GRCh38
rs947007473 Chr.7: 27155554-27155554 on Build GRCh38
rs934330087 Chr.7: 27155653-27155653 on Build GRCh38
rs752472910 Chr.7: 27155415-27155415 on Build GRCh38
rs904265926 Chr.7: 27155397-27155397 on Build GRCh38
rs565707189 Chr.7: 27155577-27155577 on Build GRCh38
rs992982202 Chr.7: 27155555-27155555 on Build GRCh38
rs970352051 Chr.7: 27155711-27155711 on Build GRCh38
rs759131860 Chr.7: 27155331-27155331 on Build GRCh38
rs1027990604 Chr.7: 27155758-27155758 on Build GRCh38
rs890260519 Chr.7: 27155434-27155434 on Build GRCh38
rs530294500 Chr.7: 27155486-27155486 on Build GRCh38
rs949034287 Chr.7: 27155616-27155616 on Build GRCh38
rs900415413 Chr.7: 27155689-27155689 on Build GRCh38
rs550056611 Chr.7: 27155493-27155493 on Build GRCh38
rs112834876 Chr.7: 27155497-27155497 on Build GRCh38
rs552697857 Chr.7: 27155688-27155688 on Build GRCh38
rs994721760 Chr.7: 27155695-27155695 on Build GRCh38
rs1043112889 Chr.7: 27155642-27155642 on Build GRCh38
rs528383842 Chr.7: 27155683-27155683 on Build GRCh38
rs916137665 Chr.7: 27155608-27155608 on Build GRCh38
rs916184110 Chr.7: 27155323-27155323 on Build GRCh38
rs78707165 Chr.7: 27155557-27155557 on Build GRCh38
rs1034947737 Chr.7: 27155352-27155352 on Build GRCh38
rs933678988 Chr.7: 27155536-27155536 on Build GRCh38
rs746609188 Chr.7: 27155664-27155664 on Build GRCh38
rs999577139 Chr.7: 27155343-27155343 on Build GRCh38
rs541713046 Chr.7: 27155330-27155330 on Build GRCh38
rs992699178 Chr.7: 27155357-27155357 on Build GRCh38
rs116870730 Chr.7: 27155550-27155550 on Build GRCh38
rs960695901 Chr.7: 27155353-27155353 on Build GRCh38
rs1052859169 Chr.7: 27155419-27155419 on Build GRCh38
rs984063895 Chr.7: 27155712-27155712 on Build GRCh38
rs948373405 Chr.7: 27155371-27155371 on Build GRCh38
rs1012228072 Chr.7: 27155697-27155697 on Build GRCh38
rs532073461 Chr.7: 27155378-27155378 on Build GRCh38
rs982793295 Chr.7: 27155542-27155542 on Build GRCh38
rs373459845 Chr.7: 27155719-27155719 on Build GRCh38
rs969900326 Chr.7: 27155404-27155404 on Build GRCh38
rs972480543 Chr.7: 27155488-27155488 on Build GRCh38
rs757198248 Chr.7: 27155648-27155648 on Build GRCh38
rs909731037 Chr.7: 27155435-27155435 on Build GRCh38
rs149037629 Chr.7: 27155540-27155540 on Build GRCh38
rs890232623 Chr.7: 27155679-27155679 on Build GRCh38
rs1008722376 Chr.7: 27155680-27155680 on Build GRCh38
rs902695410 Chr.7: 27155615-27155615 on Build GRCh38
rs753836911 Chr.7: 27155633-27155633 on Build GRCh38

View Full Product Details