Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 44207296-44207720 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
YKT6
Entrez Gene ID
10652
Gene Name
YKT6 v-SNARE homolog (S. cerevisiae)
Gene Aliases
-
UniGene
Hs.520794
Gene Chromosome Location
Chr.7: 44200968-44214294 on Build GRCh38
Transcripts
NM_006555.3 XM_005249582.4 XM_017011682.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7398120 c.370G>A p.D124N Chr.7: 44207469-44207469 on Build GRCh38
316620 c.327C>G p.D109E Chr.7: 44207426-44207426 on Build GRCh38
1089929 c.331C>T p.P111S Chr.7: 44207430-44207430 on Build GRCh38
6377803 c.295G>T p.D99Y Chr.7: 44207394-44207394 on Build GRCh38
3431617 c.316G>A p.D106N Chr.7: 44207415-44207415 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs958530865 Chr.7: 44207651-44207651 on Build GRCh38
rs963366243 Chr.7: 44207451-44207451 on Build GRCh38
rs773656627 Chr.7: 44207525-44207525 on Build GRCh38
rs911688948 Chr.7: 44207577-44207577 on Build GRCh38
rs886340279 Chr.7: 44207333-44207333 on Build GRCh38
rs531266934 Chr.7: 44207414-44207414 on Build GRCh38
rs201188604 Chr.7: 44207476-44207476 on Build GRCh38
rs368991261 Chr.7: 44207349-44207349 on Build GRCh38
rs959201807 Chr.7: 44207490-44207490 on Build GRCh38
rs773570225 Chr.7: 44207403-44207403 on Build GRCh38
rs534414096 Chr.7: 44207521-44207521 on Build GRCh38
rs776960825 Chr.7: 44207517-44207517 on Build GRCh38
rs753555118 Chr.7: 44207459-44207459 on Build GRCh38
rs748449947 Chr.7: 44207523-44207523 on Build GRCh38
rs777046711 Chr.7: 44207356-44207356 on Build GRCh38
rs755488653 Chr.7: 44207592-44207592 on Build GRCh38
rs766896881 Chr.7: 44207408-44207408 on Build GRCh38
rs911361350 Chr.7: 44207685-44207685 on Build GRCh38
rs780221434 Chr.7: 44207509-44207509 on Build GRCh38
rs988280811 Chr.7: 44207611-44207611 on Build GRCh38
rs769011365 Chr.7: 44207355-44207355 on Build GRCh38
rs369320488 Chr.7: 44207498-44207498 on Build GRCh38
rs755707761 Chr.7: 44207427-44207427 on Build GRCh38
rs374204381 Chr.7: 44207358-44207358 on Build GRCh38
rs3824065 Chr.7: 44207659-44207659 on Build GRCh38
rs935116941 Chr.7: 44207582-44207582 on Build GRCh38
rs112426773 Chr.7: 44207440-44207440 on Build GRCh38
rs1005359668 Chr.7: 44207327-44207327 on Build GRCh38
rs148034974 Chr.7: 44207441-44207441 on Build GRCh38
rs144641826 Chr.7: 44207466-44207466 on Build GRCh38
rs770004205 Chr.7: 44207524-44207524 on Build GRCh38
rs779427854 Chr.7: 44207606-44207606 on Build GRCh38
rs1047412556 Chr.7: 44207330-44207330 on Build GRCh38
rs761879672 Chr.7: 44207514-44207515 on Build GRCh38
rs184369884 Chr.7: 44207548-44207548 on Build GRCh38
rs1014962983 Chr.7: 44207428-44207428 on Build GRCh38
rs747157899 Chr.7: 44207350-44207350 on Build GRCh38
rs570507763 Chr.7: 44207471-44207471 on Build GRCh38
rs765672936 Chr.7: 44207372-44207372 on Build GRCh38
rs546729927 Chr.7: 44207594-44207594 on Build GRCh38
rs768921588 Chr.7: 44207513-44207513 on Build GRCh38
rs867489664 Chr.7: 44207563-44207563 on Build GRCh38
rs576873431 Chr.7: 44207684-44207684 on Build GRCh38
rs143284758 Chr.7: 44207404-44207404 on Build GRCh38

View Full Product Details