Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 48229684-48230181 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCA13
Entrez Gene ID
154664
Gene Name
ATP binding cassette subfamily A member 13
Gene Aliases
-
UniGene
Hs.226568
Gene Chromosome Location
Chr.7: 48171424-48647497 on Build GRCh38
Transcripts
NM_152701.4 XM_011515130.2 XM_011515131.2 XM_011515132.2 XM_011515133.2 XM_011515134.2 XM_011515136.2 XM_011515137.2 XM_011515138.2 XM_011515139.2 XM_011515141.2 XM_011515142.2 XM_011515143.2 XM_011515144.2 XM_011515145.2 XM_011515146.2 XM_011515147.2 XM_011515148.2 XM_011515149.2 XM_017011767.1 XM_017011768.1 XM_017011769.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3639265 c.521C>T p.S174F Chr.7: 48229878-48229878 on Build GRCh38
5450386 c.681T>G p.T227T Chr.7: 48229873-48229873 on Build GRCh38
1243886 c.633-7G>T p.? Chr.7: 48229818-48229818 on Build GRCh38
4607994 c.523C>A p.Q175K Chr.7: 48229880-48229880 on Build GRCh38
5358500 c.721T>C p.S241P Chr.7: 48229913-48229913 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754523019 Chr.7: 48230132-48230132 on Build GRCh38
rs746062169 Chr.7: 48229951-48229951 on Build GRCh38
rs759794098 Chr.7: 48229862-48229862 on Build GRCh38
rs754992472 Chr.7: 48229786-48229786 on Build GRCh38
rs143409784 Chr.7: 48229983-48229983 on Build GRCh38
rs993259662 Chr.7: 48229731-48229731 on Build GRCh38
rs201583707 Chr.7: 48229866-48229866 on Build GRCh38
rs753345982 Chr.7: 48230084-48230084 on Build GRCh38
rs147152680 Chr.7: 48229998-48229998 on Build GRCh38
rs943294218 Chr.7: 48230022-48230022 on Build GRCh38
rs776172158 Chr.7: 48229870-48229870 on Build GRCh38
rs60553397 Chr.7: 48230082-48230082 on Build GRCh38
rs887535447 Chr.7: 48229762-48229762 on Build GRCh38
rs200641598 Chr.7: 48229873-48229873 on Build GRCh38
rs376234689 Chr.7: 48229993-48229993 on Build GRCh38
rs1033365995 Chr.7: 48229836-48229836 on Build GRCh38
rs553080181 Chr.7: 48230024-48230024 on Build GRCh38
rs772898193 Chr.7: 48229962-48229962 on Build GRCh38
rs928377927 Chr.7: 48229734-48229734 on Build GRCh38
rs774832105 Chr.7: 48229855-48229855 on Build GRCh38
rs751236227 Chr.7: 48229980-48229980 on Build GRCh38
rs762339192 Chr.7: 48229969-48229969 on Build GRCh38
rs767439790 Chr.7: 48229941-48229941 on Build GRCh38
rs1047610797 Chr.7: 48230065-48230065 on Build GRCh38
rs535590945 Chr.7: 48230035-48230035 on Build GRCh38
rs780918542 Chr.7: 48229922-48229922 on Build GRCh38
rs991280259 Chr.7: 48230137-48230137 on Build GRCh38
rs189097947 Chr.7: 48230076-48230076 on Build GRCh38
rs771051125 Chr.7: 48229846-48229846 on Build GRCh38
rs1032490065 Chr.7: 48230149-48230149 on Build GRCh38
rs746766927 Chr.7: 48229775-48229775 on Build GRCh38
rs538538580 Chr.7: 48230115-48230115 on Build GRCh38
rs566926858 Chr.7: 48230025-48230025 on Build GRCh38
rs781079950 Chr.7: 48229788-48229788 on Build GRCh38
rs1041010216 Chr.7: 48229924-48229924 on Build GRCh38
rs761332043 Chr.7: 48229954-48229954 on Build GRCh38
rs369511345 Chr.7: 48229911-48229911 on Build GRCh38
rs751198485 Chr.7: 48229992-48229992 on Build GRCh38
rs375042376 Chr.7: 48229890-48229890 on Build GRCh38
rs773221319 Chr.7: 48229819-48229819 on Build GRCh38
rs750344364 Chr.7: 48230001-48230001 on Build GRCh38
rs978286196 Chr.7: 48229930-48229930 on Build GRCh38
rs771229596 Chr.7: 48229936-48229936 on Build GRCh38
rs199770689 Chr.7: 48229885-48229885 on Build GRCh38
rs1025608003 Chr.7: 48230131-48230131 on Build GRCh38
rs558531694 Chr.7: 48230151-48230151 on Build GRCh38
rs978636502 Chr.7: 48230138-48230138 on Build GRCh38
rs374813284 Chr.7: 48229841-48229841 on Build GRCh38
rs748100869 Chr.7: 48229926-48229926 on Build GRCh38
rs754765359 Chr.7: 48229915-48229915 on Build GRCh38
rs748041785 Chr.7: 48229794-48229794 on Build GRCh38
rs755976001 Chr.7: 48229929-48229929 on Build GRCh38
rs769887352 Chr.7: 48229818-48229818 on Build GRCh38
rs534778852 Chr.7: 48229914-48229914 on Build GRCh38
rs1013833224 Chr.7: 48230122-48230122 on Build GRCh38
rs761213176 Chr.7: 48229880-48229880 on Build GRCh38
rs184391356 Chr.7: 48229754-48229754 on Build GRCh38
rs77288702 Chr.7: 48229772-48229772 on Build GRCh38
rs929098529 Chr.7: 48230036-48230036 on Build GRCh38
rs894846126 Chr.7: 48230112-48230112 on Build GRCh38
rs867087150 Chr.7: 48229878-48229878 on Build GRCh38
rs769275074 Chr.7: 48229955-48229955 on Build GRCh38
rs758129436 Chr.7: 48229904-48229904 on Build GRCh38

View Full Product Details