Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 48239153-48239653 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCA13
Entrez Gene ID
154664
Gene Name
ATP binding cassette subfamily A member 13
Gene Aliases
-
UniGene
Hs.226568
Gene Chromosome Location
Chr.7: 48171424-48647497 on Build GRCh38
Transcripts
NM_152701.4 XM_011515130.2 XM_011515131.2 XM_011515132.2 XM_011515133.2 XM_011515134.2 XM_011515136.2 XM_011515137.2 XM_011515138.2 XM_011515139.2 XM_011515141.2 XM_011515142.2 XM_011515143.2 XM_011515144.2 XM_011515145.2 XM_011515146.2 XM_011515147.2 XM_011515148.2 XM_011515149.2 XM_017011767.1 XM_017011768.1 XM_017011769.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5500422 c.883C>T p.R295* Chr.7: 48239391-48239391 on Build GRCh38
1730565 c.812G>T p.G271V Chr.7: 48239320-48239320 on Build GRCh38
5997181 c.817G>A p.V273I Chr.7: 48239325-48239325 on Build GRCh38
4425280 c.884G>T p.R295L Chr.7: 48239392-48239392 on Build GRCh38
7184989 c.1050A>G p.R350R Chr.7: 48239393-48239393 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771029431 Chr.7: 48239403-48239403 on Build GRCh38
rs773306119 Chr.7: 48239210-48239210 on Build GRCh38
rs754249338 Chr.7: 48239420-48239420 on Build GRCh38
rs184899688 Chr.7: 48239225-48239225 on Build GRCh38
rs77073156 Chr.7: 48239623-48239623 on Build GRCh38
rs746508524 Chr.7: 48239289-48239289 on Build GRCh38
rs988036542 Chr.7: 48239339-48239339 on Build GRCh38
rs777774021 Chr.7: 48239392-48239392 on Build GRCh38
rs571268950 Chr.7: 48239519-48239519 on Build GRCh38
rs898761433 Chr.7: 48239585-48239585 on Build GRCh38
rs751688443 Chr.7: 48239232-48239232 on Build GRCh38
rs761639177 Chr.7: 48239205-48239205 on Build GRCh38
rs769329816 Chr.7: 48239391-48239391 on Build GRCh38
rs759340838 Chr.7: 48239409-48239409 on Build GRCh38
rs767453139 Chr.7: 48239325-48239325 on Build GRCh38
rs568824703 Chr.7: 48239341-48239341 on Build GRCh38
rs886828270 Chr.7: 48239363-48239363 on Build GRCh38
rs565281133 Chr.7: 48239202-48239202 on Build GRCh38
rs755216273 Chr.7: 48239241-48239241 on Build GRCh38
rs756530750 Chr.7: 48239269-48239269 on Build GRCh38
rs746668343 Chr.7: 48239201-48239201 on Build GRCh38
rs768234280 Chr.7: 48239290-48239290 on Build GRCh38
rs995670850 Chr.7: 48239597-48239597 on Build GRCh38
rs191218815 Chr.7: 48239543-48239543 on Build GRCh38
rs776793698 Chr.7: 48239385-48239385 on Build GRCh38
rs1002901742 Chr.7: 48239470-48239470 on Build GRCh38
rs932422641 Chr.7: 48239486-48239486 on Build GRCh38
rs527973661 Chr.7: 48239211-48239211 on Build GRCh38
rs769646194 Chr.7: 48239317-48239317 on Build GRCh38
rs200742821 Chr.7: 48239197-48239197 on Build GRCh38
rs557423069 Chr.7: 48239475-48239475 on Build GRCh38
rs989980736 Chr.7: 48239264-48239264 on Build GRCh38
rs776340044 Chr.7: 48239204-48239204 on Build GRCh38
rs202132236 Chr.7: 48239319-48239319 on Build GRCh38
rs373369519 Chr.7: 48239323-48239323 on Build GRCh38
rs752982701 Chr.7: 48239337-48239337 on Build GRCh38
rs745550291 Chr.7: 48239191-48239191 on Build GRCh38
rs762197437 Chr.7: 48239425-48239425 on Build GRCh38
rs764286598 Chr.7: 48239418-48239418 on Build GRCh38
rs760688539 Chr.7: 48239285-48239285 on Build GRCh38
rs754164235 Chr.7: 48239220-48239221 on Build GRCh38
rs374766244 Chr.7: 48239250-48239250 on Build GRCh38
rs748905254 Chr.7: 48239453-48239453 on Build GRCh38
rs779605323 Chr.7: 48239286-48239286 on Build GRCh38
rs765536978 Chr.7: 48239427-48239427 on Build GRCh38
rs770393544 Chr.7: 48239257-48239257 on Build GRCh38
rs375095559 Chr.7: 48239293-48239293 on Build GRCh38
rs890756594 Chr.7: 48239537-48239537 on Build GRCh38
rs779362400 Chr.7: 48239356-48239356 on Build GRCh38
rs868864507 Chr.7: 48239399-48239400 on Build GRCh38
rs989473035 Chr.7: 48239559-48239559 on Build GRCh38
rs1037257145 Chr.7: 48239558-48239558 on Build GRCh38
rs752137903 Chr.7: 48239437-48239437 on Build GRCh38
rs1025122028 Chr.7: 48239373-48239373 on Build GRCh38
rs773712093 Chr.7: 48239272-48239272 on Build GRCh38
rs377574566 Chr.7: 48239434-48239434 on Build GRCh38
rs772204219 Chr.7: 48239411-48239411 on Build GRCh38
rs747844367 Chr.7: 48239388-48239388 on Build GRCh38
rs764902783 Chr.7: 48239206-48239206 on Build GRCh38
rs761022343 Chr.7: 48239416-48239416 on Build GRCh38
rs748634549 Chr.7: 48239400-48239401 on Build GRCh38
rs780510882 Chr.7: 48239367-48239367 on Build GRCh38
rs766667248 Chr.7: 48239216-48239216 on Build GRCh38
rs759626375 Chr.7: 48239324-48239324 on Build GRCh38
rs749073031 Chr.7: 48239395-48239395 on Build GRCh38
rs537956232 Chr.7: 48239414-48239414 on Build GRCh38
rs1021063956 Chr.7: 48239579-48239579 on Build GRCh38
rs747708470 Chr.7: 48239316-48239316 on Build GRCh38
rs987537179 Chr.7: 48239621-48239624 on Build GRCh38
rs750956255 Chr.7: 48239431-48239431 on Build GRCh38
rs779428329 Chr.7: 48239371-48239382 on Build GRCh38
rs753039235 Chr.7: 48239251-48239251 on Build GRCh38
rs144218923 Chr.7: 48239270-48239270 on Build GRCh38
rs754196404 Chr.7: 48239348-48239348 on Build GRCh38
rs1050744412 Chr.7: 48239504-48239504 on Build GRCh38
rs905950188 Chr.7: 48239629-48239629 on Build GRCh38
rs750854949 Chr.7: 48239363-48239363 on Build GRCh38
rs774921371 Chr.7: 48239198-48239198 on Build GRCh38
rs755883528 Chr.7: 48239439-48239439 on Build GRCh38
rs774456970 Chr.7: 48239404-48239404 on Build GRCh38
rs754541374 Chr.7: 48239366-48239366 on Build GRCh38
rs372915489 Chr.7: 48239271-48239271 on Build GRCh38
rs770982616 Chr.7: 48239322-48239322 on Build GRCh38
rs912389777 Chr.7: 48239278-48239278 on Build GRCh38
rs757818995 Chr.7: 48239351-48239351 on Build GRCh38
rs759726634 Chr.7: 48239479-48239479 on Build GRCh38
rs777532982 Chr.7: 48239445-48239445 on Build GRCh38
rs549984444 Chr.7: 48239320-48239320 on Build GRCh38
rs774832539 Chr.7: 48239257-48239257 on Build GRCh38
rs769029109 Chr.7: 48239208-48239208 on Build GRCh38
rs188702317 Chr.7: 48239533-48239533 on Build GRCh38
rs1016260413 Chr.7: 48239228-48239228 on Build GRCh38

View Full Product Details