Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 55912498-55912956 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF713
Entrez Gene ID
349075
Gene Name
zinc finger protein 713
Gene Aliases
-
UniGene
Hs.660834
Gene Chromosome Location
Chr.7: 55887277-55942225 on Build GRCh38
Transcripts
NM_182633.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1330571 c.10G>T p.E4* Chr.7: 55912685-55912685 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs537915650 Chr.7: 55912603-55912603 on Build GRCh38
rs767145299 Chr.7: 55912727-55912727 on Build GRCh38
rs544276123 Chr.7: 55912856-55912856 on Build GRCh38
rs200710334 Chr.7: 55912710-55912710 on Build GRCh38
rs960077513 Chr.7: 55912700-55912700 on Build GRCh38
rs138200048 Chr.7: 55912904-55912904 on Build GRCh38
rs929751185 Chr.7: 55912851-55912851 on Build GRCh38
rs760020803 Chr.7: 55912741-55912743 on Build GRCh38
rs777014868 Chr.7: 55912702-55912702 on Build GRCh38
rs770344628 Chr.7: 55912634-55912634 on Build GRCh38
rs377250498 Chr.7: 55912726-55912726 on Build GRCh38
rs1016914070 Chr.7: 55912602-55912602 on Build GRCh38
rs979086846 Chr.7: 55912554-55912554 on Build GRCh38
rs1040719683 Chr.7: 55912801-55912801 on Build GRCh38
rs771269090 Chr.7: 55912694-55912695 on Build GRCh38
rs777729446 Chr.7: 55912618-55912618 on Build GRCh38
rs117873186 Chr.7: 55912834-55912834 on Build GRCh38
rs1025342688 Chr.7: 55912534-55912534 on Build GRCh38
rs762280717 Chr.7: 55912709-55912709 on Build GRCh38
rs367619077 Chr.7: 55912685-55912685 on Build GRCh38
rs757133435 Chr.7: 55912775-55912775 on Build GRCh38
rs140984938 Chr.7: 55912688-55912688 on Build GRCh38
rs751006174 Chr.7: 55912597-55912597 on Build GRCh38
rs370561974 Chr.7: 55912541-55912541 on Build GRCh38
rs187720861 Chr.7: 55912522-55912522 on Build GRCh38
rs143004989 Chr.7: 55912711-55912711 on Build GRCh38
rs1005600260 Chr.7: 55912883-55912884 on Build GRCh38
rs752630161 Chr.7: 55912733-55912733 on Build GRCh38
rs570978645 Chr.7: 55912598-55912598 on Build GRCh38
rs748594836 Chr.7: 55912624-55912624 on Build GRCh38
rs540034861 Chr.7: 55912759-55912759 on Build GRCh38
rs911120632 Chr.7: 55912754-55912754 on Build GRCh38
rs758010326 Chr.7: 55912593-55912593 on Build GRCh38
rs1040866704 Chr.7: 55912808-55912808 on Build GRCh38
rs192654523 Chr.7: 55912640-55912640 on Build GRCh38
rs1050259992 Chr.7: 55912902-55912902 on Build GRCh38
rs144400687 Chr.7: 55912675-55912675 on Build GRCh38
rs566543625 Chr.7: 55912933-55912933 on Build GRCh38
rs773449513 Chr.7: 55912676-55912676 on Build GRCh38
rs567424412 Chr.7: 55912604-55912604 on Build GRCh38
rs552765408 Chr.7: 55912595-55912595 on Build GRCh38
rs536459452 Chr.7: 55912655-55912656 on Build GRCh38
rs775909225 Chr.7: 55912644-55912644 on Build GRCh38
rs927350387 Chr.7: 55912633-55912633 on Build GRCh38
rs112359061 Chr.7: 55912852-55912852 on Build GRCh38
rs372221512 Chr.7: 55912639-55912639 on Build GRCh38
rs1031086766 Chr.7: 55912557-55912557 on Build GRCh38
rs967804206 Chr.7: 55912545-55912545 on Build GRCh38
rs950242015 Chr.7: 55912605-55912605 on Build GRCh38
rs565054397 Chr.7: 55912768-55912768 on Build GRCh38
rs777184653 Chr.7: 55912728-55912729 on Build GRCh38
rs771026384 Chr.7: 55912631-55912631 on Build GRCh38
rs569956871 Chr.7: 55912911-55912911 on Build GRCh38
rs112464141 Chr.7: 55912696-55912696 on Build GRCh38
rs187116923 Chr.7: 55912746-55912746 on Build GRCh38

View Full Product Details